Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75202731-75203268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A5
Entrez Gene ID
204962
Gene Name
solute carrier family 44 member 5
Gene Aliases
CTL5
UniGene
Hs.654821
Gene Chromosome Location
Chr.1: 75202131-75724261 on Build GRCh38
Transcripts
NM_001130058.1 NM_001320285.1 NM_001320287.1 XM_006710445.3 XM_011540986.2 XM_011540987.2 XM_017000607.1 XM_017000608.1 XM_017000609.1 XM_017000610.1 XM_017000611.1

SNP Detail

SNP ID Chromosome Location
rs34977303 Chr.1: 75202884-75202885 on Build GRCh38
rs1048987874 Chr.1: 75203184-75203184 on Build GRCh38
rs1017380318 Chr.1: 75202895-75202895 on Build GRCh38
rs1045513388 Chr.1: 75202888-75202888 on Build GRCh38
rs912892135 Chr.1: 75203185-75203185 on Build GRCh38
rs750946031 Chr.1: 75203028-75203028 on Build GRCh38
rs751009853 Chr.1: 75202984-75202985 on Build GRCh38
rs770531313 Chr.1: 75202819-75202819 on Build GRCh38
rs374835794 Chr.1: 75203199-75203199 on Build GRCh38
rs865977117 Chr.1: 75202803-75202803 on Build GRCh38
rs756413319 Chr.1: 75203183-75203183 on Build GRCh38
rs12085115 Chr.1: 75202925-75202925 on Build GRCh38
rs937093787 Chr.1: 75203224-75203224 on Build GRCh38
rs906676587 Chr.1: 75202889-75202889 on Build GRCh38
rs61796520 Chr.1: 75202948-75202948 on Build GRCh38
rs142975012 Chr.1: 75203077-75203077 on Build GRCh38
rs926717505 Chr.1: 75203103-75203103 on Build GRCh38
rs1045109810 Chr.1: 75202780-75202781 on Build GRCh38
rs957253406 Chr.1: 75203140-75203140 on Build GRCh38
rs762925989 Chr.1: 75203215-75203215 on Build GRCh38
rs570664060 Chr.1: 75202872-75202875 on Build GRCh38
rs556619738 Chr.1: 75203107-75203107 on Build GRCh38
rs1019049886 Chr.1: 75203082-75203082 on Build GRCh38
rs775274476 Chr.1: 75203194-75203194 on Build GRCh38
rs34895730 Chr.1: 75202900-75202901 on Build GRCh38
rs781513162 Chr.1: 75203000-75203000 on Build GRCh38
rs7514894 Chr.1: 75202847-75202847 on Build GRCh38
rs756816053 Chr.1: 75202993-75202993 on Build GRCh38
rs897737044 Chr.1: 75202807-75202807 on Build GRCh38
rs185419008 Chr.1: 75202789-75202789 on Build GRCh38
rs990081862 Chr.1: 75203142-75203142 on Build GRCh38
rs189780101 Chr.1: 75202915-75202915 on Build GRCh38
rs1056737222 Chr.1: 75203134-75203134 on Build GRCh38
rs559968271 Chr.1: 75203059-75203059 on Build GRCh38
rs369747046 Chr.1: 75203115-75203115 on Build GRCh38
rs368621310 Chr.1: 75202859-75202860 on Build GRCh38
rs571174178 Chr.1: 75203076-75203076 on Build GRCh38
rs527934788 Chr.1: 75202947-75202947 on Build GRCh38
rs996042835 Chr.1: 75203110-75203110 on Build GRCh38
rs868502338 Chr.1: 75202797-75202797 on Build GRCh38
rs1055297622 Chr.1: 75202921-75202921 on Build GRCh38
rs1007242301 Chr.1: 75203240-75203240 on Build GRCh38
rs529733198 Chr.1: 75202858-75202858 on Build GRCh38
rs148500011 Chr.1: 75202989-75202989 on Build GRCh38
rs966266774 Chr.1: 75203088-75203088 on Build GRCh38
rs868073778 Chr.1: 75202897-75202897 on Build GRCh38
rs778229775 Chr.1: 75202998-75203001 on Build GRCh38
rs746527187 Chr.1: 75202799-75202799 on Build GRCh38
rs935912684 Chr.1: 75202790-75202790 on Build GRCh38

View Full Product Details