Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5118673-5119170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCNA
Entrez Gene ID
5111
Gene Name
proliferating cell nuclear antigen
Gene Aliases
ATLD2
UniGene
Hs.147433
Gene Chromosome Location
Chr.20: 5114953-5126622 on Build GRCh38
Transcripts
NM_002592.2 NM_182649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
21734 c.262A>G p.I88V Chr.20: 5118826-5118826 on Build GRCh38
5638371 c.287C>G p.A96G Chr.20: 5118801-5118801 on Build GRCh38
4099503 c.226T>C p.S76P Chr.20: 5118862-5118862 on Build GRCh38
257692 c.267A>C p.T89T Chr.20: 5118821-5118821 on Build GRCh38
5821924 c.244G>T p.A82S Chr.20: 5118844-5118844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897467477 Chr.20: 5119010-5119010 on Build GRCh38
rs771828986 Chr.20: 5118856-5118856 on Build GRCh38
rs755434993 Chr.20: 5118914-5118914 on Build GRCh38
rs112672560 Chr.20: 5118768-5118768 on Build GRCh38
rs760109380 Chr.20: 5118753-5118754 on Build GRCh38
rs1006923552 Chr.20: 5118983-5118983 on Build GRCh38
rs981745353 Chr.20: 5119140-5119140 on Build GRCh38
rs372779725 Chr.20: 5118758-5118758 on Build GRCh38
rs1023187862 Chr.20: 5119077-5119077 on Build GRCh38
rs767379678 Chr.20: 5118714-5118715 on Build GRCh38
rs542727801 Chr.20: 5118958-5118958 on Build GRCh38
rs538333751 Chr.20: 5119145-5119145 on Build GRCh38
rs567693383 Chr.20: 5119129-5119129 on Build GRCh38
rs749862360 Chr.20: 5118913-5118913 on Build GRCh38
rs773793946 Chr.20: 5119062-5119062 on Build GRCh38
rs757516355 Chr.20: 5118898-5118898 on Build GRCh38
rs768225576 Chr.20: 5118833-5118833 on Build GRCh38
rs962726692 Chr.20: 5119006-5119006 on Build GRCh38
rs916261488 Chr.20: 5119107-5119107 on Build GRCh38
rs756472568 Chr.20: 5118905-5118905 on Build GRCh38
rs144755171 Chr.20: 5118860-5118860 on Build GRCh38
rs983290897 Chr.20: 5119057-5119057 on Build GRCh38
rs532302010 Chr.20: 5119007-5119007 on Build GRCh38
rs939048450 Chr.20: 5118800-5118800 on Build GRCh38
rs527652252 Chr.20: 5119080-5119080 on Build GRCh38
rs531543840 Chr.20: 5118977-5118977 on Build GRCh38
rs750213468 Chr.20: 5118724-5118724 on Build GRCh38
rs192462273 Chr.20: 5118967-5118967 on Build GRCh38
rs924600613 Chr.20: 5118749-5118749 on Build GRCh38
rs113843869 Chr.20: 5118767-5118767 on Build GRCh38
rs549488867 Chr.20: 5119003-5119003 on Build GRCh38
rs201144060 Chr.20: 5118877-5118877 on Build GRCh38
rs780224319 Chr.20: 5118906-5118906 on Build GRCh38
rs17349 Chr.20: 5118870-5118870 on Build GRCh38
rs757200275 Chr.20: 5118784-5118784 on Build GRCh38
rs778535236 Chr.20: 5118915-5118915 on Build GRCh38
rs758512349 Chr.20: 5118746-5118746 on Build GRCh38
rs570933681 Chr.20: 5118727-5118728 on Build GRCh38
rs767476611 Chr.20: 5118720-5118720 on Build GRCh38
rs767746056 Chr.20: 5118899-5118899 on Build GRCh38
rs779762293 Chr.20: 5119017-5119017 on Build GRCh38
rs920539962 Chr.20: 5119014-5119014 on Build GRCh38
rs142530566 Chr.20: 5118827-5118827 on Build GRCh38
rs757469185 Chr.20: 5118771-5118771 on Build GRCh38
rs993236559 Chr.20: 5119035-5119035 on Build GRCh38
rs199954221 Chr.20: 5118812-5118812 on Build GRCh38
rs770091696 Chr.20: 5118796-5118796 on Build GRCh38
rs760746456 Chr.20: 5118725-5118725 on Build GRCh38
rs553835879 Chr.20: 5118931-5118931 on Build GRCh38
rs751868988 Chr.20: 5118770-5118770 on Build GRCh38
rs773719325 Chr.20: 5118837-5118837 on Build GRCh38
rs768355488 Chr.20: 5119052-5119052 on Build GRCh38
rs753678672 Chr.20: 5118743-5118743 on Build GRCh38
rs764303854 Chr.20: 5118888-5118888 on Build GRCh38
rs761314717 Chr.20: 5118715-5118715 on Build GRCh38
rs1029747432 Chr.20: 5118928-5118928 on Build GRCh38
rs974449834 Chr.20: 5119011-5119011 on Build GRCh38
rs547274709 Chr.20: 5119046-5119046 on Build GRCh38
rs761623984 Chr.20: 5118853-5118853 on Build GRCh38
rs1012709232 Chr.20: 5119076-5119076 on Build GRCh38
rs776647579 Chr.20: 5118875-5118875 on Build GRCh38
rs760578899 Chr.20: 5118861-5118861 on Build GRCh38
rs939122198 Chr.20: 5119063-5119063 on Build GRCh38
rs752879172 Chr.20: 5118761-5118761 on Build GRCh38
rs25406 Chr.20: 5118990-5118990 on Build GRCh38
rs748908227 Chr.20: 5119048-5119048 on Build GRCh38
rs998514283 Chr.20: 5119096-5119096 on Build GRCh38
rs999668740 Chr.20: 5118927-5118927 on Build GRCh38
rs781573975 Chr.20: 5118777-5118777 on Build GRCh38
rs950651638 Chr.20: 5119050-5119050 on Build GRCh38
rs971375721 Chr.20: 5119126-5119126 on Build GRCh38
rs944278819 Chr.20: 5118984-5118984 on Build GRCh38
rs113838373 Chr.20: 5118934-5118934 on Build GRCh38
rs73602866 Chr.20: 5119124-5119124 on Build GRCh38
rs755958124 Chr.20: 5119000-5119000 on Build GRCh38
rs747684328 Chr.20: 5118918-5118918 on Build GRCh38
rs183918080 Chr.20: 5119071-5119071 on Build GRCh38
rs766442217 Chr.20: 5118739-5118739 on Build GRCh38
rs772420067 Chr.20: 5119103-5119103 on Build GRCh38
rs750600929 Chr.20: 5118900-5118900 on Build GRCh38
rs751564717 Chr.20: 5118896-5118896 on Build GRCh38
rs572032993 Chr.20: 5118949-5118949 on Build GRCh38

View Full Product Details