Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6770028-6770523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2
Entrez Gene ID
650
Gene Name
bone morphogenetic protein 2
Gene Aliases
BDA2, BMP2A
UniGene
Hs.73853
Gene Chromosome Location
Chr.20: 6767700-6780280 on Build GRCh38
Transcripts
NM_001200.3 XM_011529323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7225313 c.109T>G p.S37A Chr.20: 6770235-6770235 on Build GRCh38
1029273 c.25C>T p.L9L Chr.20: 6770151-6770151 on Build GRCh38
3549299 c.222G>A p.V74V Chr.20: 6770348-6770348 on Build GRCh38
1029274 c.72C>T p.L24L Chr.20: 6770198-6770198 on Build GRCh38
1029275 c.101C>T p.A34V Chr.20: 6770227-6770227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008638475 Chr.20: 6770303-6770303 on Build GRCh38
rs913432818 Chr.20: 6770244-6770244 on Build GRCh38
rs549080609 Chr.20: 6770214-6770214 on Build GRCh38
rs760484979 Chr.20: 6770386-6770386 on Build GRCh38
rs995735344 Chr.20: 6770329-6770329 on Build GRCh38
rs770559493 Chr.20: 6770462-6770462 on Build GRCh38
rs1043311715 Chr.20: 6770253-6770253 on Build GRCh38
rs977818261 Chr.20: 6770184-6770184 on Build GRCh38
rs763227368 Chr.20: 6770393-6770393 on Build GRCh38
rs750303403 Chr.20: 6770225-6770225 on Build GRCh38
rs776468417 Chr.20: 6770478-6770478 on Build GRCh38
rs796654883 Chr.20: 6770217-6770217 on Build GRCh38
rs867523846 Chr.20: 6770136-6770136 on Build GRCh38
rs755231068 Chr.20: 6770230-6770230 on Build GRCh38
rs755320693 Chr.20: 6770078-6770078 on Build GRCh38
rs767964852 Chr.20: 6770407-6770407 on Build GRCh38
rs776361118 Chr.20: 6770389-6770389 on Build GRCh38
rs936595058 Chr.20: 6770226-6770226 on Build GRCh38
rs781520906 Chr.20: 6770261-6770261 on Build GRCh38
rs765539328 Chr.20: 6770231-6770231 on Build GRCh38
rs768217398 Chr.20: 6770294-6770294 on Build GRCh38
rs769492265 Chr.20: 6770158-6770158 on Build GRCh38
rs573278139 Chr.20: 6770491-6770491 on Build GRCh38
rs36105541 Chr.20: 6770355-6770355 on Build GRCh38
rs746277910 Chr.20: 6770262-6770262 on Build GRCh38
rs776196167 Chr.20: 6770120-6770120 on Build GRCh38
rs938843062 Chr.20: 6770228-6770228 on Build GRCh38
rs754625577 Chr.20: 6770433-6770433 on Build GRCh38
rs768828191 Chr.20: 6770494-6770494 on Build GRCh38
rs752683518 Chr.20: 6770232-6770232 on Build GRCh38
rs748698168 Chr.20: 6770277-6770277 on Build GRCh38
rs762870450 Chr.20: 6770320-6770320 on Build GRCh38
rs774838556 Chr.20: 6770172-6770172 on Build GRCh38
rs557734542 Chr.20: 6770305-6770305 on Build GRCh38
rs886043711 Chr.20: 6770401-6770401 on Build GRCh38
rs778731057 Chr.20: 6770049-6770049 on Build GRCh38
rs974200174 Chr.20: 6770067-6770067 on Build GRCh38
rs773338105 Chr.20: 6770500-6770500 on Build GRCh38
rs751692947 Chr.20: 6770324-6770324 on Build GRCh38
rs2273073 Chr.20: 6770235-6770235 on Build GRCh38
rs974052081 Chr.20: 6770143-6770143 on Build GRCh38
rs1026933962 Chr.20: 6770354-6770354 on Build GRCh38
rs375798188 Chr.20: 6770409-6770409 on Build GRCh38
rs978953150 Chr.20: 6770445-6770445 on Build GRCh38
rs752982363 Chr.20: 6770306-6770306 on Build GRCh38
rs1027083347 Chr.20: 6770342-6770342 on Build GRCh38
rs960390396 Chr.20: 6770396-6770396 on Build GRCh38
rs747291949 Chr.20: 6770105-6770105 on Build GRCh38
rs949067053 Chr.20: 6770155-6770155 on Build GRCh38
rs955179163 Chr.20: 6770061-6770061 on Build GRCh38
rs773801167 Chr.20: 6770298-6770298 on Build GRCh38
rs764226242 Chr.20: 6770321-6770321 on Build GRCh38
rs762563014 Chr.20: 6770195-6770195 on Build GRCh38
rs539460039 Chr.20: 6770405-6770405 on Build GRCh38
rs778847394 Chr.20: 6770083-6770083 on Build GRCh38
rs779964077 Chr.20: 6770406-6770406 on Build GRCh38
rs373345005 Chr.20: 6770397-6770397 on Build GRCh38
rs766616005 Chr.20: 6770233-6770235 on Build GRCh38
rs372058568 Chr.20: 6770100-6770100 on Build GRCh38
rs757737368 Chr.20: 6770325-6770325 on Build GRCh38
rs769299110 Chr.20: 6770267-6770267 on Build GRCh38
rs747516210 Chr.20: 6770256-6770256 on Build GRCh38
rs369758681 Chr.20: 6770092-6770092 on Build GRCh38
rs781692600 Chr.20: 6770331-6770331 on Build GRCh38
rs761824743 Chr.20: 6770497-6770497 on Build GRCh38
rs761634402 Chr.20: 6770213-6770213 on Build GRCh38
rs773113395 Chr.20: 6770304-6770304 on Build GRCh38
rs751609144 Chr.20: 6770402-6770402 on Build GRCh38
rs746966399 Chr.20: 6770463-6770463 on Build GRCh38
rs759291983 Chr.20: 6770492-6770492 on Build GRCh38
rs377506952 Chr.20: 6770345-6770345 on Build GRCh38
rs2273074 Chr.20: 6770442-6770442 on Build GRCh38
rs767580200 Chr.20: 6770498-6770498 on Build GRCh38
rs779506433 Chr.20: 6770343-6770343 on Build GRCh38
rs961640875 Chr.20: 6770050-6770050 on Build GRCh38
rs757825179 Chr.20: 6770259-6770259 on Build GRCh38
rs761000959 Chr.20: 6770225-6770226 on Build GRCh38
rs754203178 Chr.20: 6770430-6770430 on Build GRCh38
rs754254056 Chr.20: 6770481-6770481 on Build GRCh38
rs368974118 Chr.20: 6770467-6770467 on Build GRCh38
rs959542332 Chr.20: 6770377-6770377 on Build GRCh38
rs750747484 Chr.20: 6770333-6770333 on Build GRCh38
rs959094654 Chr.20: 6770082-6770082 on Build GRCh38
rs980119171 Chr.20: 6770369-6770369 on Build GRCh38
rs763897233 Chr.20: 6770199-6770199 on Build GRCh38
rs369386883 Chr.20: 6770379-6770379 on Build GRCh38
rs1049007 Chr.20: 6770387-6770387 on Build GRCh38
rs1027134192 Chr.20: 6770068-6770068 on Build GRCh38
rs151071707 Chr.20: 6770356-6770356 on Build GRCh38
rs970071863 Chr.20: 6770186-6770186 on Build GRCh38
rs527356831 Chr.20: 6770140-6770140 on Build GRCh38
rs536048307 Chr.20: 6770240-6770240 on Build GRCh38
rs374592377 Chr.20: 6770449-6770449 on Build GRCh38
rs758803405 Chr.20: 6770098-6770098 on Build GRCh38
rs774359995 Chr.20: 6770203-6770203 on Build GRCh38
rs919924297 Chr.20: 6770150-6770150 on Build GRCh38
rs947607982 Chr.20: 6770437-6770437 on Build GRCh38
rs566450258 Chr.20: 6770107-6770107 on Build GRCh38
rs961599421 Chr.20: 6770142-6770142 on Build GRCh38
rs771718188 Chr.20: 6770452-6770452 on Build GRCh38
rs775238777 Chr.20: 6770276-6770276 on Build GRCh38
rs890872727 Chr.20: 6770293-6770293 on Build GRCh38
rs778080614 Chr.20: 6770249-6770249 on Build GRCh38
rs780452000 Chr.20: 6770428-6770428 on Build GRCh38
rs747698802 Chr.20: 6770370-6770370 on Build GRCh38
rs756645581 Chr.20: 6770418-6770418 on Build GRCh38
rs778541895 Chr.20: 6770363-6770363 on Build GRCh38
rs748311128 Chr.20: 6770085-6770085 on Build GRCh38
rs147880089 Chr.20: 6770301-6770301 on Build GRCh38
rs532833871 Chr.20: 6770048-6770048 on Build GRCh38

View Full Product Details