Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32193919-32194418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAGL2
Entrez Gene ID
5326
Gene Name
PLAG1 like zinc finger 2
Gene Aliases
ZNF900
UniGene
Hs.154104
Gene Chromosome Location
Chr.20: 32192504-32207743 on Build GRCh38
Transcripts
NM_002657.3 XM_005260436.3 XM_011528863.2 XM_011528864.2

SNP Detail

SNP ID Chromosome Location
rs577547676 Chr.20: 32194133-32194133 on Build GRCh38
rs377702503 Chr.20: 32194097-32194097 on Build GRCh38
rs921626521 Chr.20: 32194179-32194179 on Build GRCh38
rs115375105 Chr.20: 32194360-32194360 on Build GRCh38
rs1003734389 Chr.20: 32194019-32194019 on Build GRCh38
rs537745819 Chr.20: 32194093-32194093 on Build GRCh38
rs536814653 Chr.20: 32194048-32194048 on Build GRCh38
rs748108155 Chr.20: 32194309-32194309 on Build GRCh38
rs768241932 Chr.20: 32194265-32194265 on Build GRCh38
rs771253263 Chr.20: 32193991-32193992 on Build GRCh38
rs895149965 Chr.20: 32194353-32194353 on Build GRCh38
rs1012700058 Chr.20: 32194359-32194359 on Build GRCh38
rs1030824666 Chr.20: 32194112-32194112 on Build GRCh38
rs373107699 Chr.20: 32193959-32193959 on Build GRCh38
rs551235571 Chr.20: 32194079-32194079 on Build GRCh38
rs542375391 Chr.20: 32194394-32194394 on Build GRCh38
rs939620205 Chr.20: 32194314-32194314 on Build GRCh38
rs186544427 Chr.20: 32194005-32194005 on Build GRCh38
rs968155858 Chr.20: 32194080-32194080 on Build GRCh38
rs1056411797 Chr.20: 32194056-32194056 on Build GRCh38
rs975974052 Chr.20: 32194160-32194160 on Build GRCh38
rs6061216 Chr.20: 32194310-32194310 on Build GRCh38
rs908991018 Chr.20: 32194235-32194235 on Build GRCh38
rs984471363 Chr.20: 32194229-32194229 on Build GRCh38
rs776877588 Chr.20: 32194094-32194097 on Build GRCh38
rs773182655 Chr.20: 32194319-32194319 on Build GRCh38
rs1056609732 Chr.20: 32194329-32194329 on Build GRCh38
rs953126382 Chr.20: 32194219-32194219 on Build GRCh38
rs867441502 Chr.20: 32193978-32193978 on Build GRCh38
rs145984555 Chr.20: 32194078-32194078 on Build GRCh38
rs761275818 Chr.20: 32193979-32193979 on Build GRCh38
rs773666193 Chr.20: 32194290-32194290 on Build GRCh38
rs566293376 Chr.20: 32194030-32194030 on Build GRCh38
rs918734645 Chr.20: 32194311-32194311 on Build GRCh38
rs895111543 Chr.20: 32194058-32194058 on Build GRCh38
rs538368034 Chr.20: 32194146-32194146 on Build GRCh38

View Full Product Details