Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32207785-32208213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POFUT1
Entrez Gene ID
23509
Gene Name
protein O-fucosyltransferase 1
Gene Aliases
DDD2, FUT12, O-FUT, O-Fuc-T, O-FucT-1, OFUCT1
UniGene
Hs.472409
Gene Chromosome Location
Chr.20: 32207822-32238664 on Build GRCh38
Transcripts
NM_015352.1 NM_172236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4716343 c.1A>G p.M1V Chr.20: 32207942-32207942 on Build GRCh38
6751728 c.10G>T p.A4S Chr.20: 32207951-32207951 on Build GRCh38
6751724 c.97G>A p.G33S Chr.20: 32208038-32208038 on Build GRCh38
6751729 c.10G>T p.A4S Chr.20: 32207951-32207951 on Build GRCh38
5525448 c.23G>A p.R8Q Chr.20: 32207964-32207964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752049500 Chr.20: 32207891-32207891 on Build GRCh38
rs935223095 Chr.20: 32207909-32207909 on Build GRCh38
rs143528740 Chr.20: 32207918-32207918 on Build GRCh38
rs6087834 Chr.20: 32207938-32207938 on Build GRCh38
rs752465904 Chr.20: 32208055-32208055 on Build GRCh38
rs1031681243 Chr.20: 32207847-32207847 on Build GRCh38
rs182210334 Chr.20: 32208148-32208148 on Build GRCh38
rs756261411 Chr.20: 32207903-32207903 on Build GRCh38
rs997189168 Chr.20: 32207872-32207875 on Build GRCh38
rs1042889008 Chr.20: 32208021-32208021 on Build GRCh38
rs1012742821 Chr.20: 32207807-32207807 on Build GRCh38
rs1024162743 Chr.20: 32208164-32208164 on Build GRCh38
rs998786783 Chr.20: 32208082-32208082 on Build GRCh38
rs746499058 Chr.20: 32207971-32207971 on Build GRCh38
rs954673159 Chr.20: 32207848-32207848 on Build GRCh38
rs573002705 Chr.20: 32207974-32207974 on Build GRCh38
rs867609748 Chr.20: 32208056-32208056 on Build GRCh38
rs765055989 Chr.20: 32208042-32208042 on Build GRCh38
rs979690388 Chr.20: 32208179-32208179 on Build GRCh38
rs1031984829 Chr.20: 32208109-32208109 on Build GRCh38
rs1021985241 Chr.20: 32207816-32207816 on Build GRCh38
rs749309004 Chr.20: 32207951-32207951 on Build GRCh38
rs199621681 Chr.20: 32207936-32207936 on Build GRCh38
rs772695718 Chr.20: 32208006-32208006 on Build GRCh38
rs1007043675 Chr.20: 32207880-32207880 on Build GRCh38
rs935022010 Chr.20: 32207808-32207808 on Build GRCh38
rs370777228 Chr.20: 32207948-32207948 on Build GRCh38
rs1017626356 Chr.20: 32207881-32207881 on Build GRCh38
rs962929678 Chr.20: 32207882-32207882 on Build GRCh38
rs752658183 Chr.20: 32207961-32207961 on Build GRCh38
rs566580075 Chr.20: 32207875-32207875 on Build GRCh38
rs779950045 Chr.20: 32207910-32207910 on Build GRCh38
rs766503770 Chr.20: 32207949-32207949 on Build GRCh38
rs890471928 Chr.20: 32208111-32208111 on Build GRCh38
rs758385916 Chr.20: 32208115-32208115 on Build GRCh38
rs974012058 Chr.20: 32208167-32208167 on Build GRCh38
rs1051277557 Chr.20: 32207879-32207879 on Build GRCh38
rs987537322 Chr.20: 32207850-32207850 on Build GRCh38
rs758297493 Chr.20: 32207965-32207965 on Build GRCh38
rs778826164 Chr.20: 32207913-32207913 on Build GRCh38
rs746712480 Chr.20: 32207921-32207921 on Build GRCh38
rs894211893 Chr.20: 32208007-32208007 on Build GRCh38
rs773407006 Chr.20: 32208014-32208014 on Build GRCh38
rs754025432 Chr.20: 32208029-32208029 on Build GRCh38
rs777311168 Chr.20: 32207967-32207967 on Build GRCh38
rs1048409950 Chr.20: 32207962-32207962 on Build GRCh38
rs755008610 Chr.20: 32207911-32207911 on Build GRCh38
rs778630208 Chr.20: 32207955-32207955 on Build GRCh38
rs964761005 Chr.20: 32208149-32208149 on Build GRCh38
rs771687810 Chr.20: 32208171-32208171 on Build GRCh38
rs771588181 Chr.20: 32207916-32207916 on Build GRCh38
rs968663229 Chr.20: 32207832-32207832 on Build GRCh38
rs192137481 Chr.20: 32207926-32207926 on Build GRCh38
rs767656405 Chr.20: 32207897-32207897 on Build GRCh38
rs994181416 Chr.20: 32207887-32207887 on Build GRCh38
rs368172687 Chr.20: 32207940-32207940 on Build GRCh38
rs1052030104 Chr.20: 32207809-32207809 on Build GRCh38
rs575321405 Chr.20: 32207914-32207914 on Build GRCh38
rs757430659 Chr.20: 32207896-32207896 on Build GRCh38
rs368413950 Chr.20: 32208031-32208031 on Build GRCh38
rs750509980 Chr.20: 32207941-32207941 on Build GRCh38
rs1017664451 Chr.20: 32208139-32208139 on Build GRCh38
rs999297692 Chr.20: 32207840-32207840 on Build GRCh38
rs754024771 Chr.20: 32207950-32207950 on Build GRCh38
rs763234398 Chr.20: 32207928-32207928 on Build GRCh38
rs1008842715 Chr.20: 32208112-32208112 on Build GRCh38
rs750706180 Chr.20: 32207901-32207901 on Build GRCh38
rs762460177 Chr.20: 32208001-32208001 on Build GRCh38
rs745479645 Chr.20: 32207987-32207987 on Build GRCh38
rs775025070 Chr.20: 32207939-32207939 on Build GRCh38
rs960783128 Chr.20: 32207893-32207893 on Build GRCh38
rs932991722 Chr.20: 32207917-32207917 on Build GRCh38
rs765960199 Chr.20: 32208074-32208075 on Build GRCh38
rs770466202 Chr.20: 32207925-32207925 on Build GRCh38
rs769174950 Chr.20: 32207996-32207996 on Build GRCh38
rs1923095 Chr.20: 32208016-32208016 on Build GRCh38
rs916663736 Chr.20: 32207900-32207900 on Build GRCh38
rs766420398 Chr.20: 32208026-32208026 on Build GRCh38
rs780725234 Chr.20: 32207980-32207980 on Build GRCh38
rs1013575737 Chr.20: 32208116-32208116 on Build GRCh38
rs555288977 Chr.20: 32208032-32208032 on Build GRCh38
rs979270171 Chr.20: 32207902-32207902 on Build GRCh38
rs901616122 Chr.20: 32207846-32207846 on Build GRCh38
rs556847066 Chr.20: 32207839-32207839 on Build GRCh38

View Full Product Details