Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32230908-32231443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POFUT1
Entrez Gene ID
23509
Gene Name
protein O-fucosyltransferase 1
Gene Aliases
DDD2, FUT12, O-FUT, O-Fuc-T, O-FucT-1, OFUCT1
UniGene
Hs.472409
Gene Chromosome Location
Chr.20: 32207822-32238664 on Build GRCh38
Transcripts
NM_015352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4716345 c.922G>A p.V308I Chr.20: 32231005-32231005 on Build GRCh38
6751726 c.966C>T p.L322L Chr.20: 32231049-32231049 on Build GRCh38
1025702 c.932A>C p.D311A Chr.20: 32231015-32231015 on Build GRCh38
5057227 c.827delC p.L278fs*3 Chr.20: 32230910-32230910 on Build GRCh38
6751731 c.924T>A p.V308V Chr.20: 32231007-32231007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747793346 Chr.20: 32231054-32231054 on Build GRCh38
rs554230483 Chr.20: 32231393-32231393 on Build GRCh38
rs375470215 Chr.20: 32230926-32230926 on Build GRCh38
rs910312704 Chr.20: 32231371-32231371 on Build GRCh38
rs915491528 Chr.20: 32231177-32231177 on Build GRCh38
rs138613217 Chr.20: 32231314-32231314 on Build GRCh38
rs200646120 Chr.20: 32230998-32230998 on Build GRCh38
rs899095160 Chr.20: 32231259-32231259 on Build GRCh38
rs187915686 Chr.20: 32231232-32231232 on Build GRCh38
rs879239430 Chr.20: 32231016-32231016 on Build GRCh38
rs939965292 Chr.20: 32231121-32231121 on Build GRCh38
rs779519269 Chr.20: 32230927-32230927 on Build GRCh38
rs750378130 Chr.20: 32231008-32231008 on Build GRCh38
rs576092663 Chr.20: 32231422-32231422 on Build GRCh38
rs568657961 Chr.20: 32231331-32231331 on Build GRCh38
rs776072239 Chr.20: 32231093-32231093 on Build GRCh38
rs771237065 Chr.20: 32231065-32231065 on Build GRCh38
rs570784433 Chr.20: 32231316-32231316 on Build GRCh38
rs868808215 Chr.20: 32231254-32231254 on Build GRCh38
rs987061545 Chr.20: 32231369-32231369 on Build GRCh38
rs773084506 Chr.20: 32231180-32231180 on Build GRCh38
rs530132437 Chr.20: 32231114-32231114 on Build GRCh38
rs768296195 Chr.20: 32230937-32230937 on Build GRCh38
rs768479011 Chr.20: 32230962-32230962 on Build GRCh38
rs1026762872 Chr.20: 32231272-32231272 on Build GRCh38
rs774075147 Chr.20: 32231063-32231064 on Build GRCh38
rs777031988 Chr.20: 32230958-32230958 on Build GRCh38
rs760964890 Chr.20: 32230945-32230945 on Build GRCh38
rs756035083 Chr.20: 32231010-32231010 on Build GRCh38
rs1046456090 Chr.20: 32231346-32231346 on Build GRCh38
rs8118514 Chr.20: 32231362-32231362 on Build GRCh38
rs147562228 Chr.20: 32231045-32231045 on Build GRCh38
rs938089839 Chr.20: 32231356-32231356 on Build GRCh38
rs117140430 Chr.20: 32231121-32231121 on Build GRCh38
rs369910269 Chr.20: 32230942-32230942 on Build GRCh38
rs191268131 Chr.20: 32231289-32231289 on Build GRCh38
rs768100190 Chr.20: 32231032-32231032 on Build GRCh38
rs760055191 Chr.20: 32230970-32230970 on Build GRCh38
rs967396201 Chr.20: 32231327-32231327 on Build GRCh38
rs945625986 Chr.20: 32231208-32231208 on Build GRCh38
rs374302867 Chr.20: 32231231-32231231 on Build GRCh38
rs746481752 Chr.20: 32231122-32231122 on Build GRCh38
rs888943069 Chr.20: 32230935-32230935 on Build GRCh38
rs768443650 Chr.20: 32231210-32231210 on Build GRCh38
rs150166850 Chr.20: 32231290-32231290 on Build GRCh38
rs532472355 Chr.20: 32231019-32231019 on Build GRCh38
rs893631001 Chr.20: 32231400-32231400 on Build GRCh38
rs927936785 Chr.20: 32231350-32231350 on Build GRCh38
rs565027162 Chr.20: 32231005-32231005 on Build GRCh38
rs921426547 Chr.20: 32231015-32231015 on Build GRCh38
rs1033773169 Chr.20: 32231293-32231293 on Build GRCh38
rs576049320 Chr.20: 32230997-32230997 on Build GRCh38
rs977516721 Chr.20: 32231212-32231212 on Build GRCh38
rs940248495 Chr.20: 32231340-32231341 on Build GRCh38
rs78861748 Chr.20: 32230983-32230983 on Build GRCh38
rs765455438 Chr.20: 32230975-32230975 on Build GRCh38
rs764229773 Chr.20: 32231112-32231112 on Build GRCh38
rs754106797 Chr.20: 32231298-32231298 on Build GRCh38
rs3818219 Chr.20: 32231239-32231239 on Build GRCh38
rs943212258 Chr.20: 32231244-32231244 on Build GRCh38
rs770418536 Chr.20: 32231086-32231086 on Build GRCh38
rs369674226 Chr.20: 32231294-32231294 on Build GRCh38
rs35968884 Chr.20: 32231020-32231020 on Build GRCh38
rs924289794 Chr.20: 32231217-32231217 on Build GRCh38
rs763138301 Chr.20: 32231096-32231096 on Build GRCh38
rs752809888 Chr.20: 32230982-32230982 on Build GRCh38
rs906650878 Chr.20: 32231146-32231146 on Build GRCh38
rs924313083 Chr.20: 32231336-32231336 on Build GRCh38
rs893611719 Chr.20: 32231193-32231193 on Build GRCh38
rs921447605 Chr.20: 32231309-32231309 on Build GRCh38
rs1010704331 Chr.20: 32231403-32231403 on Build GRCh38
rs752799506 Chr.20: 32231354-32231354 on Build GRCh38
rs766024912 Chr.20: 32231269-32231269 on Build GRCh38
rs749000411 Chr.20: 32230928-32230928 on Build GRCh38
rs943096143 Chr.20: 32231394-32231394 on Build GRCh38
rs971244053 Chr.20: 32231026-32231026 on Build GRCh38
rs1033959028 Chr.20: 32231191-32231191 on Build GRCh38
rs563809285 Chr.20: 32231066-32231066 on Build GRCh38
rs17268666 Chr.20: 32231047-32231047 on Build GRCh38
rs746434007 Chr.20: 32231076-32231076 on Build GRCh38
rs140900043 Chr.20: 32230950-32230950 on Build GRCh38
rs755144523 Chr.20: 32231315-32231315 on Build GRCh38

View Full Product Details