Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45843300-45843811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT8
Entrez Gene ID
10005
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
HNAACTE, NAP1, PTE-1, PTE-2, PTE1, PTE2, hACTE-III, hTE
UniGene
Hs.444776
Gene Chromosome Location
Chr.20: 45841721-45857409 on Build GRCh38
Transcripts
NM_005469.3 XM_005260239.3 XM_011528480.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4098873 c.818A>G p.Y273C Chr.20: 45843550-45843550 on Build GRCh38
5822747 c.700G>A p.A234T Chr.20: 45843668-45843668 on Build GRCh38
6592338 c.806A>G p.H269R Chr.20: 45843562-45843562 on Build GRCh38
3972570 c.720G>C p.L240L Chr.20: 45843648-45843648 on Build GRCh38
5553196 c.821A>G p.E274G Chr.20: 45843547-45843547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753359447 Chr.20: 45843676-45843676 on Build GRCh38
rs773702461 Chr.20: 45843576-45843576 on Build GRCh38
rs771368576 Chr.20: 45843738-45843738 on Build GRCh38
rs78931499 Chr.20: 45843354-45843354 on Build GRCh38
rs762380579 Chr.20: 45843571-45843571 on Build GRCh38
rs55931834 Chr.20: 45843524-45843524 on Build GRCh38
rs759885718 Chr.20: 45843769-45843769 on Build GRCh38
rs754586310 Chr.20: 45843399-45843399 on Build GRCh38
rs139975225 Chr.20: 45843675-45843675 on Build GRCh38
rs780820973 Chr.20: 45843403-45843403 on Build GRCh38
rs754676114 Chr.20: 45843521-45843521 on Build GRCh38
rs752269233 Chr.20: 45843527-45843527 on Build GRCh38
rs544115603 Chr.20: 45843543-45843543 on Build GRCh38
rs983974499 Chr.20: 45843355-45843355 on Build GRCh38
rs200396914 Chr.20: 45843670-45843670 on Build GRCh38
rs775259155 Chr.20: 45843674-45843674 on Build GRCh38
rs756689298 Chr.20: 45843682-45843682 on Build GRCh38
rs780341655 Chr.20: 45843626-45843626 on Build GRCh38
rs369757435 Chr.20: 45843669-45843669 on Build GRCh38
rs761330408 Chr.20: 45843779-45843779 on Build GRCh38
rs773688686 Chr.20: 45843783-45843783 on Build GRCh38
rs775505195 Chr.20: 45843562-45843562 on Build GRCh38
rs772205269 Chr.20: 45843503-45843503 on Build GRCh38
rs1039195501 Chr.20: 45843379-45843379 on Build GRCh38
rs759721618 Chr.20: 45843501-45843501 on Build GRCh38
rs139286507 Chr.20: 45843564-45843564 on Build GRCh38
rs775625443 Chr.20: 45843506-45843506 on Build GRCh38
rs913493174 Chr.20: 45843704-45843704 on Build GRCh38
rs372337546 Chr.20: 45843687-45843687 on Build GRCh38
rs554163736 Chr.20: 45843433-45843433 on Build GRCh38
rs1046934308 Chr.20: 45843358-45843358 on Build GRCh38
rs184954994 Chr.20: 45843421-45843421 on Build GRCh38
rs536319264 Chr.20: 45843448-45843448 on Build GRCh38
rs144748462 Chr.20: 45843572-45843572 on Build GRCh38
rs751205424 Chr.20: 45843360-45843360 on Build GRCh38
rs772295216 Chr.20: 45843554-45843554 on Build GRCh38
rs1019330555 Chr.20: 45843418-45843418 on Build GRCh38
rs143094082 Chr.20: 45843568-45843568 on Build GRCh38
rs41282772 Chr.20: 45843513-45843513 on Build GRCh38
rs536167577 Chr.20: 45843742-45843742 on Build GRCh38
rs202141299 Chr.20: 45843581-45843581 on Build GRCh38
rs773878511 Chr.20: 45843719-45843719 on Build GRCh38
rs760614101 Chr.20: 45843512-45843512 on Build GRCh38
rs749132839 Chr.20: 45843542-45843542 on Build GRCh38
rs574809856 Chr.20: 45843578-45843578 on Build GRCh38
rs376653145 Chr.20: 45843693-45843693 on Build GRCh38
rs886991917 Chr.20: 45843582-45843582 on Build GRCh38
rs758146041 Chr.20: 45843623-45843623 on Build GRCh38
rs572458228 Chr.20: 45843468-45843468 on Build GRCh38
rs764700901 Chr.20: 45843782-45843782 on Build GRCh38
rs752252558 Chr.20: 45843580-45843580 on Build GRCh38
rs370026049 Chr.20: 45843528-45843528 on Build GRCh38
rs375281702 Chr.20: 45843353-45843353 on Build GRCh38
rs150578949 Chr.20: 45843647-45843647 on Build GRCh38
rs750307207 Chr.20: 45843686-45843686 on Build GRCh38
rs142815229 Chr.20: 45843607-45843607 on Build GRCh38
rs773792632 Chr.20: 45843661-45843661 on Build GRCh38
rs377650044 Chr.20: 45843727-45843727 on Build GRCh38
rs762288247 Chr.20: 45843518-45843518 on Build GRCh38
rs920614138 Chr.20: 45843344-45843344 on Build GRCh38
rs762794020 Chr.20: 45843784-45843784 on Build GRCh38
rs770441032 Chr.20: 45843651-45843651 on Build GRCh38
rs769955280 Chr.20: 45843717-45843717 on Build GRCh38
rs148501184 Chr.20: 45843574-45843574 on Build GRCh38
rs886792316 Chr.20: 45843359-45843359 on Build GRCh38
rs774580087 Chr.20: 45843481-45843481 on Build GRCh38
rs746146702 Chr.20: 45843550-45843550 on Build GRCh38
rs747173631 Chr.20: 45843627-45843627 on Build GRCh38
rs779004029 Chr.20: 45843407-45843407 on Build GRCh38
rs76000398 Chr.20: 45843752-45843752 on Build GRCh38
rs774564527 Chr.20: 45843741-45843741 on Build GRCh38
rs747872501 Chr.20: 45843482-45843482 on Build GRCh38
rs778763267 Chr.20: 45843619-45843619 on Build GRCh38
rs138190275 Chr.20: 45843361-45843361 on Build GRCh38
rs368271642 Chr.20: 45843479-45843479 on Build GRCh38
rs772146726 Chr.20: 45843475-45843475 on Build GRCh38
rs755144727 Chr.20: 45843695-45843695 on Build GRCh38
rs200606705 Chr.20: 45843720-45843720 on Build GRCh38
rs763492655 Chr.20: 45843592-45843592 on Build GRCh38
rs1003324229 Chr.20: 45843749-45843749 on Build GRCh38
rs757171467 Chr.20: 45843609-45843609 on Build GRCh38
rs748247124 Chr.20: 45843713-45843713 on Build GRCh38
rs766256775 Chr.20: 45843785-45843785 on Build GRCh38
rs768197600 Chr.20: 45843773-45843773 on Build GRCh38
rs374190319 Chr.20: 45843447-45843447 on Build GRCh38
rs768567394 Chr.20: 45843567-45843567 on Build GRCh38
rs765672228 Chr.20: 45843520-45843520 on Build GRCh38
rs745667158 Chr.20: 45843620-45843620 on Build GRCh38
rs748725572 Chr.20: 45843649-45843649 on Build GRCh38
rs768818481 Chr.20: 45843634-45843634 on Build GRCh38
rs746078591 Chr.20: 45843432-45843432 on Build GRCh38
rs968894175 Chr.20: 45843710-45843710 on Build GRCh38
rs776226725 Chr.20: 45843778-45843778 on Build GRCh38
rs779680625 Chr.20: 45843692-45843692 on Build GRCh38
rs562671523 Chr.20: 45843545-45843545 on Build GRCh38
rs777798412 Chr.20: 45843539-45843539 on Build GRCh38
rs1009312067 Chr.20: 45843492-45843492 on Build GRCh38
rs754256237 Chr.20: 45843515-45843515 on Build GRCh38
rs369064144 Chr.20: 45843699-45843699 on Build GRCh38

View Full Product Details