Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62394775-62395307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABLES2
Entrez Gene ID
81928
Gene Name
Cdk5 and Abl enzyme substrate 2
Gene Aliases
C20orf150, dJ908M14.2, ik3-2
UniGene
Hs.301040
Gene Chromosome Location
Chr.20: 62388630-62407283 on Build GRCh38
Transcripts
NM_031215.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7331179 c.584A>G p.Y195C Chr.20: 62394958-62394958 on Build GRCh38
6425295 c.574G>T p.V192F Chr.20: 62394968-62394968 on Build GRCh38
6617209 c.599G>A p.R200Q Chr.20: 62394943-62394943 on Build GRCh38
1028894 c.550C>T p.R184W Chr.20: 62394992-62394992 on Build GRCh38
2151918 c.605+4A>G p.? Chr.20: 62394933-62394933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149555287 Chr.20: 62395205-62395205 on Build GRCh38
rs199898744 Chr.20: 62395041-62395041 on Build GRCh38
rs375959062 Chr.20: 62394914-62394914 on Build GRCh38
rs78353357 Chr.20: 62394871-62394871 on Build GRCh38
rs1017115330 Chr.20: 62395033-62395034 on Build GRCh38
rs202210166 Chr.20: 62395048-62395048 on Build GRCh38
rs113051244 Chr.20: 62395058-62395058 on Build GRCh38
rs780455800 Chr.20: 62395050-62395050 on Build GRCh38
rs114213331 Chr.20: 62395083-62395083 on Build GRCh38
rs369778548 Chr.20: 62394923-62394923 on Build GRCh38
rs906416033 Chr.20: 62395098-62395098 on Build GRCh38
rs774682016 Chr.20: 62394909-62394909 on Build GRCh38
rs761106683 Chr.20: 62394932-62394932 on Build GRCh38
rs146261288 Chr.20: 62395055-62395055 on Build GRCh38
rs893620848 Chr.20: 62395156-62395156 on Build GRCh38
rs538594542 Chr.20: 62395059-62395059 on Build GRCh38
rs776055207 Chr.20: 62394939-62394939 on Build GRCh38
rs190847259 Chr.20: 62395121-62395121 on Build GRCh38
rs533957370 Chr.20: 62394820-62394820 on Build GRCh38
rs375112262 Chr.20: 62394944-62394944 on Build GRCh38
rs550054984 Chr.20: 62395057-62395057 on Build GRCh38
rs993444574 Chr.20: 62395204-62395204 on Build GRCh38
rs769446768 Chr.20: 62395008-62395008 on Build GRCh38
rs573853105 Chr.20: 62395230-62395230 on Build GRCh38
rs965194314 Chr.20: 62395200-62395200 on Build GRCh38
rs1021653930 Chr.20: 62395174-62395174 on Build GRCh38
rs373211561 Chr.20: 62395027-62395027 on Build GRCh38
rs544403047 Chr.20: 62395249-62395249 on Build GRCh38
rs373923225 Chr.20: 62394943-62394943 on Build GRCh38
rs1028015028 Chr.20: 62395239-62395239 on Build GRCh38
rs756173252 Chr.20: 62394898-62394898 on Build GRCh38
rs762727571 Chr.20: 62395019-62395019 on Build GRCh38
rs370153452 Chr.20: 62395047-62395047 on Build GRCh38
rs369586037 Chr.20: 62394949-62394949 on Build GRCh38
rs2427311 Chr.20: 62395067-62395067 on Build GRCh38
rs920222915 Chr.20: 62394838-62394838 on Build GRCh38
rs761161808 Chr.20: 62395007-62395007 on Build GRCh38
rs1050447299 Chr.20: 62395134-62395134 on Build GRCh38
rs373241073 Chr.20: 62394991-62394991 on Build GRCh38
rs556408555 Chr.20: 62395250-62395250 on Build GRCh38
rs746103333 Chr.20: 62394983-62394983 on Build GRCh38
rs150535079 Chr.20: 62394958-62394958 on Build GRCh38
rs386816019 Chr.20: 62394871-62394873 on Build GRCh38
rs200844164 Chr.20: 62395054-62395054 on Build GRCh38
rs1046489608 Chr.20: 62395086-62395086 on Build GRCh38
rs780074393 Chr.20: 62394893-62394893 on Build GRCh38
rs889239166 Chr.20: 62395152-62395152 on Build GRCh38
rs996160141 Chr.20: 62395209-62395209 on Build GRCh38
rs893774924 Chr.20: 62394895-62394895 on Build GRCh38
rs758213967 Chr.20: 62394955-62394955 on Build GRCh38
rs772954205 Chr.20: 62395010-62395010 on Build GRCh38
rs2427310 Chr.20: 62394873-62394873 on Build GRCh38
rs149234266 Chr.20: 62394978-62394978 on Build GRCh38
rs940586011 Chr.20: 62394824-62394824 on Build GRCh38
rs970573987 Chr.20: 62394812-62394812 on Build GRCh38
rs906826656 Chr.20: 62394877-62394877 on Build GRCh38
rs1030763534 Chr.20: 62395219-62395219 on Build GRCh38
rs561928086 Chr.20: 62394957-62394957 on Build GRCh38
rs978363738 Chr.20: 62394819-62394819 on Build GRCh38
rs751356232 Chr.20: 62395025-62395025 on Build GRCh38
rs951992744 Chr.20: 62395240-62395240 on Build GRCh38
rs930414780 Chr.20: 62394841-62394841 on Build GRCh38
rs776201668 Chr.20: 62395005-62395005 on Build GRCh38
rs749493749 Chr.20: 62394903-62394903 on Build GRCh38
rs138669550 Chr.20: 62394969-62394969 on Build GRCh38
rs375897717 Chr.20: 62395022-62395022 on Build GRCh38
rs777748745 Chr.20: 62394901-62394901 on Build GRCh38
rs186502881 Chr.20: 62395026-62395026 on Build GRCh38
rs999862116 Chr.20: 62395107-62395107 on Build GRCh38
rs1052597369 Chr.20: 62395271-62395271 on Build GRCh38
rs999696692 Chr.20: 62394882-62394882 on Build GRCh38
rs763041045 Chr.20: 62395037-62395038 on Build GRCh38
rs941774819 Chr.20: 62394879-62394879 on Build GRCh38
rs981132822 Chr.20: 62395263-62395263 on Build GRCh38
rs906483351 Chr.20: 62394941-62394941 on Build GRCh38
rs771104317 Chr.20: 62394908-62394908 on Build GRCh38
rs372865056 Chr.20: 62394897-62394897 on Build GRCh38
rs547933927 Chr.20: 62395042-62395042 on Build GRCh38
rs1010721714 Chr.20: 62395163-62395163 on Build GRCh38
rs763631148 Chr.20: 62394936-62394936 on Build GRCh38
rs2427309 Chr.20: 62394830-62394830 on Build GRCh38
rs578197176 Chr.20: 62395270-62395270 on Build GRCh38
rs369094971 Chr.20: 62394887-62394887 on Build GRCh38
rs757461175 Chr.20: 62394979-62394979 on Build GRCh38
rs182142205 Chr.20: 62394915-62394915 on Build GRCh38
rs538259962 Chr.20: 62394874-62394874 on Build GRCh38
rs908154055 Chr.20: 62395243-62395243 on Build GRCh38
rs567502952 Chr.20: 62394850-62394850 on Build GRCh38
rs971121730 Chr.20: 62395260-62395260 on Build GRCh38
rs914587282 Chr.20: 62394845-62394845 on Build GRCh38
rs141752965 Chr.20: 62394980-62394980 on Build GRCh38
rs765237042 Chr.20: 62394852-62394852 on Build GRCh38
rs1008953154 Chr.20: 62395185-62395185 on Build GRCh38
rs956125995 Chr.20: 62394833-62394833 on Build GRCh38
rs769103109 Chr.20: 62394993-62394993 on Build GRCh38
rs747530163 Chr.20: 62394992-62394992 on Build GRCh38
rs1018793241 Chr.20: 62395194-62395194 on Build GRCh38
rs990074860 Chr.20: 62394839-62394839 on Build GRCh38
rs772372837 Chr.20: 62394987-62394987 on Build GRCh38
rs775594319 Chr.20: 62395033-62395034 on Build GRCh38
rs928148298 Chr.20: 62395267-62395267 on Build GRCh38
rs961852097 Chr.20: 62395197-62395197 on Build GRCh38
rs752680200 Chr.20: 62395037-62395037 on Build GRCh38
rs573945287 Chr.20: 62394959-62394959 on Build GRCh38

View Full Product Details