Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62564772-62565306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR1-1HG
Entrez Gene ID
128826
Gene Name
MIR1-1 host gene
Gene Aliases
C20orf166, MIR133A2HG, dJ353C17.1
UniGene
Hs.86507
Gene Chromosome Location
Chr.20: 62550453-62570764 on Build GRCh38
Transcripts
NM_001302812.1 NM_178463.3 NR_029676.1

Gene Symbol
MIR133A2
Entrez Gene ID
406923
Gene Name
microRNA 133a-2
Gene Aliases
MIRN133A2, mir-133a-2
UniGene
Gene Chromosome Location
Chr.20: 62564912-62565013 on Build GRCh38
Transcripts
NM_001302812.1 NM_178463.3 NR_029676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5664486 c.114G>T p.Q38H Chr.20: 62565094-62565094 on Build GRCh38
1327509 c.118C>G p.Q40E Chr.20: 62565098-62565098 on Build GRCh38
1713767 c.40C>T p.P14S Chr.20: 62565020-62565020 on Build GRCh38
7358373 c.46G>A p.A16T Chr.20: 62565026-62565026 on Build GRCh38
4431078 c.41C>T p.P14L Chr.20: 62565021-62565021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374274740 Chr.20: 62565016-62565016 on Build GRCh38
rs111916454 Chr.20: 62565065-62565065 on Build GRCh38
rs782172092 Chr.20: 62564995-62564995 on Build GRCh38
rs199563235 Chr.20: 62565021-62565021 on Build GRCh38
rs1030709767 Chr.20: 62565070-62565070 on Build GRCh38
rs547709642 Chr.20: 62564795-62564795 on Build GRCh38
rs376376498 Chr.20: 62564906-62564906 on Build GRCh38
rs781934657 Chr.20: 62565068-62565068 on Build GRCh38
rs561962767 Chr.20: 62565011-62565011 on Build GRCh38
rs781862477 Chr.20: 62565117-62565117 on Build GRCh38
rs984116360 Chr.20: 62565178-62565178 on Build GRCh38
rs372265930 Chr.20: 62565034-62565034 on Build GRCh38
rs782422161 Chr.20: 62565071-62565071 on Build GRCh38
rs776590484 Chr.20: 62565003-62565003 on Build GRCh38
rs781959727 Chr.20: 62564878-62564878 on Build GRCh38
rs373830147 Chr.20: 62565025-62565025 on Build GRCh38
rs944144887 Chr.20: 62564859-62564859 on Build GRCh38
rs386816038 Chr.20: 62564835-62564838 on Build GRCh38
rs935142998 Chr.20: 62565282-62565282 on Build GRCh38
rs6062250 Chr.20: 62565216-62565216 on Build GRCh38
rs985664540 Chr.20: 62565122-62565122 on Build GRCh38
rs948067590 Chr.20: 62565192-62565192 on Build GRCh38
rs374291696 Chr.20: 62565121-62565121 on Build GRCh38
rs781978688 Chr.20: 62565040-62565040 on Build GRCh38
rs782199575 Chr.20: 62564896-62564896 on Build GRCh38
rs139918515 Chr.20: 62565001-62565001 on Build GRCh38
rs782449996 Chr.20: 62565063-62565063 on Build GRCh38
rs1024828602 Chr.20: 62564958-62564958 on Build GRCh38
rs369575411 Chr.20: 62564978-62564978 on Build GRCh38
rs782168781 Chr.20: 62565069-62565069 on Build GRCh38
rs782550297 Chr.20: 62564888-62564888 on Build GRCh38
rs1049891199 Chr.20: 62564851-62564851 on Build GRCh38
rs558775444 Chr.20: 62564877-62564877 on Build GRCh38
rs782692159 Chr.20: 62565064-62565064 on Build GRCh38
rs45547937 Chr.20: 62564810-62564810 on Build GRCh38
rs782105206 Chr.20: 62565002-62565002 on Build GRCh38
rs970474523 Chr.20: 62564804-62564804 on Build GRCh38
rs782792513 Chr.20: 62564999-62564999 on Build GRCh38
rs13040413 Chr.20: 62564893-62564893 on Build GRCh38
rs376144505 Chr.20: 62565104-62565104 on Build GRCh38
rs11905281 Chr.20: 62564837-62564837 on Build GRCh38
rs374881483 Chr.20: 62565150-62565150 on Build GRCh38
rs782594260 Chr.20: 62564895-62564895 on Build GRCh38
rs782526298 Chr.20: 62565066-62565066 on Build GRCh38
rs782482679 Chr.20: 62565057-62565057 on Build GRCh38
rs781967665 Chr.20: 62564931-62564931 on Build GRCh38
rs782542471 Chr.20: 62565013-62565013 on Build GRCh38
rs1048884252 Chr.20: 62565051-62565051 on Build GRCh38
rs782241284 Chr.20: 62564884-62564884 on Build GRCh38
rs782216286 Chr.20: 62565060-62565060 on Build GRCh38
rs782363736 Chr.20: 62565081-62565081 on Build GRCh38
rs545537539 Chr.20: 62565067-62565067 on Build GRCh38
rs13040566 Chr.20: 62564830-62564830 on Build GRCh38
rs369105864 Chr.20: 62565134-62565134 on Build GRCh38
rs782130509 Chr.20: 62565077-62565077 on Build GRCh38
rs782427313 Chr.20: 62565022-62565022 on Build GRCh38
rs537551832 Chr.20: 62564850-62564850 on Build GRCh38
rs781919565 Chr.20: 62565036-62565036 on Build GRCh38
rs985568356 Chr.20: 62565032-62565032 on Build GRCh38
rs781808460 Chr.20: 62565061-62565061 on Build GRCh38
rs931292537 Chr.20: 62565047-62565047 on Build GRCh38
rs6062251 Chr.20: 62565060-62565060 on Build GRCh38
rs781833632 Chr.20: 62564869-62564869 on Build GRCh38
rs530330353 Chr.20: 62565133-62565133 on Build GRCh38
rs782482851 Chr.20: 62565119-62565119 on Build GRCh38
rs782085627 Chr.20: 62564959-62564959 on Build GRCh38
rs782791397 Chr.20: 62565041-62565041 on Build GRCh38
rs111509987 Chr.20: 62565026-62565026 on Build GRCh38
rs200375711 Chr.20: 62565009-62565009 on Build GRCh38
rs916568872 Chr.20: 62565191-62565191 on Build GRCh38
rs897142278 Chr.20: 62564879-62564879 on Build GRCh38
rs376229320 Chr.20: 62564857-62564857 on Build GRCh38
rs566334362 Chr.20: 62565094-62565094 on Build GRCh38
rs553267926 Chr.20: 62564984-62564984 on Build GRCh38
rs373489679 Chr.20: 62564898-62564898 on Build GRCh38
rs781929120 Chr.20: 62564989-62564989 on Build GRCh38
rs11907343 Chr.20: 62564835-62564835 on Build GRCh38
rs781867772 Chr.20: 62565056-62565056 on Build GRCh38
rs782017709 Chr.20: 62565074-62565074 on Build GRCh38
rs112279683 Chr.20: 62565044-62565044 on Build GRCh38
rs782688865 Chr.20: 62564905-62564905 on Build GRCh38
rs782309448 Chr.20: 62565008-62565008 on Build GRCh38
rs918707127 Chr.20: 62565043-62565043 on Build GRCh38
rs906449490 Chr.20: 62565280-62565280 on Build GRCh38
rs782637945 Chr.20: 62564889-62564889 on Build GRCh38
rs781849464 Chr.20: 62564890-62564890 on Build GRCh38
rs936360936 Chr.20: 62564829-62564829 on Build GRCh38
rs782068525 Chr.20: 62564871-62564871 on Build GRCh38
rs781818103 Chr.20: 62565010-62565010 on Build GRCh38
rs912616837 Chr.20: 62564858-62564858 on Build GRCh38
rs541311917 Chr.20: 62564924-62564924 on Build GRCh38
rs782202410 Chr.20: 62564815-62564815 on Build GRCh38
rs368184486 Chr.20: 62565017-62565017 on Build GRCh38
rs977726493 Chr.20: 62564807-62564807 on Build GRCh38
rs782692383 Chr.20: 62564832-62564832 on Build GRCh38

View Full Product Details