Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 13630708-13631171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POTED
Entrez Gene ID
317754
Gene Name
POTE ankyrin domain family member D
Gene Aliases
A26B3, ANKRD21, CT104.1, POTE, POTE-21, POTE21
UniGene
Hs.742365
Gene Chromosome Location
Chr.21: 13610081-13641585 on Build GRCh38
Transcripts
NM_174981.3 XM_006723997.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3963856 c.1402T>A p.Y468N Chr.21: 13631100-13631100 on Build GRCh38
109732 c.1343C>T p.P448L Chr.21: 13631041-13631041 on Build GRCh38
6412754 c.1306G>T p.G436C Chr.21: 13631004-13631004 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777632712 Chr.21: 13631142-13631142 on Build GRCh38
rs937658823 Chr.21: 13630941-13630941 on Build GRCh38
rs200810073 Chr.21: 13631050-13631050 on Build GRCh38
rs372424274 Chr.21: 13630797-13630797 on Build GRCh38
rs903687732 Chr.21: 13631104-13631104 on Build GRCh38
rs1009472192 Chr.21: 13631046-13631046 on Build GRCh38
rs919460799 Chr.21: 13630830-13630830 on Build GRCh38
rs200065764 Chr.21: 13630994-13630994 on Build GRCh38
rs1056080561 Chr.21: 13631002-13631002 on Build GRCh38
rs896404546 Chr.21: 13631023-13631023 on Build GRCh38
rs150193699 Chr.21: 13631041-13631041 on Build GRCh38
rs374137772 Chr.21: 13630888-13630888 on Build GRCh38
rs2150356 Chr.21: 13630748-13630748 on Build GRCh38
rs1042168208 Chr.21: 13631084-13631084 on Build GRCh38
rs568367816 Chr.21: 13631059-13631059 on Build GRCh38
rs3896203 Chr.21: 13630774-13630774 on Build GRCh38
rs368861889 Chr.21: 13631063-13631063 on Build GRCh38
rs1000262860 Chr.21: 13631122-13631122 on Build GRCh38
rs1033523972 Chr.21: 13631132-13631132 on Build GRCh38
rs201618299 Chr.21: 13630938-13630938 on Build GRCh38
rs2150357 Chr.21: 13630860-13630860 on Build GRCh38

View Full Product Details