Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33520285-33520800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GART
Entrez Gene ID
2618
Gene Name
phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase
Gene Aliases
AIRS, GARS, GARTF, PAIS, PGFT, PRGS
UniGene
Hs.473648
Gene Chromosome Location
Chr.21: 33503931-33542917 on Build GRCh38
Transcripts
NM_000819.4 NM_001136005.1 NM_001136006.1 XM_005260941.1 XM_006723989.1 XM_006723990.1 XM_011529526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713944 c.1616C>T p.S539F Chr.21: 33520450-33520450 on Build GRCh38
1661948 c.1606G>C p.D536H Chr.21: 33520460-33520460 on Build GRCh38
725259 c.1540C>T p.Q514* Chr.21: 33520526-33520526 on Build GRCh38
5858542 c.1658C>T p.A553V Chr.21: 33520408-33520408 on Build GRCh38
5883625 c.1504-1G>A p.? Chr.21: 33520563-33520563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767895094 Chr.21: 33520444-33520444 on Build GRCh38
rs756149272 Chr.21: 33520473-33520473 on Build GRCh38
rs77864216 Chr.21: 33520559-33520559 on Build GRCh38
rs972958508 Chr.21: 33520518-33520518 on Build GRCh38
rs1046487787 Chr.21: 33520554-33520554 on Build GRCh38
rs1005984591 Chr.21: 33520642-33520642 on Build GRCh38
rs745634040 Chr.21: 33520534-33520534 on Build GRCh38
rs368534728 Chr.21: 33520613-33520613 on Build GRCh38
rs894735903 Chr.21: 33520697-33520697 on Build GRCh38
rs752886369 Chr.21: 33520464-33520464 on Build GRCh38
rs181940225 Chr.21: 33520764-33520764 on Build GRCh38
rs918763823 Chr.21: 33520519-33520519 on Build GRCh38
rs774238157 Chr.21: 33520380-33520380 on Build GRCh38
rs146964652 Chr.21: 33520412-33520412 on Build GRCh38
rs778891157 Chr.21: 33520526-33520526 on Build GRCh38
rs1031183260 Chr.21: 33520770-33520770 on Build GRCh38
rs749455383 Chr.21: 33520374-33520374 on Build GRCh38
rs776161541 Chr.21: 33520408-33520408 on Build GRCh38
rs775296695 Chr.21: 33520539-33520539 on Build GRCh38
rs1014365969 Chr.21: 33520712-33520712 on Build GRCh38
rs889013093 Chr.21: 33520645-33520645 on Build GRCh38
rs2834232 Chr.21: 33520589-33520589 on Build GRCh38
rs767851839 Chr.21: 33520596-33520596 on Build GRCh38
rs532083274 Chr.21: 33520769-33520769 on Build GRCh38
rs965370291 Chr.21: 33520699-33520699 on Build GRCh38
rs768074739 Chr.21: 33520543-33520543 on Build GRCh38
rs761604103 Chr.21: 33520341-33520341 on Build GRCh38
rs745830138 Chr.21: 33520394-33520394 on Build GRCh38
rs912670615 Chr.21: 33520512-33520512 on Build GRCh38
rs1028845102 Chr.21: 33520475-33520475 on Build GRCh38
rs758767216 Chr.21: 33520320-33520320 on Build GRCh38
rs749543929 Chr.21: 33520506-33520506 on Build GRCh38
rs781291902 Chr.21: 33520353-33520353 on Build GRCh38
rs906257945 Chr.21: 33520348-33520348 on Build GRCh38
rs757514401 Chr.21: 33520515-33520515 on Build GRCh38
rs543067712 Chr.21: 33520637-33520637 on Build GRCh38
rs748152476 Chr.21: 33520368-33520368 on Build GRCh38
rs777185578 Chr.21: 33520567-33520568 on Build GRCh38
rs977841712 Chr.21: 33520766-33520766 on Build GRCh38
rs370801092 Chr.21: 33520599-33520599 on Build GRCh38
rs1025708730 Chr.21: 33520756-33520756 on Build GRCh38
rs760101056 Chr.21: 33520403-33520403 on Build GRCh38
rs985851117 Chr.21: 33520508-33520508 on Build GRCh38
rs772102457 Chr.21: 33520399-33520399 on Build GRCh38
rs970434259 Chr.21: 33520723-33520723 on Build GRCh38
rs759887101 Chr.21: 33520592-33520592 on Build GRCh38
rs376062510 Chr.21: 33520726-33520726 on Build GRCh38
rs543241010 Chr.21: 33520551-33520551 on Build GRCh38
rs140192894 Chr.21: 33520373-33520373 on Build GRCh38
rs80229002 Chr.21: 33520629-33520629 on Build GRCh38
rs780338177 Chr.21: 33520331-33520331 on Build GRCh38
rs764111073 Chr.21: 33520610-33520610 on Build GRCh38
rs756594445 Chr.21: 33520608-33520608 on Build GRCh38
rs755530140 Chr.21: 33520421-33520421 on Build GRCh38
rs1034565332 Chr.21: 33520663-33520663 on Build GRCh38
rs1006930741 Chr.21: 33520665-33520665 on Build GRCh38
rs767298098 Chr.21: 33520370-33520370 on Build GRCh38
rs773819383 Chr.21: 33520323-33520323 on Build GRCh38
rs1020024918 Chr.21: 33520679-33520679 on Build GRCh38
rs147558295 Chr.21: 33520417-33520417 on Build GRCh38
rs777519167 Chr.21: 33520320-33520320 on Build GRCh38
rs370280197 Chr.21: 33520540-33520540 on Build GRCh38
rs1010780712 Chr.21: 33520427-33520427 on Build GRCh38
rs199737548 Chr.21: 33520377-33520377 on Build GRCh38
rs752087713 Chr.21: 33520334-33520334 on Build GRCh38
rs1024465731 Chr.21: 33520720-33520720 on Build GRCh38
rs1046177142 Chr.21: 33520328-33520328 on Build GRCh38
rs777847396 Chr.21: 33520489-33520489 on Build GRCh38
rs761497734 Chr.21: 33520566-33520566 on Build GRCh38
rs954057307 Chr.21: 33520500-33520500 on Build GRCh38
rs766025319 Chr.21: 33520323-33520325 on Build GRCh38
rs755442316 Chr.21: 33520345-33520345 on Build GRCh38
rs931428106 Chr.21: 33520521-33520521 on Build GRCh38
rs1048693398 Chr.21: 33520527-33520527 on Build GRCh38
rs970483510 Chr.21: 33520340-33520340 on Build GRCh38
rs554416036 Chr.21: 33520402-33520402 on Build GRCh38
rs35927582 Chr.21: 33520537-33520537 on Build GRCh38

View Full Product Details