Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38053932-38054433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCR4
Entrez Gene ID
10281
Gene Name
Down syndrome critical region 4
Gene Aliases
DCRB, DSCRB
UniGene
Hs.23251
Gene Chromosome Location
Chr.21: 38054011-38121360 on Build GRCh38
Transcripts
NM_005867.3

SNP Detail

SNP ID Chromosome Location
rs1003248789 Chr.21: 38054104-38054104 on Build GRCh38
rs776450849 Chr.21: 38054287-38054287 on Build GRCh38
rs907767964 Chr.21: 38053954-38053954 on Build GRCh38
rs1014163812 Chr.21: 38054242-38054242 on Build GRCh38
rs561769477 Chr.21: 38054256-38054256 on Build GRCh38
rs777677362 Chr.21: 38054150-38054150 on Build GRCh38
rs371149735 Chr.21: 38054402-38054402 on Build GRCh38
rs577203855 Chr.21: 38054374-38054374 on Build GRCh38
rs538708609 Chr.21: 38054106-38054106 on Build GRCh38
rs529192640 Chr.21: 38054335-38054335 on Build GRCh38
rs541266727 Chr.21: 38054180-38054180 on Build GRCh38
rs911032299 Chr.21: 38054118-38054118 on Build GRCh38
rs748264806 Chr.21: 38054007-38054007 on Build GRCh38
rs946217498 Chr.21: 38054061-38054061 on Build GRCh38
rs965274204 Chr.21: 38054324-38054324 on Build GRCh38
rs975933106 Chr.21: 38054252-38054252 on Build GRCh38
rs145422464 Chr.21: 38054355-38054355 on Build GRCh38
rs572595803 Chr.21: 38054179-38054179 on Build GRCh38
rs1054511942 Chr.21: 38053986-38053987 on Build GRCh38
rs1055116520 Chr.21: 38054214-38054214 on Build GRCh38
rs2070349 Chr.21: 38053958-38053958 on Build GRCh38
rs951998501 Chr.21: 38054177-38054177 on Build GRCh38
rs746972310 Chr.21: 38054165-38054165 on Build GRCh38
rs1018425722 Chr.21: 38054316-38054316 on Build GRCh38
rs1056143198 Chr.21: 38054134-38054134 on Build GRCh38
rs1046685706 Chr.21: 38054248-38054248 on Build GRCh38
rs907444794 Chr.21: 38054078-38054078 on Build GRCh38
rs569694758 Chr.21: 38053964-38053964 on Build GRCh38
rs772202162 Chr.21: 38054027-38054027 on Build GRCh38
rs895346469 Chr.21: 38054225-38054225 on Build GRCh38
rs922060357 Chr.21: 38053976-38053976 on Build GRCh38
rs976048988 Chr.21: 38053961-38053961 on Build GRCh38
rs965933940 Chr.21: 38054223-38054223 on Build GRCh38
rs373471679 Chr.21: 38054401-38054401 on Build GRCh38
rs951266233 Chr.21: 38054349-38054349 on Build GRCh38
rs985301926 Chr.21: 38054077-38054077 on Build GRCh38
rs558407911 Chr.21: 38054138-38054138 on Build GRCh38
rs932072066 Chr.21: 38053979-38053979 on Build GRCh38
rs929360998 Chr.21: 38054166-38054166 on Build GRCh38
rs373987284 Chr.21: 38054143-38054143 on Build GRCh38
rs942022905 Chr.21: 38054395-38054395 on Build GRCh38
rs1055251039 Chr.21: 38054396-38054396 on Build GRCh38
rs868631508 Chr.21: 38054300-38054300 on Build GRCh38
rs2070350 Chr.21: 38054254-38054254 on Build GRCh38
rs543383281 Chr.21: 38054326-38054326 on Build GRCh38
rs574808476 Chr.21: 38054271-38054271 on Build GRCh38
rs942096413 Chr.21: 38054199-38054199 on Build GRCh38

View Full Product Details