Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 5025166-5024661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICOSLG
Entrez Gene ID
23308
Gene Name
inducible T-cell costimulator ligand
Gene Aliases
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, LICOS
UniGene
Hs.14155
Gene Chromosome Location
Chr.21: 44216980-44241447 on Build GRCh38
Transcripts
NM_001283050.1 NM_001283051.1 NM_001283052.1 NM_015259.5 XM_011529514.2 XM_011529515.2 XM_011529516.2 XM_017028310.1 XM_006723899.2 XM_006723900.2 XM_011546078.2 XM_011546079.1

Gene Symbol
LOC102723996
Entrez Gene ID
102723996
Gene Name
ICOS ligand
Gene Aliases
-
UniGene
Hs.14155
Gene Chromosome Location
Chr.21: 5022044-5046678 on Build GRCh38
Transcripts
NM_001283050.1 NM_001283051.1 NM_001283052.1 NM_015259.5 XM_011529514.2 XM_011529515.2 XM_011529516.2 XM_017028310.1 XM_006723899.2 XM_006723900.2 XM_011546078.2 XM_011546079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4425975 c.51A>T p.R17R Chr.21: 44238452-44238452 on Build GRCh38
5631913 c.39C>T p.F13F Chr.21: 44238464-44238464 on Build GRCh38
1031232 c.49C>T p.R17* Chr.21: 44238454-44238454 on Build GRCh38
4425974 c.51A>T p.R17R Chr.21: 44238452-44238452 on Build GRCh38
5631912 c.39C>T p.F13F Chr.21: 44238464-44238464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942785551 Chr.21: 44238367-44238367 on Build GRCh38
rs117712254 Chr.21: 44238775-44238775 on Build GRCh38
rs560346684 Chr.21: 44238794-44238794 on Build GRCh38
rs753788754 Chr.21: 44238476-44238476 on Build GRCh38
rs55815224 Chr.21: 44238655-44238655 on Build GRCh38
rs547488608 Chr.21: 44238647-44238647 on Build GRCh38
rs771402330 Chr.21: 44238409-44238409 on Build GRCh38
rs778128955 Chr.21: 44238522-44238522 on Build GRCh38
rs911194104 Chr.21: 44238656-44238656 on Build GRCh38
rs143721741 Chr.21: 44238703-44238703 on Build GRCh38
rs189341723 Chr.21: 44238355-44238355 on Build GRCh38
rs11700561 Chr.21: 44238742-44238742 on Build GRCh38
rs541764909 Chr.21: 44238793-44238793 on Build GRCh38
rs1021279194 Chr.21: 44238554-44238554 on Build GRCh38
rs138156492 Chr.21: 44238560-44238560 on Build GRCh38
rs375330678 Chr.21: 44238509-44238509 on Build GRCh38
rs773045580 Chr.21: 44238437-44238437 on Build GRCh38
rs997436880 Chr.21: 44238418-44238418 on Build GRCh38
rs553512605 Chr.21: 44238743-44238743 on Build GRCh38
rs868687678 Chr.21: 44238516-44238516 on Build GRCh38
rs564530443 Chr.21: 44238436-44238436 on Build GRCh38
rs201294559 Chr.21: 44238534-44238534 on Build GRCh38
rs555915870 Chr.21: 44238740-44238740 on Build GRCh38
rs748198368 Chr.21: 44238709-44238709 on Build GRCh38
rs745509562 Chr.21: 44238520-44238521 on Build GRCh38
rs754822519 Chr.21: 44238479-44238479 on Build GRCh38
rs375392868 Chr.21: 44238420-44238420 on Build GRCh38
rs753468634 Chr.21: 44238510-44238510 on Build GRCh38
rs371788513 Chr.21: 44238457-44238457 on Build GRCh38
rs139012386 Chr.21: 44238741-44238741 on Build GRCh38
rs56124762 Chr.21: 44238591-44238591 on Build GRCh38
rs760455624 Chr.21: 44238453-44238453 on Build GRCh38
rs773443944 Chr.21: 44238754-44238754 on Build GRCh38
rs143526126 Chr.21: 44238521-44238521 on Build GRCh38
rs976822919 Chr.21: 44238697-44238697 on Build GRCh38
rs972869503 Chr.21: 44238600-44238600 on Build GRCh38
rs756999659 Chr.21: 44238563-44238563 on Build GRCh38
rs148852823 Chr.21: 44238361-44238361 on Build GRCh38
rs372368630 Chr.21: 44238417-44238417 on Build GRCh38
rs752756475 Chr.21: 44238520-44238520 on Build GRCh38
rs757610300 Chr.21: 44238540-44238540 on Build GRCh38
rs745771300 Chr.21: 44238644-44238644 on Build GRCh38
rs1039290643 Chr.21: 44238374-44238374 on Build GRCh38
rs533269829 Chr.21: 44238677-44238677 on Build GRCh38
rs544058839 Chr.21: 44238782-44238782 on Build GRCh38
rs761487517 Chr.21: 44238435-44238435 on Build GRCh38
rs933174109 Chr.21: 44238739-44238739 on Build GRCh38
rs55811546 Chr.21: 44238804-44238804 on Build GRCh38
rs116563117 Chr.21: 44238773-44238773 on Build GRCh38
rs1025889870 Chr.21: 44238612-44238612 on Build GRCh38
rs780768988 Chr.21: 44238492-44238493 on Build GRCh38
rs960166696 Chr.21: 44238594-44238594 on Build GRCh38
rs952978052 Chr.21: 44238809-44238809 on Build GRCh38
rs369564508 Chr.21: 44238439-44238439 on Build GRCh38
rs763739571 Chr.21: 44238434-44238434 on Build GRCh38
rs942796282 Chr.21: 44238662-44238662 on Build GRCh38
rs146223343 Chr.21: 44238733-44238733 on Build GRCh38
rs777191995 Chr.21: 44238416-44238416 on Build GRCh38
rs556313264 Chr.21: 44238744-44238744 on Build GRCh38
rs112539578 Chr.21: 44238704-44238704 on Build GRCh38
rs1001270373 Chr.21: 44238567-44238567 on Build GRCh38
rs369357407 Chr.21: 44238542-44238542 on Build GRCh38
rs755593004 Chr.21: 44238373-44238373 on Build GRCh38

View Full Product Details