Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 27979651-27980172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
UniGene
Hs.387856
Gene Chromosome Location
Chr.22: 27978014-28679865 on Build GRCh38
Transcripts
NM_001145418.1 XM_005261405.2 XM_006724171.3 XM_011530018.2 XM_017028673.1 NR_026963.1

Gene Symbol
TTC28-AS1
Entrez Gene ID
284900
Gene Name
TTC28 antisense RNA 1
Gene Aliases
TTC28-AS, TTC28AS, dJ353E16.2
UniGene
Hs.602319
Gene Chromosome Location
Chr.22: 27919376-28002679 on Build GRCh38
Transcripts
NM_001145418.1 XM_005261405.2 XM_006724171.3 XM_011530018.2 XM_017028673.1 NR_026963.1

SNP Detail

SNP ID Chromosome Location
rs904332609 Chr.22: 27980150-27980151 on Build GRCh38
rs1010113893 Chr.22: 27979815-27979815 on Build GRCh38
rs1033995654 Chr.22: 27979856-27979856 on Build GRCh38
rs568708743 Chr.22: 27979772-27979772 on Build GRCh38
rs1020165020 Chr.22: 27979907-27979907 on Build GRCh38
rs949565216 Chr.22: 27980087-27980087 on Build GRCh38
rs546106618 Chr.22: 27979854-27979855 on Build GRCh38
rs906943872 Chr.22: 27979800-27979800 on Build GRCh38
rs931297379 Chr.22: 27980029-27980029 on Build GRCh38
rs371650490 Chr.22: 27980150-27980150 on Build GRCh38
rs942740130 Chr.22: 27979688-27979688 on Build GRCh38
rs138952358 Chr.22: 27980023-27980023 on Build GRCh38
rs962098336 Chr.22: 27979996-27979996 on Build GRCh38
rs747334227 Chr.22: 27979742-27979742 on Build GRCh38
rs931576013 Chr.22: 27979721-27979721 on Build GRCh38
rs954844136 Chr.22: 27979909-27979909 on Build GRCh38
rs1057315151 Chr.22: 27979775-27979775 on Build GRCh38
rs542502578 Chr.22: 27980104-27980104 on Build GRCh38
rs374386377 Chr.22: 27980055-27980055 on Build GRCh38
rs370360376 Chr.22: 27979880-27979880 on Build GRCh38
rs887282993 Chr.22: 27979776-27979776 on Build GRCh38
rs967656614 Chr.22: 27979941-27979941 on Build GRCh38
rs113242466 Chr.22: 27979935-27979935 on Build GRCh38
rs558588189 Chr.22: 27979942-27979942 on Build GRCh38
rs1013720797 Chr.22: 27979797-27979797 on Build GRCh38
rs912677379 Chr.22: 27979968-27979968 on Build GRCh38
rs892800261 Chr.22: 27980148-27980148 on Build GRCh38
rs1034071247 Chr.22: 27980140-27980140 on Build GRCh38
rs948307500 Chr.22: 27980025-27980025 on Build GRCh38
rs546433745 Chr.22: 27980135-27980135 on Build GRCh38
rs2294820 Chr.22: 27979687-27979687 on Build GRCh38
rs200082161 Chr.22: 27979812-27979812 on Build GRCh38
rs557687487 Chr.22: 27980129-27980129 on Build GRCh38
rs575864167 Chr.22: 27980134-27980134 on Build GRCh38
rs1001230010 Chr.22: 27979806-27979806 on Build GRCh38
rs2294821 Chr.22: 27979799-27979799 on Build GRCh38
rs1042913832 Chr.22: 27980141-27980141 on Build GRCh38
rs552584545 Chr.22: 27979708-27979708 on Build GRCh38
rs981403923 Chr.22: 27980042-27980042 on Build GRCh38
rs190076060 Chr.22: 27980012-27980012 on Build GRCh38
rs986589853 Chr.22: 27979946-27979946 on Build GRCh38
rs367653830 Chr.22: 27980127-27980127 on Build GRCh38
rs527710847 Chr.22: 27980077-27980077 on Build GRCh38
rs938461088 Chr.22: 27979727-27979727 on Build GRCh38
rs184690170 Chr.22: 27979702-27979702 on Build GRCh38
rs957035210 Chr.22: 27979876-27979876 on Build GRCh38
rs60080097 Chr.22: 27980013-27980013 on Build GRCh38

View Full Product Details