Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 87346961-87347506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMO4
Entrez Gene ID
8543
Gene Name
LIM domain only 4
Gene Aliases
-
UniGene
Hs.436792
Gene Chromosome Location
Chr.1: 87328468-87348923 on Build GRCh38
Transcripts
NM_006769.3 XM_005271291.3

SNP Detail

SNP ID Chromosome Location
rs548191707 Chr.1: 87347419-87347419 on Build GRCh38
rs879084212 Chr.1: 87347347-87347347 on Build GRCh38
rs12133888 Chr.1: 87347266-87347266 on Build GRCh38
rs1007840010 Chr.1: 87347083-87347083 on Build GRCh38
rs542843121 Chr.1: 87347460-87347461 on Build GRCh38
rs960791331 Chr.1: 87347477-87347478 on Build GRCh38
rs1056770551 Chr.1: 87346995-87346995 on Build GRCh38
rs919811041 Chr.1: 87347176-87347176 on Build GRCh38
rs973891236 Chr.1: 87347142-87347142 on Build GRCh38
rs61773265 Chr.1: 87347459-87347459 on Build GRCh38
rs892712712 Chr.1: 87347034-87347034 on Build GRCh38
rs954875009 Chr.1: 87347373-87347373 on Build GRCh38
rs1027200428 Chr.1: 87347150-87347150 on Build GRCh38
rs932840834 Chr.1: 87347416-87347416 on Build GRCh38
rs970330963 Chr.1: 87347355-87347355 on Build GRCh38
rs373903920 Chr.1: 87347179-87347179 on Build GRCh38
rs945579917 Chr.1: 87347230-87347230 on Build GRCh38
rs886969914 Chr.1: 87347216-87347216 on Build GRCh38
rs1012553000 Chr.1: 87347246-87347246 on Build GRCh38
rs765923988 Chr.1: 87347165-87347169 on Build GRCh38
rs1035496645 Chr.1: 87347476-87347476 on Build GRCh38
rs925549484 Chr.1: 87347408-87347408 on Build GRCh38
rs938526987 Chr.1: 87347217-87347217 on Build GRCh38
rs191576035 Chr.1: 87347345-87347345 on Build GRCh38
rs148718742 Chr.1: 87347406-87347406 on Build GRCh38
rs890825160 Chr.1: 87347081-87347081 on Build GRCh38
rs914173520 Chr.1: 87347229-87347229 on Build GRCh38
rs1007867535 Chr.1: 87347296-87347296 on Build GRCh38
rs890863288 Chr.1: 87347286-87347286 on Build GRCh38
rs527347845 Chr.1: 87347446-87347446 on Build GRCh38
rs530707658 Chr.1: 87347441-87347441 on Build GRCh38
rs931212862 Chr.1: 87347387-87347387 on Build GRCh38
rs76142413 Chr.1: 87347461-87347461 on Build GRCh38
rs973369324 Chr.1: 87347384-87347384 on Build GRCh38
rs754211457 Chr.1: 87347068-87347068 on Build GRCh38
rs768385098 Chr.1: 87347407-87347407 on Build GRCh38
rs1015493635 Chr.1: 87347089-87347089 on Build GRCh38
rs142253228 Chr.1: 87347424-87347424 on Build GRCh38
rs144272791 Chr.1: 87347348-87347348 on Build GRCh38
rs576717567 Chr.1: 87347043-87347043 on Build GRCh38
rs987600081 Chr.1: 87347088-87347088 on Build GRCh38
rs557590697 Chr.1: 87347314-87347314 on Build GRCh38
rs767598130 Chr.1: 87347090-87347090 on Build GRCh38
rs763479467 Chr.1: 87347036-87347036 on Build GRCh38
rs903979870 Chr.1: 87347242-87347242 on Build GRCh38
rs961254954 Chr.1: 87347129-87347129 on Build GRCh38
rs1001795250 Chr.1: 87347475-87347475 on Build GRCh38
rs758244402 Chr.1: 87347382-87347382 on Build GRCh38
rs529857787 Chr.1: 87347413-87347413 on Build GRCh38
rs755235614 Chr.1: 87347069-87347069 on Build GRCh38
rs943756883 Chr.1: 87347436-87347437 on Build GRCh38
rs992171336 Chr.1: 87347377-87347377 on Build GRCh38
rs778857302 Chr.1: 87347251-87347251 on Build GRCh38
rs182867198 Chr.1: 87347356-87347356 on Build GRCh38
rs988066208 Chr.1: 87347374-87347374 on Build GRCh38
rs1021334208 Chr.1: 87347259-87347259 on Build GRCh38
rs954937057 Chr.1: 87347075-87347075 on Build GRCh38
rs779836389 Chr.1: 87347385-87347385 on Build GRCh38
rs910642414 Chr.1: 87347116-87347116 on Build GRCh38
rs1048817916 Chr.1: 87347471-87347471 on Build GRCh38
rs897087403 Chr.1: 87347326-87347326 on Build GRCh38
rs939333125 Chr.1: 87346990-87346990 on Build GRCh38
rs149593144 Chr.1: 87347219-87347219 on Build GRCh38
rs541145285 Chr.1: 87347461-87347461 on Build GRCh38
rs1043907047 Chr.1: 87347231-87347231 on Build GRCh38
rs926579624 Chr.1: 87346986-87346986 on Build GRCh38
rs541106347 Chr.1: 87347375-87347375 on Build GRCh38
rs908473517 Chr.1: 87347412-87347412 on Build GRCh38
rs1047334938 Chr.1: 87347200-87347200 on Build GRCh38
rs943352644 Chr.1: 87347133-87347133 on Build GRCh38
rs1056859644 Chr.1: 87347218-87347218 on Build GRCh38
rs930694317 Chr.1: 87346984-87346984 on Build GRCh38
rs12033984 Chr.1: 87346985-87346985 on Build GRCh38
rs776928218 Chr.1: 87347283-87347285 on Build GRCh38
rs995173294 Chr.1: 87347461-87347463 on Build GRCh38
rs575950007 Chr.1: 87347329-87347329 on Build GRCh38
rs576966074 Chr.1: 87347035-87347035 on Build GRCh38
rs6658480 Chr.1: 87347159-87347159 on Build GRCh38
rs373238612 Chr.1: 87347215-87347215 on Build GRCh38
rs756130672 Chr.1: 87347182-87347182 on Build GRCh38
rs1049976277 Chr.1: 87347273-87347273 on Build GRCh38
rs754355874 Chr.1: 87347364-87347365 on Build GRCh38

View Full Product Details