Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31326465-31326975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PATZ1
Entrez Gene ID
23598
Gene Name
POZ/BTB and AT hook containing zinc finger 1
Gene Aliases
MAZR, PATZ, RIAZ, ZBTB19, ZNF278, ZSG, dJ400N23
UniGene
Hs.731398
Gene Chromosome Location
Chr.22: 31325804-31346263 on Build GRCh38
Transcripts
NM_014323.2 NM_032050.1 NM_032052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3681417 c.2055A>C p.E685D Chr.22: 31326900-31326900 on Build GRCh38
6738121 c.2047G>T p.G683W Chr.22: 31326908-31326908 on Build GRCh38
1242621 c.2007G>A p.A669A Chr.22: 31326948-31326948 on Build GRCh38
726608 c.2063G>C p.*688S Chr.22: 31326892-31326892 on Build GRCh38
273076 c.2006C>T p.A669V Chr.22: 31326949-31326949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040166408 Chr.22: 31326550-31326550 on Build GRCh38
rs776815518 Chr.22: 31326862-31326864 on Build GRCh38
rs921681135 Chr.22: 31326868-31326868 on Build GRCh38
rs896286812 Chr.22: 31326906-31326906 on Build GRCh38
rs979502195 Chr.22: 31326894-31326894 on Build GRCh38
rs183633830 Chr.22: 31326829-31326829 on Build GRCh38
rs958783247 Chr.22: 31326539-31326539 on Build GRCh38
rs2240425 Chr.22: 31326933-31326933 on Build GRCh38
rs59795126 Chr.22: 31326828-31326828 on Build GRCh38
rs200180132 Chr.22: 31326883-31326883 on Build GRCh38
rs912711050 Chr.22: 31326838-31326838 on Build GRCh38
rs931649763 Chr.22: 31326564-31326564 on Build GRCh38
rs751644130 Chr.22: 31326880-31326880 on Build GRCh38
rs200485962 Chr.22: 31326870-31326870 on Build GRCh38
rs540767601 Chr.22: 31326926-31326926 on Build GRCh38
rs926248956 Chr.22: 31326645-31326645 on Build GRCh38
rs140254785 Chr.22: 31326498-31326498 on Build GRCh38
rs890197659 Chr.22: 31326771-31326771 on Build GRCh38
rs570092149 Chr.22: 31326576-31326576 on Build GRCh38
rs936275018 Chr.22: 31326911-31326911 on Build GRCh38
rs946062518 Chr.22: 31326584-31326584 on Build GRCh38
rs778881735 Chr.22: 31326921-31326921 on Build GRCh38
rs745896752 Chr.22: 31326862-31326863 on Build GRCh38
rs532553708 Chr.22: 31326589-31326589 on Build GRCh38
rs763469287 Chr.22: 31326949-31326949 on Build GRCh38
rs552784478 Chr.22: 31326738-31326738 on Build GRCh38
rs552870719 Chr.22: 31326680-31326680 on Build GRCh38
rs117283032 Chr.22: 31326871-31326871 on Build GRCh38
rs1039886785 Chr.22: 31326704-31326704 on Build GRCh38
rs560384984 Chr.22: 31326948-31326948 on Build GRCh38
rs760382199 Chr.22: 31326856-31326856 on Build GRCh38
rs999434766 Chr.22: 31326657-31326657 on Build GRCh38
rs568056532 Chr.22: 31326932-31326932 on Build GRCh38
rs1014988606 Chr.22: 31326819-31326819 on Build GRCh38
rs140642260 Chr.22: 31326897-31326897 on Build GRCh38
rs768562503 Chr.22: 31326919-31326919 on Build GRCh38
rs903767891 Chr.22: 31326654-31326654 on Build GRCh38
rs539183539 Chr.22: 31326727-31326727 on Build GRCh38
rs951595293 Chr.22: 31326822-31326822 on Build GRCh38
rs958565314 Chr.22: 31326840-31326840 on Build GRCh38
rs771418683 Chr.22: 31326929-31326929 on Build GRCh38
rs969375738 Chr.22: 31326807-31326807 on Build GRCh38
rs896349102 Chr.22: 31326687-31326687 on Build GRCh38
rs1051503067 Chr.22: 31326764-31326764 on Build GRCh38
rs756377516 Chr.22: 31326907-31326907 on Build GRCh38
rs944420539 Chr.22: 31326513-31326513 on Build GRCh38
rs779781336 Chr.22: 31326913-31326913 on Build GRCh38
rs1024144635 Chr.22: 31326777-31326777 on Build GRCh38
rs1043611918 Chr.22: 31326639-31326639 on Build GRCh38
rs573042481 Chr.22: 31326820-31326820 on Build GRCh38
rs765639616 Chr.22: 31326858-31326858 on Build GRCh38
rs144494015 Chr.22: 31326898-31326898 on Build GRCh38
rs753932457 Chr.22: 31326547-31326547 on Build GRCh38
rs2240424 Chr.22: 31326900-31326900 on Build GRCh38
rs770739464 Chr.22: 31326943-31326943 on Build GRCh38
rs977811178 Chr.22: 31326598-31326598 on Build GRCh38
rs753135036 Chr.22: 31326860-31326860 on Build GRCh38
rs992870154 Chr.22: 31326562-31326562 on Build GRCh38
rs566816448 Chr.22: 31326493-31326493 on Build GRCh38
rs760197285 Chr.22: 31326937-31326937 on Build GRCh38
rs1004535205 Chr.22: 31326794-31326794 on Build GRCh38
rs936226800 Chr.22: 31326701-31326701 on Build GRCh38
rs931736793 Chr.22: 31326754-31326754 on Build GRCh38
rs749125837 Chr.22: 31326918-31326918 on Build GRCh38
rs1013592509 Chr.22: 31326707-31326707 on Build GRCh38
rs995908223 Chr.22: 31326813-31326813 on Build GRCh38
rs917334649 Chr.22: 31326583-31326583 on Build GRCh38

View Full Product Details