Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36191413-36191949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL4
Entrez Gene ID
80832
Gene Name
apolipoprotein L4
Gene Aliases
APOL-IV, APOLIV
UniGene
Hs.115099
Gene Chromosome Location
Chr.22: 36189128-36204833 on Build GRCh38
Transcripts
NM_030643.4 NM_145660.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183272 c.424G>A p.A142T Chr.22: 36191704-36191704 on Build GRCh38
444930 c.252G>A p.K84K Chr.22: 36191879-36191879 on Build GRCh38
1283458 c.518C>A p.A173D Chr.22: 36191610-36191610 on Build GRCh38
4602970 c.331_332insAG p.F111fs*1 Chr.22: 36191796-36191797 on Build GRCh38
4608954 c.567C>A p.S189R Chr.22: 36191561-36191561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758551863 Chr.22: 36191892-36191892 on Build GRCh38
rs764671998 Chr.22: 36191701-36191701 on Build GRCh38
rs559666388 Chr.22: 36191767-36191767 on Build GRCh38
rs761143263 Chr.22: 36191860-36191860 on Build GRCh38
rs759059896 Chr.22: 36191610-36191610 on Build GRCh38
rs202160988 Chr.22: 36191796-36191796 on Build GRCh38
rs944667155 Chr.22: 36191747-36191747 on Build GRCh38
rs2227169 Chr.22: 36191906-36191906 on Build GRCh38
rs750941188 Chr.22: 36191711-36191711 on Build GRCh38
rs773152139 Chr.22: 36191587-36191587 on Build GRCh38
rs779680139 Chr.22: 36191459-36191459 on Build GRCh38
rs552292512 Chr.22: 36191700-36191700 on Build GRCh38
rs761069234 Chr.22: 36191716-36191716 on Build GRCh38
rs567877022 Chr.22: 36191852-36191852 on Build GRCh38
rs757616875 Chr.22: 36191558-36191558 on Build GRCh38
rs561038536 Chr.22: 36191611-36191611 on Build GRCh38
rs372907993 Chr.22: 36191473-36191473 on Build GRCh38
rs373049856 Chr.22: 36191704-36191704 on Build GRCh38
rs746989675 Chr.22: 36191776-36191776 on Build GRCh38
rs770675134 Chr.22: 36191691-36191691 on Build GRCh38
rs767460047 Chr.22: 36191520-36191520 on Build GRCh38
rs371861029 Chr.22: 36191655-36191655 on Build GRCh38
rs930352349 Chr.22: 36191854-36191854 on Build GRCh38
rs373653186 Chr.22: 36191580-36191580 on Build GRCh38
rs576629880 Chr.22: 36191521-36191521 on Build GRCh38
rs375647382 Chr.22: 36191605-36191605 on Build GRCh38
rs772137246 Chr.22: 36191672-36191672 on Build GRCh38
rs759790461 Chr.22: 36191873-36191873 on Build GRCh38
rs769468185 Chr.22: 36191819-36191819 on Build GRCh38
rs1048822421 Chr.22: 36191874-36191874 on Build GRCh38
rs2227167 Chr.22: 36191438-36191438 on Build GRCh38
rs532062993 Chr.22: 36191659-36191659 on Build GRCh38
rs570294062 Chr.22: 36191916-36191916 on Build GRCh38
rs113584116 Chr.22: 36191802-36191802 on Build GRCh38
rs757778102 Chr.22: 36191630-36191630 on Build GRCh38
rs368675825 Chr.22: 36191640-36191640 on Build GRCh38
rs768195971 Chr.22: 36191842-36191842 on Build GRCh38
rs750148218 Chr.22: 36191444-36191444 on Build GRCh38
rs780300775 Chr.22: 36191648-36191648 on Build GRCh38
rs866616562 Chr.22: 36191796-36191797 on Build GRCh38
rs911795877 Chr.22: 36191727-36191727 on Build GRCh38
rs536811185 Chr.22: 36191881-36191881 on Build GRCh38
rs753389439 Chr.22: 36191543-36191543 on Build GRCh38
rs773097290 Chr.22: 36191677-36191677 on Build GRCh38
rs753129069 Chr.22: 36191462-36191462 on Build GRCh38
rs765575092 Chr.22: 36191879-36191879 on Build GRCh38
rs372303780 Chr.22: 36191800-36191801 on Build GRCh38
rs774239962 Chr.22: 36191516-36191516 on Build GRCh38
rs564007068 Chr.22: 36191512-36191512 on Build GRCh38
rs371910438 Chr.22: 36191751-36191751 on Build GRCh38
rs757150141 Chr.22: 36191904-36191904 on Build GRCh38
rs574540918 Chr.22: 36191614-36191614 on Build GRCh38
rs746034036 Chr.22: 36191569-36191569 on Build GRCh38
rs769626300 Chr.22: 36191922-36191922 on Build GRCh38
rs777777455 Chr.22: 36191894-36191894 on Build GRCh38
rs749006492 Chr.22: 36191925-36191925 on Build GRCh38
rs770750848 Chr.22: 36191786-36191786 on Build GRCh38
rs541276993 Chr.22: 36191593-36191593 on Build GRCh38
rs755653825 Chr.22: 36191448-36191448 on Build GRCh38
rs78582347 Chr.22: 36191840-36191840 on Build GRCh38
rs370544575 Chr.22: 36191628-36191628 on Build GRCh38
rs202197928 Chr.22: 36191570-36191570 on Build GRCh38
rs758359131 Chr.22: 36191770-36191770 on Build GRCh38
rs765746722 Chr.22: 36191540-36191540 on Build GRCh38
rs777827259 Chr.22: 36191773-36191773 on Build GRCh38
rs373464413 Chr.22: 36191752-36191752 on Build GRCh38
rs888584406 Chr.22: 36191876-36191876 on Build GRCh38
rs2227168 Chr.22: 36191463-36191463 on Build GRCh38
rs780866198 Chr.22: 36191792-36191792 on Build GRCh38
rs369164521 Chr.22: 36191489-36191489 on Build GRCh38
rs1003525857 Chr.22: 36191638-36191638 on Build GRCh38
rs72487618 Chr.22: 36191579-36191579 on Build GRCh38
rs924008020 Chr.22: 36191488-36191488 on Build GRCh38
rs3075364 Chr.22: 36191801-36191802 on Build GRCh38
rs915017970 Chr.22: 36191678-36191678 on Build GRCh38
rs752852742 Chr.22: 36191763-36191763 on Build GRCh38
rs774933689 Chr.22: 36191838-36191838 on Build GRCh38
rs1035980977 Chr.22: 36191801-36191801 on Build GRCh38
rs766720139 Chr.22: 36191738-36191738 on Build GRCh38
rs774986936 Chr.22: 36191703-36191703 on Build GRCh38
rs745461165 Chr.22: 36191794-36191794 on Build GRCh38
rs132700 Chr.22: 36191656-36191656 on Build GRCh38
rs764644992 Chr.22: 36191548-36191548 on Build GRCh38
rs766082814 Chr.22: 36191600-36191600 on Build GRCh38
rs567799767 Chr.22: 36191479-36191479 on Build GRCh38
rs374527418 Chr.22: 36191863-36191863 on Build GRCh38
rs746839143 Chr.22: 36191680-36191680 on Build GRCh38
rs375845529 Chr.22: 36191483-36191483 on Build GRCh38
rs869115251 Chr.22: 36191802-36191803 on Build GRCh38
rs751459617 Chr.22: 36191896-36191896 on Build GRCh38
rs373026421 Chr.22: 36191493-36191493 on Build GRCh38
rs556656952 Chr.22: 36191518-36191518 on Build GRCh38
rs146308862 Chr.22: 36191634-36191634 on Build GRCh38
rs749408154 Chr.22: 36191574-36191574 on Build GRCh38
rs112894680 Chr.22: 36191803-36191803 on Build GRCh38
rs767805122 Chr.22: 36191631-36191631 on Build GRCh38
rs760431363 Chr.22: 36191531-36191531 on Build GRCh38
rs748038530 Chr.22: 36191660-36191660 on Build GRCh38
rs768034738 Chr.22: 36191705-36191705 on Build GRCh38
rs771338528 Chr.22: 36191481-36191481 on Build GRCh38
rs71314965 Chr.22: 36191542-36191542 on Build GRCh38
rs754076165 Chr.22: 36191746-36191746 on Build GRCh38
rs777929187 Chr.22: 36191464-36191464 on Build GRCh38
rs185794495 Chr.22: 36191578-36191578 on Build GRCh38
rs373611761 Chr.22: 36191557-36191557 on Build GRCh38
rs1005347908 Chr.22: 36191822-36191822 on Build GRCh38
rs372227838 Chr.22: 36191615-36191615 on Build GRCh38
rs370797006 Chr.22: 36191573-36191573 on Build GRCh38
rs4435550 Chr.22: 36191780-36191780 on Build GRCh38
rs915112917 Chr.22: 36191502-36191502 on Build GRCh38
rs776344717 Chr.22: 36191695-36191695 on Build GRCh38
rs376873369 Chr.22: 36191546-36191546 on Build GRCh38
rs888624607 Chr.22: 36191597-36191597 on Build GRCh38
rs781596763 Chr.22: 36191562-36191562 on Build GRCh38

View Full Product Details