Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 11582208-11582709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VGLL4
Entrez Gene ID
9686
Gene Name
vestigial like family member 4
Gene Aliases
VGL-4
UniGene
Hs.740389
Gene Chromosome Location
Chr.3: 11556067-11720768 on Build GRCh38
Transcripts
NM_001128219.2 NM_001128220.2 NM_001284390.1 NM_001284391.1 NM_014667.3 XM_011534267.2 XM_011534268.1 XM_011534269.1 XM_017007541.1 XM_017007542.1

SNP Detail

SNP ID Chromosome Location
rs769395143 Chr.3: 11582306-11582306 on Build GRCh38
rs978474464 Chr.3: 11582463-11582463 on Build GRCh38
rs1043087987 Chr.3: 11582317-11582317 on Build GRCh38
rs765947019 Chr.3: 11582324-11582324 on Build GRCh38
rs888824955 Chr.3: 11582557-11582557 on Build GRCh38
rs984373348 Chr.3: 11582441-11582441 on Build GRCh38
rs562097161 Chr.3: 11582663-11582663 on Build GRCh38
rs772761631 Chr.3: 11582309-11582309 on Build GRCh38
rs978545970 Chr.3: 11582435-11582454 on Build GRCh38
rs940146815 Chr.3: 11582514-11582514 on Build GRCh38
rs551505449 Chr.3: 11582435-11582435 on Build GRCh38
rs1025561789 Chr.3: 11582413-11582413 on Build GRCh38
rs897578022 Chr.3: 11582307-11582307 on Build GRCh38
rs752553890 Chr.3: 11582328-11582328 on Build GRCh38
rs913499872 Chr.3: 11582458-11582458 on Build GRCh38
rs751484775 Chr.3: 11582339-11582339 on Build GRCh38
rs1020812827 Chr.3: 11582653-11582653 on Build GRCh38
rs1032176451 Chr.3: 11582387-11582387 on Build GRCh38
rs1001267490 Chr.3: 11582375-11582375 on Build GRCh38
rs753204562 Chr.3: 11582475-11582475 on Build GRCh38
rs758845187 Chr.3: 11582549-11582549 on Build GRCh38
rs1029043421 Chr.3: 11582640-11582640 on Build GRCh38
rs762410615 Chr.3: 11582321-11582321 on Build GRCh38
rs895526590 Chr.3: 11582524-11582524 on Build GRCh38
rs904291089 Chr.3: 11582344-11582344 on Build GRCh38
rs536939038 Chr.3: 11582442-11582442 on Build GRCh38
rs946270463 Chr.3: 11582462-11582462 on Build GRCh38
rs777808241 Chr.3: 11582302-11582302 on Build GRCh38
rs559763250 Chr.3: 11582395-11582395 on Build GRCh38
rs374191515 Chr.3: 11582278-11582278 on Build GRCh38
rs756345144 Chr.3: 11582329-11582329 on Build GRCh38
rs1036489757 Chr.3: 11582288-11582288 on Build GRCh38
rs547554543 Chr.3: 11582372-11582372 on Build GRCh38
rs925597229 Chr.3: 11582465-11582465 on Build GRCh38
rs548616286 Chr.3: 11582525-11582525 on Build GRCh38
rs544104845 Chr.3: 11582615-11582615 on Build GRCh38
rs780559419 Chr.3: 11582428-11582430 on Build GRCh38
rs951339842 Chr.3: 11582417-11582417 on Build GRCh38
rs757240311 Chr.3: 11582347-11582347 on Build GRCh38
rs774225342 Chr.3: 11582326-11582326 on Build GRCh38
rs779152984 Chr.3: 11582349-11582349 on Build GRCh38
rs576852730 Chr.3: 11582614-11582614 on Build GRCh38
rs988012532 Chr.3: 11582446-11582446 on Build GRCh38
rs183288851 Chr.3: 11582327-11582327 on Build GRCh38
rs566902123 Chr.3: 11582537-11582537 on Build GRCh38
rs755555021 Chr.3: 11582622-11582625 on Build GRCh38
rs145171310 Chr.3: 11582612-11582612 on Build GRCh38
rs187843368 Chr.3: 11582546-11582546 on Build GRCh38
rs1051410795 Chr.3: 11582556-11582556 on Build GRCh38
rs954767053 Chr.3: 11582660-11582660 on Build GRCh38
rs1020853478 Chr.3: 11582672-11582672 on Build GRCh38
rs1037222754 Chr.3: 11582526-11582526 on Build GRCh38
rs1007505865 Chr.3: 11582377-11582377 on Build GRCh38
rs772513989 Chr.3: 11582265-11582265 on Build GRCh38
rs756779565 Chr.3: 11582397-11582397 on Build GRCh38
rs926647309 Chr.3: 11582438-11582438 on Build GRCh38
rs982505868 Chr.3: 11582418-11582418 on Build GRCh38
rs192882159 Chr.3: 11582587-11582587 on Build GRCh38
rs569623302 Chr.3: 11582439-11582439 on Build GRCh38
rs561130315 Chr.3: 11582277-11582277 on Build GRCh38
rs183541253 Chr.3: 11582590-11582590 on Build GRCh38
rs1009404446 Chr.3: 11582396-11582396 on Build GRCh38
rs970849952 Chr.3: 11582402-11582402 on Build GRCh38
rs937108283 Chr.3: 11582508-11582508 on Build GRCh38
rs780863692 Chr.3: 11582613-11582613 on Build GRCh38
rs539650654 Chr.3: 11582603-11582603 on Build GRCh38
rs778134538 Chr.3: 11582582-11582582 on Build GRCh38
rs562059042 Chr.3: 11582275-11582275 on Build GRCh38
rs543875783 Chr.3: 11582258-11582258 on Build GRCh38
rs533002545 Chr.3: 11582399-11582399 on Build GRCh38
rs1009785549 Chr.3: 11582616-11582616 on Build GRCh38
rs753919573 Chr.3: 11582346-11582346 on Build GRCh38
rs1016652639 Chr.3: 11582433-11582433 on Build GRCh38
rs765875936 Chr.3: 11582461-11582461 on Build GRCh38
rs576765950 Chr.3: 11582252-11582252 on Build GRCh38

View Full Product Details