Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19947395-19947897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB5A
Entrez Gene ID
5868
Gene Name
RAB5A, member RAS oncogene family
Gene Aliases
RAB5
UniGene
Hs.475663
Gene Chromosome Location
Chr.3: 19947080-19985175 on Build GRCh38
Transcripts
NM_001292048.1 NM_004162.4

SNP Detail

SNP ID Chromosome Location
rs537238139 Chr.3: 19947501-19947501 on Build GRCh38
rs1051676032 Chr.3: 19947617-19947617 on Build GRCh38
rs968186942 Chr.3: 19947843-19947843 on Build GRCh38
rs537348857 Chr.3: 19947825-19947825 on Build GRCh38
rs4858133 Chr.3: 19947541-19947541 on Build GRCh38
rs958319571 Chr.3: 19947494-19947494 on Build GRCh38
rs370378591 Chr.3: 19947427-19947427 on Build GRCh38
rs773303409 Chr.3: 19947797-19947797 on Build GRCh38
rs70937044 Chr.3: 19947671-19947671 on Build GRCh38
rs545949813 Chr.3: 19947630-19947630 on Build GRCh38
rs564344317 Chr.3: 19947665-19947665 on Build GRCh38
rs942595502 Chr.3: 19947543-19947543 on Build GRCh38
rs931928436 Chr.3: 19947593-19947593 on Build GRCh38
rs566240462 Chr.3: 19947475-19947475 on Build GRCh38
rs963883319 Chr.3: 19947508-19947508 on Build GRCh38
rs1000357944 Chr.3: 19947822-19947822 on Build GRCh38
rs147229022 Chr.3: 19947490-19947490 on Build GRCh38
rs559434718 Chr.3: 19947752-19947752 on Build GRCh38
rs36113684 Chr.3: 19947614-19947615 on Build GRCh38
rs1051685649 Chr.3: 19947875-19947875 on Build GRCh38
rs1041806606 Chr.3: 19947784-19947784 on Build GRCh38
rs898886318 Chr.3: 19947703-19947703 on Build GRCh38
rs368836151 Chr.3: 19947851-19947851 on Build GRCh38
rs1039351701 Chr.3: 19947544-19947544 on Build GRCh38
rs141294840 Chr.3: 19947827-19947827 on Build GRCh38
rs900851111 Chr.3: 19947545-19947545 on Build GRCh38
rs890125992 Chr.3: 19947445-19947446 on Build GRCh38
rs909724639 Chr.3: 19947718-19947718 on Build GRCh38
rs915322702 Chr.3: 19947840-19947840 on Build GRCh38
rs1032514416 Chr.3: 19947615-19947615 on Build GRCh38
rs947224093 Chr.3: 19947561-19947561 on Build GRCh38
rs902958261 Chr.3: 19947582-19947582 on Build GRCh38
rs77756562 Chr.3: 19947870-19947870 on Build GRCh38
rs951586348 Chr.3: 19947691-19947691 on Build GRCh38
rs900317349 Chr.3: 19947588-19947588 on Build GRCh38
rs112312922 Chr.3: 19947654-19947654 on Build GRCh38
rs368413653 Chr.3: 19947424-19947424 on Build GRCh38
rs548256170 Chr.3: 19947786-19947786 on Build GRCh38
rs966919967 Chr.3: 19947452-19947452 on Build GRCh38
rs569954001 Chr.3: 19947789-19947789 on Build GRCh38
rs890135777 Chr.3: 19947627-19947627 on Build GRCh38
rs575574352 Chr.3: 19947605-19947605 on Build GRCh38
rs983930949 Chr.3: 19947706-19947706 on Build GRCh38
rs970754451 Chr.3: 19947517-19947517 on Build GRCh38
rs1030028463 Chr.3: 19947433-19947433 on Build GRCh38
rs918666767 Chr.3: 19947677-19947677 on Build GRCh38
rs372701791 Chr.3: 19947567-19947567 on Build GRCh38
rs1035200276 Chr.3: 19947790-19947790 on Build GRCh38
rs6781143 Chr.3: 19947737-19947737 on Build GRCh38
rs988677500 Chr.3: 19947849-19947861 on Build GRCh38
rs188259696 Chr.3: 19947547-19947547 on Build GRCh38
rs902932894 Chr.3: 19947791-19947791 on Build GRCh38
rs535081026 Chr.3: 19947853-19947853 on Build GRCh38
rs542242917 Chr.3: 19947556-19947556 on Build GRCh38
rs115902578 Chr.3: 19947504-19947504 on Build GRCh38
rs978687122 Chr.3: 19947479-19947479 on Build GRCh38
rs386659066 Chr.3: 19947424-19947427 on Build GRCh38
rs1023563995 Chr.3: 19947644-19947644 on Build GRCh38
rs547192725 Chr.3: 19947743-19947743 on Build GRCh38
rs767518516 Chr.3: 19947785-19947785 on Build GRCh38
rs370279152 Chr.3: 19947446-19947447 on Build GRCh38
rs1048732679 Chr.3: 19947540-19947540 on Build GRCh38
rs569996874 Chr.3: 19947498-19947498 on Build GRCh38
rs939956694 Chr.3: 19947580-19947580 on Build GRCh38
rs918392525 Chr.3: 19947531-19947531 on Build GRCh38
rs900882462 Chr.3: 19947418-19947418 on Build GRCh38
rs956706910 Chr.3: 19947821-19947821 on Build GRCh38
rs116252091 Chr.3: 19947505-19947505 on Build GRCh38
rs1042864706 Chr.3: 19947585-19947585 on Build GRCh38
rs929808130 Chr.3: 19947539-19947539 on Build GRCh38
rs35001626 Chr.3: 19947751-19947751 on Build GRCh38
rs958246484 Chr.3: 19947620-19947620 on Build GRCh38
rs973574232 Chr.3: 19947535-19947535 on Build GRCh38
rs529638299 Chr.3: 19947771-19947771 on Build GRCh38
rs982232426 Chr.3: 19947525-19947525 on Build GRCh38
rs922534797 Chr.3: 19947764-19947764 on Build GRCh38
rs528722594 Chr.3: 19947491-19947491 on Build GRCh38
rs140617353 Chr.3: 19947574-19947574 on Build GRCh38
rs774722512 Chr.3: 19947648-19947648 on Build GRCh38

View Full Product Details