Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19950688-19951185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB5A
Entrez Gene ID
5868
Gene Name
RAB5A, member RAS oncogene family
Gene Aliases
RAB5
UniGene
Hs.475663
Gene Chromosome Location
Chr.3: 19947080-19985175 on Build GRCh38
Transcripts
NM_001292048.1 NM_004162.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412013 c.20C>A p.T7K Chr.3: 19950918-19950918 on Build GRCh38
4719858 c.34C>G p.P12A Chr.3: 19950932-19950932 on Build GRCh38
6761587 c.134T>A p.F45Y Chr.3: 19951032-19951032 on Build GRCh38
6224943 c.31G>A p.G11R Chr.3: 19950929-19950929 on Build GRCh38
3592018 c.30C>T p.N10N Chr.3: 19950928-19950928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036335227 Chr.3: 19951077-19951077 on Build GRCh38
rs774566349 Chr.3: 19950906-19950906 on Build GRCh38
rs543002874 Chr.3: 19950785-19950785 on Build GRCh38
rs771004325 Chr.3: 19950970-19950970 on Build GRCh38
rs1007804178 Chr.3: 19951147-19951147 on Build GRCh38
rs749444935 Chr.3: 19950889-19950889 on Build GRCh38
rs747992546 Chr.3: 19951027-19951027 on Build GRCh38
rs1019273362 Chr.3: 19950765-19950765 on Build GRCh38
rs765895417 Chr.3: 19951061-19951061 on Build GRCh38
rs781710069 Chr.3: 19950862-19950862 on Build GRCh38
rs149313297 Chr.3: 19950964-19950964 on Build GRCh38
rs371868725 Chr.3: 19950851-19950851 on Build GRCh38
rs189794197 Chr.3: 19950773-19950773 on Build GRCh38
rs200047397 Chr.3: 19950909-19950909 on Build GRCh38
rs11717172 Chr.3: 19950820-19950820 on Build GRCh38
rs755357753 Chr.3: 19950856-19950856 on Build GRCh38
rs10510496 Chr.3: 19951096-19951096 on Build GRCh38
rs1045999103 Chr.3: 19950858-19950858 on Build GRCh38
rs111819761 Chr.3: 19950913-19950913 on Build GRCh38
rs115981030 Chr.3: 19950896-19950896 on Build GRCh38
rs772319638 Chr.3: 19951008-19951008 on Build GRCh38
rs762827236 Chr.3: 19951048-19951048 on Build GRCh38
rs749885739 Chr.3: 19950928-19950928 on Build GRCh38
rs532008844 Chr.3: 19950846-19950846 on Build GRCh38
rs770930318 Chr.3: 19950890-19950890 on Build GRCh38
rs770467311 Chr.3: 19950876-19950876 on Build GRCh38
rs149206559 Chr.3: 19950938-19950938 on Build GRCh38
rs781375870 Chr.3: 19951054-19951054 on Build GRCh38
rs764398542 Chr.3: 19951101-19951101 on Build GRCh38
rs1007656246 Chr.3: 19950941-19950941 on Build GRCh38
rs775197969 Chr.3: 19951031-19951031 on Build GRCh38
rs11545950 Chr.3: 19950952-19950952 on Build GRCh38
rs765282057 Chr.3: 19950917-19950917 on Build GRCh38
rs180981510 Chr.3: 19951100-19951100 on Build GRCh38
rs778287571 Chr.3: 19950956-19950956 on Build GRCh38
rs148447810 Chr.3: 19950985-19950985 on Build GRCh38
rs754321196 Chr.3: 19951111-19951111 on Build GRCh38
rs764547158 Chr.3: 19950931-19950931 on Build GRCh38
rs6773437 Chr.3: 19950940-19950940 on Build GRCh38
rs748645733 Chr.3: 19950873-19950873 on Build GRCh38
rs754561415 Chr.3: 19951111-19951112 on Build GRCh38
rs35986369 Chr.3: 19950975-19950976 on Build GRCh38
rs752050318 Chr.3: 19950853-19950856 on Build GRCh38
rs147139196 Chr.3: 19950953-19950953 on Build GRCh38
rs994934469 Chr.3: 19950717-19950717 on Build GRCh38
rs201114014 Chr.3: 19951060-19951060 on Build GRCh38
rs1027463591 Chr.3: 19950725-19950725 on Build GRCh38
rs57181336 Chr.3: 19950711-19950711 on Build GRCh38
rs756202716 Chr.3: 19951132-19951132 on Build GRCh38
rs774047411 Chr.3: 19951067-19951067 on Build GRCh38
rs184924584 Chr.3: 19950735-19950735 on Build GRCh38
rs953313807 Chr.3: 19950727-19950727 on Build GRCh38
rs941150846 Chr.3: 19950740-19950740 on Build GRCh38
rs746068565 Chr.3: 19950991-19950991 on Build GRCh38
rs748616736 Chr.3: 19951098-19951098 on Build GRCh38
rs966029028 Chr.3: 19950801-19950801 on Build GRCh38
rs777711883 Chr.3: 19950881-19950881 on Build GRCh38
rs773130913 Chr.3: 19951034-19951034 on Build GRCh38
rs781668564 Chr.3: 19950942-19950942 on Build GRCh38
rs759262097 Chr.3: 19951071-19951071 on Build GRCh38
rs779811315 Chr.3: 19950848-19950848 on Build GRCh38
rs374073544 Chr.3: 19951088-19951088 on Build GRCh38
rs144198452 Chr.3: 19950914-19950914 on Build GRCh38
rs369479155 Chr.3: 19950802-19950802 on Build GRCh38
rs761706329 Chr.3: 19950916-19950916 on Build GRCh38
rs139006514 Chr.3: 19950930-19950930 on Build GRCh38
rs889239494 Chr.3: 19951110-19951110 on Build GRCh38
rs1049159601 Chr.3: 19951104-19951104 on Build GRCh38
rs951397737 Chr.3: 19951076-19951076 on Build GRCh38
rs760256312 Chr.3: 19950815-19950815 on Build GRCh38

View Full Product Details