Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19984199-19984738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PP2D1
Entrez Gene ID
151649
Gene Name
protein phosphatase 2C like domain containing 1
Gene Aliases
C3orf48
UniGene
Hs.733161
Gene Chromosome Location
Chr.3: 19979961-20012273 on Build GRCh38
Transcripts
NR_027694.1 NM_001292048.1 NM_004162.4

Gene Symbol
RAB5A
Entrez Gene ID
5868
Gene Name
RAB5A, member RAS oncogene family
Gene Aliases
RAB5
UniGene
Hs.475663
Gene Chromosome Location
Chr.3: 19947080-19985175 on Build GRCh38
Transcripts
NR_027694.1 NM_001292048.1 NM_004162.4

SNP Detail

SNP ID Chromosome Location
rs537345201 Chr.3: 19984429-19984429 on Build GRCh38
rs148311294 Chr.3: 19984315-19984315 on Build GRCh38
rs529989810 Chr.3: 19984549-19984549 on Build GRCh38
rs759393574 Chr.3: 19984265-19984265 on Build GRCh38
rs753656222 Chr.3: 19984223-19984223 on Build GRCh38
rs569586973 Chr.3: 19984414-19984414 on Build GRCh38
rs762686643 Chr.3: 19984314-19984314 on Build GRCh38
rs988254056 Chr.3: 19984450-19984450 on Build GRCh38
rs548150020 Chr.3: 19984408-19984408 on Build GRCh38
rs200703696 Chr.3: 19984286-19984286 on Build GRCh38
rs1042896296 Chr.3: 19984489-19984489 on Build GRCh38
rs878925852 Chr.3: 19984621-19984621 on Build GRCh38
rs142990198 Chr.3: 19984594-19984594 on Build GRCh38
rs750373227 Chr.3: 19984371-19984371 on Build GRCh38
rs764984761 Chr.3: 19984357-19984357 on Build GRCh38
rs931955247 Chr.3: 19984558-19984558 on Build GRCh38
rs764777175 Chr.3: 19984283-19984283 on Build GRCh38
rs947242784 Chr.3: 19984488-19984488 on Build GRCh38
rs756691250 Chr.3: 19984642-19984642 on Build GRCh38
rs890134387 Chr.3: 19984577-19984577 on Build GRCh38
rs935290463 Chr.3: 19984445-19984445 on Build GRCh38
rs982764216 Chr.3: 19984432-19984432 on Build GRCh38
rs751066724 Chr.3: 19984575-19984575 on Build GRCh38
rs553016901 Chr.3: 19984611-19984611 on Build GRCh38
rs541250739 Chr.3: 19984330-19984333 on Build GRCh38
rs1030031005 Chr.3: 19984297-19984297 on Build GRCh38
rs576145141 Chr.3: 19984631-19984631 on Build GRCh38
rs915298886 Chr.3: 19984479-19984479 on Build GRCh38
rs761678627 Chr.3: 19984247-19984247 on Build GRCh38
rs996479516 Chr.3: 19984295-19984295 on Build GRCh38
rs1011333515 Chr.3: 19984252-19984252 on Build GRCh38
rs772634040 Chr.3: 19984299-19984299 on Build GRCh38
rs570265786 Chr.3: 19984503-19984503 on Build GRCh38
rs1014700341 Chr.3: 19984621-19984621 on Build GRCh38
rs969659106 Chr.3: 19984415-19984419 on Build GRCh38
rs45542931 Chr.3: 19984433-19984433 on Build GRCh38
rs540230533 Chr.3: 19984569-19984569 on Build GRCh38
rs574899435 Chr.3: 19984664-19984664 on Build GRCh38
rs900334977 Chr.3: 19984491-19984491 on Build GRCh38
rs916730670 Chr.3: 19984415-19984415 on Build GRCh38
rs573813685 Chr.3: 19984366-19984366 on Build GRCh38
rs536451350 Chr.3: 19984382-19984382 on Build GRCh38
rs1023844739 Chr.3: 19984662-19984662 on Build GRCh38

View Full Product Details