Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 31621933-31622452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STT3B
Entrez Gene ID
201595
Gene Name
STT3B, catalytic subunit of the oligosaccharyltransferase complex
Gene Aliases
CDG1X, SIMP, STT3-B
UniGene
Hs.475812
Gene Chromosome Location
Chr.3: 31532501-31637622 on Build GRCh38
Transcripts
NM_178862.2 XM_017005857.1 XM_017005858.1 XM_017005859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6331929 c.1381A>T p.M461L Chr.3: 31622150-31622150 on Build GRCh38
7306257 c.1381A>C p.M461L Chr.3: 31622150-31622150 on Build GRCh38
1043570 c.1375C>T p.R459* Chr.3: 31622144-31622144 on Build GRCh38
6295418 c.1402G>T p.V468L Chr.3: 31622171-31622171 on Build GRCh38
1753157 c.1435G>T p.V479F Chr.3: 31622204-31622204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896848010 Chr.3: 31622117-31622117 on Build GRCh38
rs1019747156 Chr.3: 31622368-31622368 on Build GRCh38
rs989338969 Chr.3: 31622410-31622410 on Build GRCh38
rs745834383 Chr.3: 31622074-31622074 on Build GRCh38
rs773579456 Chr.3: 31622351-31622351 on Build GRCh38
rs749665192 Chr.3: 31622171-31622171 on Build GRCh38
rs763454701 Chr.3: 31622188-31622188 on Build GRCh38
rs778721341 Chr.3: 31622070-31622070 on Build GRCh38
rs768417967 Chr.3: 31622110-31622110 on Build GRCh38
rs746754679 Chr.3: 31622064-31622065 on Build GRCh38
rs551957725 Chr.3: 31622251-31622251 on Build GRCh38
rs747786174 Chr.3: 31622333-31622333 on Build GRCh38
rs765539767 Chr.3: 31622175-31622175 on Build GRCh38
rs138738231 Chr.3: 31622042-31622042 on Build GRCh38
rs925125269 Chr.3: 31622031-31622031 on Build GRCh38
rs949120022 Chr.3: 31622072-31622072 on Build GRCh38
rs543727898 Chr.3: 31622002-31622002 on Build GRCh38
rs10662891 Chr.3: 31622023-31622024 on Build GRCh38
rs766813650 Chr.3: 31622189-31622189 on Build GRCh38
rs746554485 Chr.3: 31622317-31622317 on Build GRCh38
rs748595130 Chr.3: 31622385-31622385 on Build GRCh38
rs559861697 Chr.3: 31622111-31622111 on Build GRCh38
rs773524257 Chr.3: 31622177-31622177 on Build GRCh38
rs755019626 Chr.3: 31622194-31622194 on Build GRCh38
rs1054980595 Chr.3: 31622041-31622041 on Build GRCh38
rs937805346 Chr.3: 31622037-31622037 on Build GRCh38
rs755205429 Chr.3: 31622050-31622050 on Build GRCh38
rs767831091 Chr.3: 31622224-31622224 on Build GRCh38
rs905900632 Chr.3: 31622164-31622164 on Build GRCh38
rs547979609 Chr.3: 31621966-31621966 on Build GRCh38
rs1057184599 Chr.3: 31622323-31622323 on Build GRCh38
rs538159071 Chr.3: 31622341-31622341 on Build GRCh38
rs372199533 Chr.3: 31622081-31622081 on Build GRCh38
rs141872505 Chr.3: 31622302-31622302 on Build GRCh38
rs890053753 Chr.3: 31622329-31622329 on Build GRCh38
rs71097434 Chr.3: 31622025-31622026 on Build GRCh38
rs199741348 Chr.3: 31622026-31622026 on Build GRCh38
rs143950703 Chr.3: 31622167-31622167 on Build GRCh38
rs1044406953 Chr.3: 31622108-31622108 on Build GRCh38
rs547760757 Chr.3: 31622046-31622046 on Build GRCh38
rs775157879 Chr.3: 31622104-31622104 on Build GRCh38
rs780701285 Chr.3: 31622320-31622320 on Build GRCh38
rs758472135 Chr.3: 31622270-31622270 on Build GRCh38
rs112888755 Chr.3: 31622023-31622023 on Build GRCh38
rs117459028 Chr.3: 31622377-31622377 on Build GRCh38
rs751705264 Chr.3: 31622193-31622193 on Build GRCh38
rs562518923 Chr.3: 31622013-31622013 on Build GRCh38
rs199910542 Chr.3: 31622356-31622356 on Build GRCh38
rs369285498 Chr.3: 31622252-31622252 on Build GRCh38
rs750189081 Chr.3: 31622264-31622264 on Build GRCh38
rs374931507 Chr.3: 31622107-31622107 on Build GRCh38
rs965091684 Chr.3: 31621977-31621977 on Build GRCh38
rs913224683 Chr.3: 31621985-31621985 on Build GRCh38
rs752886119 Chr.3: 31622225-31622225 on Build GRCh38
rs929460891 Chr.3: 31621989-31621989 on Build GRCh38
rs781267393 Chr.3: 31622060-31622060 on Build GRCh38
rs151177461 Chr.3: 31622021-31622022 on Build GRCh38
rs587777217 Chr.3: 31622328-31622328 on Build GRCh38
rs918338427 Chr.3: 31621987-31621987 on Build GRCh38
rs375438745 Chr.3: 31622067-31622067 on Build GRCh38
rs768027749 Chr.3: 31622319-31622319 on Build GRCh38
rs80348727 Chr.3: 31621984-31621984 on Build GRCh38
rs1020274486 Chr.3: 31622421-31622422 on Build GRCh38
rs146175720 Chr.3: 31622424-31622424 on Build GRCh38
rs979790077 Chr.3: 31622010-31622010 on Build GRCh38
rs1006982705 Chr.3: 31622335-31622335 on Build GRCh38
rs66468423 Chr.3: 31622024-31622025 on Build GRCh38
rs965392186 Chr.3: 31622383-31622383 on Build GRCh38
rs1010050804 Chr.3: 31622422-31622422 on Build GRCh38

View Full Product Details