Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36528719-36529216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAC
Entrez Gene ID
6769
Gene Name
SH3 and cysteine rich domain
Gene Aliases
STAC1
UniGene
Hs.56045
Gene Chromosome Location
Chr.3: 36380455-36548007 on Build GRCh38
Transcripts
NM_001292049.1 NM_003149.2 XM_011534036.2 XM_011534037.2 XM_011534038.2 XM_017007083.1 XM_017007084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380532 c.1010A>C p.N337T Chr.3: 36528885-36528885 on Build GRCh38
5790985 c.999C>A p.F333L Chr.3: 36528874-36528874 on Build GRCh38
5002035 c.1030C>A p.Q344K Chr.3: 36528905-36528905 on Build GRCh38
1309042 c.1089G>C p.Q363H Chr.3: 36528964-36528964 on Build GRCh38
5791515 c.989G>C p.R330T Chr.3: 36528864-36528864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75665620 Chr.3: 36528787-36528787 on Build GRCh38
rs940258701 Chr.3: 36529022-36529022 on Build GRCh38
rs914479723 Chr.3: 36528858-36528858 on Build GRCh38
rs916115105 Chr.3: 36528966-36528966 on Build GRCh38
rs749833239 Chr.3: 36529020-36529020 on Build GRCh38
rs754952678 Chr.3: 36528818-36528818 on Build GRCh38
rs374926738 Chr.3: 36528961-36528961 on Build GRCh38
rs977323951 Chr.3: 36529127-36529127 on Build GRCh38
rs901134298 Chr.3: 36529071-36529071 on Build GRCh38
rs577888046 Chr.3: 36528842-36528842 on Build GRCh38
rs574154750 Chr.3: 36528875-36528875 on Build GRCh38
rs1035593421 Chr.3: 36529114-36529114 on Build GRCh38
rs771690470 Chr.3: 36528751-36528751 on Build GRCh38
rs775182793 Chr.3: 36528757-36528757 on Build GRCh38
rs145748087 Chr.3: 36528948-36528948 on Build GRCh38
rs368298937 Chr.3: 36528942-36528942 on Build GRCh38
rs1028357923 Chr.3: 36529076-36529076 on Build GRCh38
rs779619398 Chr.3: 36528862-36528862 on Build GRCh38
rs763465147 Chr.3: 36528796-36528796 on Build GRCh38
rs559679564 Chr.3: 36528928-36528928 on Build GRCh38
rs751135400 Chr.3: 36529000-36529000 on Build GRCh38
rs545352424 Chr.3: 36529006-36529006 on Build GRCh38
rs769430475 Chr.3: 36529013-36529013 on Build GRCh38
rs867501410 Chr.3: 36528973-36528973 on Build GRCh38
rs960855599 Chr.3: 36529151-36529151 on Build GRCh38
rs144959929 Chr.3: 36528850-36528850 on Build GRCh38
rs1011159980 Chr.3: 36529082-36529085 on Build GRCh38
rs775814562 Chr.3: 36528767-36528768 on Build GRCh38
rs771436116 Chr.3: 36528936-36528936 on Build GRCh38
rs767656854 Chr.3: 36528819-36528819 on Build GRCh38
rs771381284 Chr.3: 36529030-36529030 on Build GRCh38
rs372092346 Chr.3: 36528949-36528949 on Build GRCh38
rs780719553 Chr.3: 36528893-36528893 on Build GRCh38
rs762407584 Chr.3: 36528784-36528784 on Build GRCh38
rs751810041 Chr.3: 36528815-36528815 on Build GRCh38
rs764493570 Chr.3: 36528787-36528787 on Build GRCh38
rs1031372153 Chr.3: 36529031-36529031 on Build GRCh38
rs371539813 Chr.3: 36528825-36528825 on Build GRCh38
rs1037448351 Chr.3: 36529050-36529050 on Build GRCh38
rs556587047 Chr.3: 36528777-36528777 on Build GRCh38
rs1031791601 Chr.3: 36529129-36529129 on Build GRCh38
rs398051808 Chr.3: 36528777-36528777 on Build GRCh38
rs759832462 Chr.3: 36528944-36528944 on Build GRCh38
rs747654301 Chr.3: 36529010-36529010 on Build GRCh38
rs778365806 Chr.3: 36529017-36529017 on Build GRCh38
rs370981732 Chr.3: 36529112-36529112 on Build GRCh38
rs758135966 Chr.3: 36528982-36528982 on Build GRCh38
rs757156979 Chr.3: 36528826-36528826 on Build GRCh38
rs969597971 Chr.3: 36529109-36529109 on Build GRCh38
rs549973415 Chr.3: 36529086-36529086 on Build GRCh38
rs999010050 Chr.3: 36529021-36529021 on Build GRCh38
rs541572410 Chr.3: 36528901-36528901 on Build GRCh38
rs1022515027 Chr.3: 36529083-36529083 on Build GRCh38
rs746500990 Chr.3: 36528767-36528767 on Build GRCh38
rs375565038 Chr.3: 36528833-36528833 on Build GRCh38
rs879092859 Chr.3: 36528768-36528769 on Build GRCh38
rs766700851 Chr.3: 36528811-36528811 on Build GRCh38
rs776947956 Chr.3: 36528782-36528782 on Build GRCh38
rs1007331102 Chr.3: 36529093-36529093 on Build GRCh38
rs201731723 Chr.3: 36528855-36528855 on Build GRCh38
rs948852008 Chr.3: 36528991-36528991 on Build GRCh38
rs924265203 Chr.3: 36528945-36528945 on Build GRCh38
rs746690171 Chr.3: 36528867-36528867 on Build GRCh38
rs368545283 Chr.3: 36528790-36528790 on Build GRCh38
rs148142776 Chr.3: 36529073-36529073 on Build GRCh38
rs977503451 Chr.3: 36528939-36528939 on Build GRCh38
rs763342034 Chr.3: 36528929-36528929 on Build GRCh38
rs761123421 Chr.3: 36528777-36528778 on Build GRCh38
rs779923636 Chr.3: 36528990-36528990 on Build GRCh38
rs941336769 Chr.3: 36528904-36528904 on Build GRCh38
rs11305660 Chr.3: 36528768-36528768 on Build GRCh38
rs770406342 Chr.3: 36528915-36528915 on Build GRCh38
rs867013407 Chr.3: 36528857-36528857 on Build GRCh38
rs754633520 Chr.3: 36529003-36529003 on Build GRCh38
rs775459507 Chr.3: 36529036-36529036 on Build GRCh38
rs969046625 Chr.3: 36529123-36529123 on Build GRCh38

View Full Product Details