Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46374007-46374519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR5
Entrez Gene ID
1234
Gene Name
C-C motif chemokine receptor 5 (gene/pseudogene)
Gene Aliases
CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
UniGene
Hs.450802
Gene Chromosome Location
Chr.3: 46370142-46376206 on Build GRCh38
Transcripts
NM_000579.3 NM_001100168.1 NR_125406.1

Gene Symbol
LOC102724297
Entrez Gene ID
102724297
Gene Name
uncharacterized LOC102724297
Gene Aliases
-
UniGene
Hs.622021
Gene Chromosome Location
Chr.3: 46364660-46407059 on Build GRCh38
Transcripts
NM_000579.3 NM_001100168.1 NR_125406.1

SNP Detail

SNP ID Chromosome Location
rs1057136344 Chr.3: 46374341-46374341 on Build GRCh38
rs574715855 Chr.3: 46374428-46374428 on Build GRCh38
rs746407806 Chr.3: 46374486-46374486 on Build GRCh38
rs570782804 Chr.3: 46374246-46374246 on Build GRCh38
rs962500340 Chr.3: 46374282-46374282 on Build GRCh38
rs536872608 Chr.3: 46374036-46374036 on Build GRCh38
rs752397660 Chr.3: 46374157-46374157 on Build GRCh38
rs764395007 Chr.3: 46374247-46374247 on Build GRCh38
rs1000094225 Chr.3: 46374473-46374473 on Build GRCh38
rs867834481 Chr.3: 46374049-46374049 on Build GRCh38
rs866115726 Chr.3: 46374302-46374302 on Build GRCh38
rs552382275 Chr.3: 46374243-46374243 on Build GRCh38
rs902385298 Chr.3: 46374491-46374491 on Build GRCh38
rs369515918 Chr.3: 46374460-46374460 on Build GRCh38
rs143589063 Chr.3: 46374324-46374324 on Build GRCh38
rs1035658784 Chr.3: 46374105-46374105 on Build GRCh38
rs950293317 Chr.3: 46374079-46374079 on Build GRCh38
rs146080174 Chr.3: 46374073-46374073 on Build GRCh38
rs763665714 Chr.3: 46374318-46374318 on Build GRCh38
rs1010142611 Chr.3: 46374143-46374143 on Build GRCh38
rs971625297 Chr.3: 46374328-46374328 on Build GRCh38
rs543618933 Chr.3: 46374419-46374419 on Build GRCh38
rs1017056005 Chr.3: 46374260-46374260 on Build GRCh38
rs1029273904 Chr.3: 46374225-46374225 on Build GRCh38
rs914611071 Chr.3: 46374443-46374443 on Build GRCh38
rs1001552690 Chr.3: 46374066-46374066 on Build GRCh38
rs1038296493 Chr.3: 46374307-46374307 on Build GRCh38
rs1042213202 Chr.3: 46374176-46374176 on Build GRCh38
rs575840249 Chr.3: 46374379-46374379 on Build GRCh38
rs1045742755 Chr.3: 46374092-46374092 on Build GRCh38
rs964152239 Chr.3: 46374213-46374213 on Build GRCh38
rs998083568 Chr.3: 46374216-46374216 on Build GRCh38
rs978988121 Chr.3: 46374173-46374173 on Build GRCh38
rs569458865 Chr.3: 46374146-46374146 on Build GRCh38
rs976456855 Chr.3: 46374235-46374235 on Build GRCh38
rs927305902 Chr.3: 46374114-46374114 on Build GRCh38
rs758790885 Chr.3: 46374495-46374495 on Build GRCh38
rs1010175310 Chr.3: 46374385-46374385 on Build GRCh38
rs927276137 Chr.3: 46374418-46374418 on Build GRCh38
rs1018526953 Chr.3: 46374483-46374483 on Build GRCh38
rs904450707 Chr.3: 46374447-46374447 on Build GRCh38
rs535892345 Chr.3: 46374322-46374322 on Build GRCh38
rs41414147 Chr.3: 46374251-46374251 on Build GRCh38
rs757414418 Chr.3: 46374391-46374391 on Build GRCh38
rs990172616 Chr.3: 46374437-46374437 on Build GRCh38
rs183840358 Chr.3: 46374198-46374198 on Build GRCh38
rs922268195 Chr.3: 46374241-46374241 on Build GRCh38
rs748287465 Chr.3: 46374236-46374236 on Build GRCh38
rs1054984862 Chr.3: 46374152-46374152 on Build GRCh38
rs112622745 Chr.3: 46374155-46374155 on Build GRCh38
rs1011121512 Chr.3: 46374166-46374166 on Build GRCh38
rs937434983 Chr.3: 46374330-46374330 on Build GRCh38
rs973283640 Chr.3: 46374290-46374290 on Build GRCh38
rs529752751 Chr.3: 46374070-46374070 on Build GRCh38
rs953749329 Chr.3: 46374234-46374234 on Build GRCh38
rs959996868 Chr.3: 46374116-46374116 on Build GRCh38
rs568368266 Chr.3: 46374293-46374293 on Build GRCh38

View Full Product Details