Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50214315-50214815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A3
Entrez Gene ID
10991
Gene Name
solute carrier family 38 member 3
Gene Aliases
G17, NAT1, SN1, SNAT3
UniGene
Hs.76460
Gene Chromosome Location
Chr.3: 50205246-50220979 on Build GRCh38
Transcripts
NM_006841.5 XM_006712954.2

SNP Detail

SNP ID Chromosome Location
rs574312037 Chr.3: 50214364-50214364 on Build GRCh38
rs967792427 Chr.3: 50214466-50214466 on Build GRCh38
rs752987140 Chr.3: 50214685-50214685 on Build GRCh38
rs1030706825 Chr.3: 50214518-50214518 on Build GRCh38
rs576447749 Chr.3: 50214449-50214449 on Build GRCh38
rs778946609 Chr.3: 50214730-50214730 on Build GRCh38
rs987101642 Chr.3: 50214597-50214597 on Build GRCh38
rs755175005 Chr.3: 50214347-50214347 on Build GRCh38
rs371453211 Chr.3: 50214594-50214594 on Build GRCh38
rs771271929 Chr.3: 50214634-50214634 on Build GRCh38
rs771785594 Chr.3: 50214396-50214396 on Build GRCh38
rs745824606 Chr.3: 50214418-50214418 on Build GRCh38
rs774726937 Chr.3: 50214673-50214673 on Build GRCh38
rs753793719 Chr.3: 50214727-50214727 on Build GRCh38
rs766824971 Chr.3: 50214425-50214425 on Build GRCh38
rs144414224 Chr.3: 50214775-50214775 on Build GRCh38
rs764252484 Chr.3: 50214706-50214706 on Build GRCh38
rs768278720 Chr.3: 50214631-50214631 on Build GRCh38
rs759842322 Chr.3: 50214674-50214674 on Build GRCh38
rs397812056 Chr.3: 50214403-50214404 on Build GRCh38
rs777713997 Chr.3: 50214494-50214494 on Build GRCh38
rs775172352 Chr.3: 50214405-50214405 on Build GRCh38
rs555078572 Chr.3: 50214416-50214416 on Build GRCh38
rs201228948 Chr.3: 50214400-50214400 on Build GRCh38
rs758306308 Chr.3: 50214732-50214732 on Build GRCh38
rs934880786 Chr.3: 50214498-50214498 on Build GRCh38
rs867433815 Chr.3: 50214415-50214415 on Build GRCh38
rs763488558 Chr.3: 50214410-50214410 on Build GRCh38
rs942848311 Chr.3: 50214605-50214605 on Build GRCh38
rs370248110 Chr.3: 50214611-50214611 on Build GRCh38
rs73835217 Chr.3: 50214593-50214593 on Build GRCh38
rs200407654 Chr.3: 50214399-50214399 on Build GRCh38
rs182598575 Chr.3: 50214610-50214610 on Build GRCh38
rs770830612 Chr.3: 50214389-50214389 on Build GRCh38
rs751845991 Chr.3: 50214344-50214344 on Build GRCh38
rs369567718 Chr.3: 50214483-50214483 on Build GRCh38
rs753874925 Chr.3: 50214500-50214500 on Build GRCh38
rs746729087 Chr.3: 50214769-50214769 on Build GRCh38
rs779900330 Chr.3: 50214617-50214617 on Build GRCh38
rs760289657 Chr.3: 50214341-50214351 on Build GRCh38
rs767840163 Chr.3: 50214682-50214682 on Build GRCh38
rs912819728 Chr.3: 50214598-50214598 on Build GRCh38
rs750366583 Chr.3: 50214731-50214731 on Build GRCh38
rs781260081 Chr.3: 50214348-50214348 on Build GRCh38
rs748156306 Chr.3: 50214349-50214349 on Build GRCh38
rs752897089 Chr.3: 50214452-50214452 on Build GRCh38
rs866210794 Chr.3: 50214434-50214434 on Build GRCh38
rs775991638 Chr.3: 50214632-50214632 on Build GRCh38
rs779990297 Chr.3: 50214768-50214768 on Build GRCh38
rs186974017 Chr.3: 50214791-50214791 on Build GRCh38
rs375618794 Chr.3: 50214700-50214700 on Build GRCh38
rs780758291 Chr.3: 50214790-50214790 on Build GRCh38
rs766620379 Chr.3: 50214337-50214337 on Build GRCh38
rs746628117 Chr.3: 50214618-50214618 on Build GRCh38
rs377107626 Chr.3: 50214515-50214515 on Build GRCh38
rs745529637 Chr.3: 50214394-50214394 on Build GRCh38
rs869255780 Chr.3: 50214404-50214405 on Build GRCh38
rs749153056 Chr.3: 50214384-50214384 on Build GRCh38
rs535248730 Chr.3: 50214406-50214406 on Build GRCh38
rs767684444 Chr.3: 50214451-50214451 on Build GRCh38
rs761340501 Chr.3: 50214619-50214619 on Build GRCh38
rs558686936 Chr.3: 50214580-50214580 on Build GRCh38
rs774007796 Chr.3: 50214392-50214392 on Build GRCh38
rs1012097925 Chr.3: 50214419-50214419 on Build GRCh38
rs372869563 Chr.3: 50214501-50214501 on Build GRCh38
rs896115593 Chr.3: 50214510-50214510 on Build GRCh38
rs867226595 Chr.3: 50214448-50214448 on Build GRCh38
rs924876703 Chr.3: 50214412-50214412 on Build GRCh38
rs756074239 Chr.3: 50214358-50214358 on Build GRCh38
rs776066457 Chr.3: 50214402-50214403 on Build GRCh38

View Full Product Details