Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52527910-52528404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NT5DC2
Entrez Gene ID
64943
Gene Name
5'-nucleotidase domain containing 2
Gene Aliases
-
UniGene
Hs.84753
Gene Chromosome Location
Chr.3: 52524369-52535077 on Build GRCh38
Transcripts
NM_001134231.1 NM_022908.2 XM_005265406.2 XM_006713303.3 XM_006713305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944366 c.675C>T p.F225F Chr.3: 52528279-52528279 on Build GRCh38
1047063 c.552C>A p.F184L Chr.3: 52528291-52528291 on Build GRCh38
1047062 c.691C>T p.L231F Chr.3: 52528043-52528043 on Build GRCh38
6730554 c.659C>T p.T220M Chr.3: 52528184-52528184 on Build GRCh38
731086 c.549G>A p.Q183Q Chr.3: 52528294-52528294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777201386 Chr.3: 52528210-52528210 on Build GRCh38
rs751747147 Chr.3: 52528331-52528332 on Build GRCh38
rs777335352 Chr.3: 52528115-52528115 on Build GRCh38
rs576338420 Chr.3: 52528070-52528070 on Build GRCh38
rs747643714 Chr.3: 52528022-52528022 on Build GRCh38
rs767918056 Chr.3: 52528080-52528080 on Build GRCh38
rs748054021 Chr.3: 52528355-52528355 on Build GRCh38
rs765292611 Chr.3: 52528058-52528058 on Build GRCh38
rs768295656 Chr.3: 52528350-52528350 on Build GRCh38
rs749231831 Chr.3: 52528276-52528276 on Build GRCh38
rs200698696 Chr.3: 52528352-52528352 on Build GRCh38
rs769775780 Chr.3: 52528251-52528251 on Build GRCh38
rs760884787 Chr.3: 52528302-52528302 on Build GRCh38
rs549530537 Chr.3: 52528281-52528281 on Build GRCh38
rs745499065 Chr.3: 52528238-52528238 on Build GRCh38
rs532284676 Chr.3: 52528284-52528284 on Build GRCh38
rs759599650 Chr.3: 52528054-52528054 on Build GRCh38
rs753888001 Chr.3: 52527957-52527957 on Build GRCh38
rs756486787 Chr.3: 52528203-52528203 on Build GRCh38
rs552491402 Chr.3: 52528339-52528339 on Build GRCh38
rs775427316 Chr.3: 52528059-52528059 on Build GRCh38
rs762941427 Chr.3: 52528183-52528183 on Build GRCh38
rs772754777 Chr.3: 52528038-52528038 on Build GRCh38
rs773483301 Chr.3: 52528294-52528294 on Build GRCh38
rs750168661 Chr.3: 52528340-52528340 on Build GRCh38
rs780489426 Chr.3: 52528013-52528013 on Build GRCh38
rs770688443 Chr.3: 52528134-52528134 on Build GRCh38
rs546135402 Chr.3: 52527980-52527980 on Build GRCh38
rs923671031 Chr.3: 52528128-52528128 on Build GRCh38
rs372686433 Chr.3: 52528346-52528346 on Build GRCh38
rs201535413 Chr.3: 52528232-52528232 on Build GRCh38
rs577577576 Chr.3: 52527966-52527966 on Build GRCh38
rs764118430 Chr.3: 52528184-52528184 on Build GRCh38
rs746544496 Chr.3: 52528214-52528214 on Build GRCh38
rs746663345 Chr.3: 52528039-52528039 on Build GRCh38
rs374478667 Chr.3: 52528063-52528063 on Build GRCh38
rs746684792 Chr.3: 52528124-52528124 on Build GRCh38
rs148169309 Chr.3: 52528068-52528068 on Build GRCh38
rs756757184 Chr.3: 52528216-52528216 on Build GRCh38
rs751744449 Chr.3: 52527989-52527989 on Build GRCh38
rs371707270 Chr.3: 52528160-52528160 on Build GRCh38
rs931381109 Chr.3: 52528012-52528012 on Build GRCh38
rs1047169669 Chr.3: 52528247-52528247 on Build GRCh38
rs1024618100 Chr.3: 52528025-52528025 on Build GRCh38
rs202071593 Chr.3: 52528195-52528195 on Build GRCh38
rs774466643 Chr.3: 52528185-52528185 on Build GRCh38
rs199782369 Chr.3: 52528365-52528365 on Build GRCh38
rs753194045 Chr.3: 52528338-52528338 on Build GRCh38
rs569551234 Chr.3: 52528283-52528283 on Build GRCh38
rs201941712 Chr.3: 52528155-52528155 on Build GRCh38
rs749113363 Chr.3: 52528349-52528349 on Build GRCh38
rs765774818 Chr.3: 52528333-52528333 on Build GRCh38
rs901416219 Chr.3: 52528143-52528144 on Build GRCh38
rs766697581 Chr.3: 52528310-52528310 on Build GRCh38
rs749748227 Chr.3: 52528099-52528099 on Build GRCh38
rs1056333700 Chr.3: 52528274-52528274 on Build GRCh38
rs759773944 Chr.3: 52528321-52528321 on Build GRCh38
rs776539707 Chr.3: 52528043-52528043 on Build GRCh38
rs758461013 Chr.3: 52527984-52527984 on Build GRCh38
rs776378305 Chr.3: 52528141-52528141 on Build GRCh38
rs759546240 Chr.3: 52527958-52527958 on Build GRCh38
rs377675801 Chr.3: 52527982-52527982 on Build GRCh38
rs981996462 Chr.3: 52527971-52527971 on Build GRCh38
rs375582198 Chr.3: 52527948-52527948 on Build GRCh38
rs756431676 Chr.3: 52528089-52528089 on Build GRCh38
rs780937578 Chr.3: 52528159-52528159 on Build GRCh38
rs375132600 Chr.3: 52528189-52528189 on Build GRCh38
rs994188437 Chr.3: 52527981-52527981 on Build GRCh38
rs1034971816 Chr.3: 52528285-52528285 on Build GRCh38
rs893547843 Chr.3: 52528222-52528222 on Build GRCh38
rs201139153 Chr.3: 52527949-52527949 on Build GRCh38
rs775345734 Chr.3: 52528182-52528182 on Build GRCh38
rs374674749 Chr.3: 52527993-52527993 on Build GRCh38
rs1020645125 Chr.3: 52528239-52528239 on Build GRCh38
rs750808882 Chr.3: 52528081-52528081 on Build GRCh38
rs200356286 Chr.3: 52528320-52528320 on Build GRCh38
rs757906160 Chr.3: 52528107-52528107 on Build GRCh38
rs193221218 Chr.3: 52527988-52527988 on Build GRCh38
rs200461145 Chr.3: 52528029-52528029 on Build GRCh38
rs878984979 Chr.3: 52528374-52528374 on Build GRCh38
rs780544513 Chr.3: 52528095-52528095 on Build GRCh38
rs757438583 Chr.3: 52528071-52528071 on Build GRCh38
rs367558577 Chr.3: 52528000-52528000 on Build GRCh38
rs966394705 Chr.3: 52528256-52528256 on Build GRCh38
rs774116030 Chr.3: 52528282-52528282 on Build GRCh38
rs868553212 Chr.3: 52528277-52528277 on Build GRCh38
rs754372952 Chr.3: 52528206-52528206 on Build GRCh38
rs766614377 Chr.3: 52528142-52528143 on Build GRCh38
rs1005720762 Chr.3: 52528268-52528268 on Build GRCh38
rs200504121 Chr.3: 52528002-52528002 on Build GRCh38
rs769724278 Chr.3: 52528006-52528006 on Build GRCh38
rs888527273 Chr.3: 52528262-52528262 on Build GRCh38
rs761832845 Chr.3: 52528188-52528188 on Build GRCh38
rs761580676 Chr.3: 52527974-52527975 on Build GRCh38
rs755482905 Chr.3: 52528207-52528207 on Build GRCh38
rs779866215 Chr.3: 52528270-52528270 on Build GRCh38
rs766755364 Chr.3: 52528204-52528204 on Build GRCh38

View Full Product Details