Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87240675-87241198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP2B
Entrez Gene ID
25978
Gene Name
charged multivesicular body protein 2B
Gene Aliases
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
UniGene
Hs.476930
Gene Chromosome Location
Chr.3: 87227263-87255548 on Build GRCh38
Transcripts
NM_001244644.1 NM_014043.3 XM_011533576.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413500 c.100G>T p.A34S Chr.3: 87240764-87240764 on Build GRCh38
109298 c.112C>A p.Q38K Chr.3: 87240776-87240776 on Build GRCh38
5490825 c.73C>T p.Q25* Chr.3: 87240737-87240737 on Build GRCh38
109883 c.115G>A p.E39K Chr.3: 87240779-87240779 on Build GRCh38
7351621 c.126+1G>A p.? Chr.3: 87240791-87240791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984655587 Chr.3: 87240878-87240878 on Build GRCh38
rs151007324 Chr.3: 87240779-87240779 on Build GRCh38
rs1037101360 Chr.3: 87241023-87241023 on Build GRCh38
rs772044188 Chr.3: 87240826-87240827 on Build GRCh38
rs769631869 Chr.3: 87240777-87240779 on Build GRCh38
rs781703226 Chr.3: 87240702-87240702 on Build GRCh38
rs1021378955 Chr.3: 87240748-87240748 on Build GRCh38
rs138886714 Chr.3: 87240728-87240728 on Build GRCh38
rs940077885 Chr.3: 87240982-87240982 on Build GRCh38
rs768113049 Chr.3: 87240845-87240845 on Build GRCh38
rs756803337 Chr.3: 87240772-87240772 on Build GRCh38
rs529384835 Chr.3: 87240881-87240881 on Build GRCh38
rs369912013 Chr.3: 87240794-87240794 on Build GRCh38
rs770968398 Chr.3: 87240729-87240729 on Build GRCh38
rs140349455 Chr.3: 87240865-87240865 on Build GRCh38
rs575545480 Chr.3: 87241041-87241041 on Build GRCh38
rs755177329 Chr.3: 87240803-87240803 on Build GRCh38
rs376846232 Chr.3: 87240775-87240775 on Build GRCh38
rs775142652 Chr.3: 87240754-87240754 on Build GRCh38
rs968948477 Chr.3: 87240777-87240777 on Build GRCh38
rs372974735 Chr.3: 87240801-87240801 on Build GRCh38
rs1007804469 Chr.3: 87240738-87240738 on Build GRCh38
rs748144379 Chr.3: 87240811-87240811 on Build GRCh38
rs781126895 Chr.3: 87240701-87240701 on Build GRCh38
rs1051723852 Chr.3: 87241122-87241122 on Build GRCh38
rs780237778 Chr.3: 87240719-87240719 on Build GRCh38
rs200322526 Chr.3: 87240720-87240720 on Build GRCh38
rs142645831 Chr.3: 87240776-87240776 on Build GRCh38
rs746197863 Chr.3: 87240703-87240703 on Build GRCh38
rs772337651 Chr.3: 87240840-87240840 on Build GRCh38
rs761359306 Chr.3: 87240887-87240887 on Build GRCh38
rs747894141 Chr.3: 87240706-87240709 on Build GRCh38
rs764648695 Chr.3: 87240835-87240836 on Build GRCh38
rs933115338 Chr.3: 87241064-87241064 on Build GRCh38
rs775755199 Chr.3: 87240827-87240828 on Build GRCh38
rs376241082 Chr.3: 87240812-87240812 on Build GRCh38
rs762064841 Chr.3: 87240844-87240844 on Build GRCh38
rs897525731 Chr.3: 87241081-87241081 on Build GRCh38
rs1013003315 Chr.3: 87241165-87241165 on Build GRCh38
rs956311381 Chr.3: 87241006-87241006 on Build GRCh38
rs544535734 Chr.3: 87241078-87241080 on Build GRCh38
rs866615436 Chr.3: 87241050-87241050 on Build GRCh38
rs916154916 Chr.3: 87240815-87240815 on Build GRCh38
rs552023510 Chr.3: 87241079-87241079 on Build GRCh38
rs559967286 Chr.3: 87240958-87240958 on Build GRCh38
rs1007401975 Chr.3: 87241141-87241141 on Build GRCh38
rs557194644 Chr.3: 87240710-87240710 on Build GRCh38
rs910438301 Chr.3: 87241074-87241074 on Build GRCh38
rs756409160 Chr.3: 87240780-87240780 on Build GRCh38
rs746162849 Chr.3: 87240782-87240785 on Build GRCh38
rs941930125 Chr.3: 87241077-87241077 on Build GRCh38
rs750983989 Chr.3: 87240778-87240778 on Build GRCh38
rs771533082 Chr.3: 87240901-87240904 on Build GRCh38
rs774383512 Chr.3: 87240777-87240780 on Build GRCh38
rs570281214 Chr.3: 87241084-87241084 on Build GRCh38
rs548963118 Chr.3: 87241132-87241132 on Build GRCh38
rs63750818 Chr.3: 87240749-87240749 on Build GRCh38
rs1043254503 Chr.3: 87241148-87241148 on Build GRCh38
rs959678900 Chr.3: 87240755-87240755 on Build GRCh38
rs922627010 Chr.3: 87241038-87241038 on Build GRCh38
rs139894940 Chr.3: 87240752-87240752 on Build GRCh38
rs761243838 Chr.3: 87240829-87240830 on Build GRCh38
rs776778264 Chr.3: 87240735-87240735 on Build GRCh38
rs762547677 Chr.3: 87240756-87240756 on Build GRCh38
rs185484639 Chr.3: 87241100-87241100 on Build GRCh38
rs374428152 Chr.3: 87240759-87240759 on Build GRCh38
rs763615051 Chr.3: 87240758-87240758 on Build GRCh38
rs779243164 Chr.3: 87240810-87240810 on Build GRCh38
rs541565598 Chr.3: 87240924-87240924 on Build GRCh38
rs985975960 Chr.3: 87241065-87241065 on Build GRCh38

View Full Product Details