Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 97243859-97244361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
UniGene
Hs.272208
Gene Chromosome Location
Chr.3: 96814581-97761532 on Build GRCh38
Transcripts
NM_001080448.2 XM_006713592.3 XM_017006210.1 XM_017006211.1 XM_017006212.1 XM_017006213.1 XM_017006214.1 XM_017006218.1 XM_017006219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6876731 c.1522G>A p.E508K Chr.3: 97244203-97244203 on Build GRCh38
5733192 c.1355G>T p.G452V Chr.3: 97244036-97244036 on Build GRCh38
1049027 c.1196C>A p.S399Y Chr.3: 97244159-97244159 on Build GRCh38
5400105 c.1072G>A p.G358R Chr.3: 97244035-97244035 on Build GRCh38
6655039 c.1514A>G p.Y505C Chr.3: 97244195-97244195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374566449 Chr.3: 97244188-97244188 on Build GRCh38
rs767590710 Chr.3: 97244110-97244110 on Build GRCh38
rs773321167 Chr.3: 97244305-97244305 on Build GRCh38
rs779311285 Chr.3: 97244298-97244298 on Build GRCh38
rs778587736 Chr.3: 97243993-97243993 on Build GRCh38
rs746371126 Chr.3: 97244172-97244172 on Build GRCh38
rs771073897 Chr.3: 97244320-97244320 on Build GRCh38
rs201738877 Chr.3: 97243988-97243988 on Build GRCh38
rs756759339 Chr.3: 97244161-97244161 on Build GRCh38
rs763227720 Chr.3: 97244089-97244089 on Build GRCh38
rs917730920 Chr.3: 97243991-97243991 on Build GRCh38
rs752702302 Chr.3: 97244247-97244247 on Build GRCh38
rs761743970 Chr.3: 97243896-97243896 on Build GRCh38
rs368962314 Chr.3: 97244001-97244001 on Build GRCh38
rs753806867 Chr.3: 97243959-97243959 on Build GRCh38
rs565852920 Chr.3: 97244099-97244099 on Build GRCh38
rs924630409 Chr.3: 97244060-97244060 on Build GRCh38
rs569601978 Chr.3: 97243910-97243910 on Build GRCh38
rs775773153 Chr.3: 97244074-97244074 on Build GRCh38
rs762039514 Chr.3: 97244196-97244196 on Build GRCh38
rs746436436 Chr.3: 97243897-97243897 on Build GRCh38
rs970608101 Chr.3: 97243894-97243894 on Build GRCh38
rs187331112 Chr.3: 97244277-97244277 on Build GRCh38
rs756618750 Chr.3: 97244265-97244265 on Build GRCh38
rs371358724 Chr.3: 97244176-97244176 on Build GRCh38
rs762796650 Chr.3: 97244251-97244251 on Build GRCh38
rs752475387 Chr.3: 97244145-97244145 on Build GRCh38
rs770135644 Chr.3: 97244067-97244067 on Build GRCh38
rs751700923 Chr.3: 97243912-97243912 on Build GRCh38
rs914858890 Chr.3: 97244224-97244224 on Build GRCh38
rs773234297 Chr.3: 97244205-97244205 on Build GRCh38
rs780859786 Chr.3: 97244164-97244164 on Build GRCh38
rs189103500 Chr.3: 97244018-97244018 on Build GRCh38
rs777388224 Chr.3: 97244038-97244038 on Build GRCh38
rs756027235 Chr.3: 97243930-97243930 on Build GRCh38
rs761845354 Chr.3: 97244109-97244109 on Build GRCh38
rs1026644477 Chr.3: 97244329-97244329 on Build GRCh38
rs757025107 Chr.3: 97244260-97244260 on Build GRCh38
rs777518741 Chr.3: 97244151-97244151 on Build GRCh38
rs891263215 Chr.3: 97244274-97244274 on Build GRCh38
rs762996903 Chr.3: 97244324-97244324 on Build GRCh38
rs758045778 Chr.3: 97244148-97244148 on Build GRCh38
rs775222679 Chr.3: 97244321-97244321 on Build GRCh38
rs780843097 Chr.3: 97244263-97244263 on Build GRCh38
rs200900494 Chr.3: 97244187-97244187 on Build GRCh38
rs9835140 Chr.3: 97244160-97244160 on Build GRCh38
rs866564290 Chr.3: 97244190-97244190 on Build GRCh38
rs55757053 Chr.3: 97244259-97244259 on Build GRCh38
rs369086136 Chr.3: 97244070-97244070 on Build GRCh38
rs190532302 Chr.3: 97244325-97244325 on Build GRCh38
rs748366898 Chr.3: 97244300-97244300 on Build GRCh38
rs373819855 Chr.3: 97244111-97244111 on Build GRCh38
rs756680143 Chr.3: 97244303-97244303 on Build GRCh38
rs535111359 Chr.3: 97244022-97244022 on Build GRCh38
rs765136384 Chr.3: 97244217-97244217 on Build GRCh38
rs780388246 Chr.3: 97244266-97244266 on Build GRCh38
rs971529766 Chr.3: 97244047-97244047 on Build GRCh38
rs755394695 Chr.3: 97244138-97244138 on Build GRCh38
rs376836753 Chr.3: 97243907-97243907 on Build GRCh38
rs373391182 Chr.3: 97244049-97244049 on Build GRCh38
rs181777860 Chr.3: 97244035-97244035 on Build GRCh38
rs774582004 Chr.3: 97244105-97244105 on Build GRCh38
rs973039162 Chr.3: 97243958-97243958 on Build GRCh38
rs536651965 Chr.3: 97244055-97244055 on Build GRCh38
rs549221199 Chr.3: 97244061-97244061 on Build GRCh38
rs760622374 Chr.3: 97244318-97244318 on Build GRCh38
rs1392696 Chr.3: 97243889-97243889 on Build GRCh38
rs534785975 Chr.3: 97244125-97244125 on Build GRCh38
rs763984183 Chr.3: 97244255-97244255 on Build GRCh38
rs766259314 Chr.3: 97243946-97243946 on Build GRCh38
rs370143584 Chr.3: 97244276-97244276 on Build GRCh38
rs751963745 Chr.3: 97244118-97244118 on Build GRCh38
rs759485796 Chr.3: 97244212-97244212 on Build GRCh38
rs767572244 Chr.3: 97244204-97244204 on Build GRCh38

View Full Product Details