Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112102600-112103101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf52
Entrez Gene ID
79669
Gene Name
chromosome 3 open reading frame 52
Gene Aliases
TTMP
UniGene
Hs.434247
Gene Chromosome Location
Chr.3: 112086335-112131095 on Build GRCh38
Transcripts
NM_001171747.1 NM_024616.2 XM_011513133.2 XM_017007181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1593213 c.327C>A p.F109L Chr.3: 112102896-112102896 on Build GRCh38
1036335 c.391G>A p.E131K Chr.3: 112102960-112102960 on Build GRCh38
1036334 c.336G>T p.M112I Chr.3: 112102905-112102905 on Build GRCh38
1593212 c.391G>A p.E131K Chr.3: 112102960-112102960 on Build GRCh38
6403773 c.301C>T p.L101F Chr.3: 112102870-112102870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577670634 Chr.3: 112102712-112102712 on Build GRCh38
rs767735572 Chr.3: 112102878-112102878 on Build GRCh38
rs986748869 Chr.3: 112103069-112103069 on Build GRCh38
rs369573804 Chr.3: 112103031-112103031 on Build GRCh38
rs1001906776 Chr.3: 112102643-112102643 on Build GRCh38
rs761898694 Chr.3: 112102994-112102994 on Build GRCh38
rs1009727610 Chr.3: 112102680-112102680 on Build GRCh38
rs954037754 Chr.3: 112102762-112102762 on Build GRCh38
rs751661933 Chr.3: 112102869-112102869 on Build GRCh38
rs145975333 Chr.3: 112102793-112102793 on Build GRCh38
rs368954149 Chr.3: 112102841-112102841 on Build GRCh38
rs776274687 Chr.3: 112102861-112102861 on Build GRCh38
rs766485401 Chr.3: 112103010-112103010 on Build GRCh38
rs778603271 Chr.3: 112102822-112102822 on Build GRCh38
rs960972235 Chr.3: 112102671-112102671 on Build GRCh38
rs192095 Chr.3: 112103016-112103016 on Build GRCh38
rs769754678 Chr.3: 112102978-112102978 on Build GRCh38
rs199685620 Chr.3: 112102959-112102959 on Build GRCh38
rs773164886 Chr.3: 112103001-112103001 on Build GRCh38
rs561508717 Chr.3: 112102673-112102673 on Build GRCh38
rs775343079 Chr.3: 112102988-112102988 on Build GRCh38
rs771869744 Chr.3: 112102827-112102827 on Build GRCh38
rs1049302495 Chr.3: 112102628-112102628 on Build GRCh38
rs759202200 Chr.3: 112102969-112102969 on Build GRCh38
rs769035694 Chr.3: 112102821-112102821 on Build GRCh38
rs201080279 Chr.3: 112102961-112102961 on Build GRCh38
rs538858661 Chr.3: 112102774-112102774 on Build GRCh38
rs776539390 Chr.3: 112102966-112102966 on Build GRCh38
rs747690830 Chr.3: 112102943-112102943 on Build GRCh38
rs570484708 Chr.3: 112102754-112102754 on Build GRCh38
rs16859188 Chr.3: 112102642-112102642 on Build GRCh38
rs1021094237 Chr.3: 112102687-112102687 on Build GRCh38
rs202233429 Chr.3: 112102889-112102889 on Build GRCh38
rs760391515 Chr.3: 112102850-112102850 on Build GRCh38
rs972364169 Chr.3: 112102676-112102676 on Build GRCh38
rs550403950 Chr.3: 112102738-112102738 on Build GRCh38
rs749866717 Chr.3: 112102818-112102818 on Build GRCh38
rs1048798470 Chr.3: 112102929-112102929 on Build GRCh38
rs79246340 Chr.3: 112102649-112102649 on Build GRCh38
rs1026572260 Chr.3: 112102968-112102968 on Build GRCh38
rs139531193 Chr.3: 112102864-112102864 on Build GRCh38
rs769915876 Chr.3: 112103050-112103050 on Build GRCh38
rs954151636 Chr.3: 112103049-112103049 on Build GRCh38
rs754119338 Chr.3: 112102902-112102902 on Build GRCh38
rs930809485 Chr.3: 112102755-112102755 on Build GRCh38
rs149820783 Chr.3: 112102935-112102935 on Build GRCh38
rs780441227 Chr.3: 112102803-112102803 on Build GRCh38
rs778046236 Chr.3: 112102881-112102881 on Build GRCh38
rs545126644 Chr.3: 112102795-112102795 on Build GRCh38
rs1049137040 Chr.3: 112102767-112102767 on Build GRCh38
rs1001985902 Chr.3: 112102951-112102951 on Build GRCh38
rs746731235 Chr.3: 112102960-112102960 on Build GRCh38
rs16859190 Chr.3: 112102900-112102900 on Build GRCh38
rs968640181 Chr.3: 112102710-112102710 on Build GRCh38
rs755553530 Chr.3: 112102925-112102925 on Build GRCh38
rs930483042 Chr.3: 112102907-112102907 on Build GRCh38
rs747635747 Chr.3: 112102825-112102825 on Build GRCh38
rs373724900 Chr.3: 112102998-112102998 on Build GRCh38
rs772958240 Chr.3: 112102845-112102845 on Build GRCh38
rs779315208 Chr.3: 112102888-112102888 on Build GRCh38
rs761845275 Chr.3: 112102870-112102870 on Build GRCh38
rs781343233 Chr.3: 112102792-112102792 on Build GRCh38
rs1022121422 Chr.3: 112102903-112102903 on Build GRCh38
rs770699957 Chr.3: 112102856-112102856 on Build GRCh38
rs868790351 Chr.3: 112102739-112102739 on Build GRCh38
rs919398165 Chr.3: 112102688-112102688 on Build GRCh38
rs771640919 Chr.3: 112102955-112102955 on Build GRCh38
rs367805861 Chr.3: 112103030-112103030 on Build GRCh38
rs562483515 Chr.3: 112102666-112102666 on Build GRCh38
rs530667987 Chr.3: 112102715-112102715 on Build GRCh38
rs548234043 Chr.3: 112102650-112102650 on Build GRCh38

View Full Product Details