Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112169034-112169475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9C1
Entrez Gene ID
285335
Gene Name
solute carrier family 9 member C1
Gene Aliases
NHE, NHE-10, SLC9A10, sperm-NHE
UniGene
Hs.680112
Gene Chromosome Location
Chr.3: 112140898-112294258 on Build GRCh38
Transcripts
NM_001320531.1 NM_183061.2 NR_135297.1 XM_006713603.2 XM_011512718.1 XM_011512719.1 XM_011512720.1 XM_011512721.1 XM_011512725.2 XM_017006246.1 XM_017006247.1 XM_017006248.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4932763 c.3042A>G p.L1014L Chr.3: 112169206-112169206 on Build GRCh38
7155318 c.2989_2990insA p.M997fs*21 Chr.3: 112169258-112169259 on Build GRCh38
1616718 c.3037C>T p.H1013Y Chr.3: 112169211-112169211 on Build GRCh38
3585361 c.2971C>T p.P991S Chr.3: 112169277-112169277 on Build GRCh38
3585360 c.3056G>A p.W1019* Chr.3: 112169058-112169058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765897852 Chr.3: 112169077-112169077 on Build GRCh38
rs762617673 Chr.3: 112169315-112169315 on Build GRCh38
rs748426980 Chr.3: 112169269-112169269 on Build GRCh38
rs772601922 Chr.3: 112169146-112169146 on Build GRCh38
rs371207483 Chr.3: 112169344-112169344 on Build GRCh38
rs548578146 Chr.3: 112169245-112169245 on Build GRCh38
rs185671757 Chr.3: 112169346-112169346 on Build GRCh38
rs774982831 Chr.3: 112169372-112169372 on Build GRCh38
rs758268069 Chr.3: 112169237-112169237 on Build GRCh38
rs764639667 Chr.3: 112169208-112169208 on Build GRCh38
rs752787219 Chr.3: 112169342-112169342 on Build GRCh38
rs778316784 Chr.3: 112169276-112169276 on Build GRCh38
rs766872135 Chr.3: 112169188-112169188 on Build GRCh38
rs373390617 Chr.3: 112169095-112169095 on Build GRCh38
rs781126177 Chr.3: 112169239-112169239 on Build GRCh38
rs748807413 Chr.3: 112169092-112169093 on Build GRCh38
rs374547341 Chr.3: 112169238-112169238 on Build GRCh38
rs1034132782 Chr.3: 112169392-112169392 on Build GRCh38
rs748372026 Chr.3: 112169101-112169101 on Build GRCh38
rs193250654 Chr.3: 112169331-112169331 on Build GRCh38
rs376063047 Chr.3: 112169378-112169378 on Build GRCh38
rs1054571244 Chr.3: 112169299-112169299 on Build GRCh38
rs774960236 Chr.3: 112169313-112169313 on Build GRCh38
rs780026426 Chr.3: 112169062-112169062 on Build GRCh38
rs770931445 Chr.3: 112169100-112169101 on Build GRCh38
rs572919256 Chr.3: 112169388-112169388 on Build GRCh38
rs548267063 Chr.3: 112169117-112169117 on Build GRCh38
rs755147870 Chr.3: 112169255-112169255 on Build GRCh38
rs140783319 Chr.3: 112169283-112169283 on Build GRCh38
rs778735105 Chr.3: 112169112-112169113 on Build GRCh38
rs769386846 Chr.3: 112169305-112169305 on Build GRCh38
rs144423530 Chr.3: 112169290-112169290 on Build GRCh38
rs377699671 Chr.3: 112169325-112169325 on Build GRCh38
rs759956304 Chr.3: 112169297-112169297 on Build GRCh38
rs534678683 Chr.3: 112169205-112169205 on Build GRCh38
rs761725218 Chr.3: 112169324-112169324 on Build GRCh38
rs912895640 Chr.3: 112169132-112169132 on Build GRCh38
rs771359542 Chr.3: 112169284-112169284 on Build GRCh38
rs755003295 Chr.3: 112169421-112169425 on Build GRCh38
rs772368591 Chr.3: 112169270-112169270 on Build GRCh38
rs931706432 Chr.3: 112169121-112169124 on Build GRCh38
rs762389373 Chr.3: 112169374-112169374 on Build GRCh38
rs147426717 Chr.3: 112169433-112169433 on Build GRCh38
rs747221159 Chr.3: 112169353-112169353 on Build GRCh38
rs771397210 Chr.3: 112169354-112169354 on Build GRCh38
rs751367471 Chr.3: 112169317-112169317 on Build GRCh38
rs1003308575 Chr.3: 112169389-112169390 on Build GRCh38
rs752494083 Chr.3: 112169078-112169078 on Build GRCh38
rs973558102 Chr.3: 112169431-112169431 on Build GRCh38
rs567972277 Chr.3: 112169266-112169266 on Build GRCh38
rs945664536 Chr.3: 112169156-112169156 on Build GRCh38
rs773686467 Chr.3: 112169169-112169169 on Build GRCh38
rs202130918 Chr.3: 112169316-112169316 on Build GRCh38
rs552373722 Chr.3: 112169080-112169080 on Build GRCh38
rs779154896 Chr.3: 112169340-112169340 on Build GRCh38
rs761288932 Chr.3: 112169170-112169170 on Build GRCh38
rs753557816 Chr.3: 112169093-112169093 on Build GRCh38
rs752414698 Chr.3: 112169217-112169217 on Build GRCh38
rs566189211 Chr.3: 112169115-112169115 on Build GRCh38
rs755981319 Chr.3: 112169335-112169335 on Build GRCh38
rs758017227 Chr.3: 112169230-112169230 on Build GRCh38
rs763928111 Chr.3: 112169233-112169233 on Build GRCh38
rs771801213 Chr.3: 112169354-112169358 on Build GRCh38
rs866280027 Chr.3: 112169406-112169406 on Build GRCh38
rs745779841 Chr.3: 112169242-112169242 on Build GRCh38
rs1001429992 Chr.3: 112169386-112169386 on Build GRCh38
rs745633514 Chr.3: 112169200-112169204 on Build GRCh38
rs768773809 Chr.3: 112169090-112169090 on Build GRCh38
rs552774208 Chr.3: 112169365-112169365 on Build GRCh38
rs777488067 Chr.3: 112169075-112169076 on Build GRCh38
rs781384284 Chr.3: 112169362-112169362 on Build GRCh38

View Full Product Details