Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119876224-119876646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSK3B
Entrez Gene ID
2932
Gene Name
glycogen synthase kinase 3 beta
Gene Aliases
-
UniGene
Hs.445733
Gene Chromosome Location
Chr.3: 119821321-120094417 on Build GRCh38
Transcripts
NM_001146156.1 NM_002093.3 XM_006713610.2 XM_006713611.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036816 c.903G>A p.W301* Chr.3: 119876419-119876419 on Build GRCh38
7315955 c.902G>C p.W301S Chr.3: 119876420-119876420 on Build GRCh38
6424956 c.881C>T p.P294L Chr.3: 119876441-119876441 on Build GRCh38
6926317 c.856C>T p.P286S Chr.3: 119876466-119876466 on Build GRCh38
6433798 c.817C>A p.L273M Chr.3: 119876505-119876505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043649183 Chr.3: 119876583-119876583 on Build GRCh38
rs202229925 Chr.3: 119876536-119876536 on Build GRCh38
rs568223760 Chr.3: 119876487-119876487 on Build GRCh38
rs767276205 Chr.3: 119876369-119876369 on Build GRCh38
rs535206260 Chr.3: 119876546-119876546 on Build GRCh38
rs137906892 Chr.3: 119876335-119876335 on Build GRCh38
rs754511299 Chr.3: 119876545-119876545 on Build GRCh38
rs949789694 Chr.3: 119876250-119876251 on Build GRCh38
rs750822479 Chr.3: 119876531-119876531 on Build GRCh38
rs765995720 Chr.3: 119876539-119876539 on Build GRCh38
rs778753553 Chr.3: 119876452-119876452 on Build GRCh38
rs553440109 Chr.3: 119876608-119876608 on Build GRCh38
rs753356510 Chr.3: 119876544-119876544 on Build GRCh38
rs757738148 Chr.3: 119876365-119876365 on Build GRCh38
rs774789474 Chr.3: 119876523-119876523 on Build GRCh38
rs771909190 Chr.3: 119876466-119876466 on Build GRCh38
rs945932989 Chr.3: 119876541-119876541 on Build GRCh38
rs201102477 Chr.3: 119876387-119876387 on Build GRCh38
rs751932713 Chr.3: 119876363-119876363 on Build GRCh38
rs1047370019 Chr.3: 119876319-119876319 on Build GRCh38
rs762372227 Chr.3: 119876394-119876394 on Build GRCh38
rs72548704 Chr.3: 119876393-119876394 on Build GRCh38
rs959999532 Chr.3: 119876621-119876621 on Build GRCh38
rs750160085 Chr.3: 119876379-119876379 on Build GRCh38
rs200373768 Chr.3: 119876490-119876490 on Build GRCh38
rs775594725 Chr.3: 119876507-119876507 on Build GRCh38
rs904762112 Chr.3: 119876596-119876596 on Build GRCh38
rs762257490 Chr.3: 119876528-119876528 on Build GRCh38
rs373688856 Chr.3: 119876561-119876561 on Build GRCh38
rs920504945 Chr.3: 119876270-119876270 on Build GRCh38
rs778764261 Chr.3: 119876253-119876253 on Build GRCh38
rs755845247 Chr.3: 119876384-119876384 on Build GRCh38
rs769178134 Chr.3: 119876521-119876521 on Build GRCh38
rs752570079 Chr.3: 119876560-119876560 on Build GRCh38
rs552556881 Chr.3: 119876261-119876261 on Build GRCh38
rs549729441 Chr.3: 119876437-119876437 on Build GRCh38
rs891702710 Chr.3: 119876479-119876479 on Build GRCh38
rs973684712 Chr.3: 119876265-119876265 on Build GRCh38
rs944390909 Chr.3: 119876364-119876364 on Build GRCh38
rs926958167 Chr.3: 119876257-119876263 on Build GRCh38
rs767936850 Chr.3: 119876529-119876529 on Build GRCh38
rs745433639 Chr.3: 119876285-119876285 on Build GRCh38
rs747930041 Chr.3: 119876455-119876455 on Build GRCh38
rs901642342 Chr.3: 119876564-119876564 on Build GRCh38
rs762995497 Chr.3: 119876508-119876508 on Build GRCh38
rs570662743 Chr.3: 119876278-119876278 on Build GRCh38
rs997021211 Chr.3: 119876573-119876573 on Build GRCh38
rs773000672 Chr.3: 119876477-119876477 on Build GRCh38
rs190878300 Chr.3: 119876614-119876614 on Build GRCh38
rs770038325 Chr.3: 119876489-119876489 on Build GRCh38
rs769275172 Chr.3: 119876406-119876406 on Build GRCh38
rs369346892 Chr.3: 119876388-119876388 on Build GRCh38
rs760354315 Chr.3: 119876533-119876533 on Build GRCh38

View Full Product Details