Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134191718-134192261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RYK
Entrez Gene ID
6259
Gene Name
receptor-like tyrosine kinase
Gene Aliases
D3S3195, JTK5, JTK5A, RYK1
UniGene
Hs.654562
Gene Chromosome Location
Chr.3: 134157133-134250834 on Build GRCh38
Transcripts
NM_001005861.2 NM_002958.3 XM_017007016.1

SNP Detail

SNP ID Chromosome Location
rs375652759 Chr.3: 134191966-134191966 on Build GRCh38
rs372101827 Chr.3: 134191923-134191923 on Build GRCh38
rs1026934980 Chr.3: 134192052-134192052 on Build GRCh38
rs778701959 Chr.3: 134191916-134191916 on Build GRCh38
rs912050878 Chr.3: 134191783-134191783 on Build GRCh38
rs370703140 Chr.3: 134191858-134191858 on Build GRCh38
rs978703231 Chr.3: 134192206-134192206 on Build GRCh38
rs147089787 Chr.3: 134191829-134191829 on Build GRCh38
rs988892691 Chr.3: 134192141-134192141 on Build GRCh38
rs562169484 Chr.3: 134191992-134191992 on Build GRCh38
rs1042591532 Chr.3: 134191769-134191769 on Build GRCh38
rs965232307 Chr.3: 134192129-134192129 on Build GRCh38
rs1036775428 Chr.3: 134191935-134191935 on Build GRCh38
rs762145212 Chr.3: 134191958-134191958 on Build GRCh38
rs765653693 Chr.3: 134191959-134191959 on Build GRCh38
rs912870149 Chr.3: 134192065-134192065 on Build GRCh38
rs780654400 Chr.3: 134191878-134191878 on Build GRCh38
rs764210355 Chr.3: 134191845-134191845 on Build GRCh38
rs772141154 Chr.3: 134191837-134191837 on Build GRCh38
rs953929849 Chr.3: 134191789-134191789 on Build GRCh38
rs777289155 Chr.3: 134191889-134191889 on Build GRCh38
rs774206383 Chr.3: 134191828-134191828 on Build GRCh38
rs554508310 Chr.3: 134191787-134191787 on Build GRCh38
rs762204556 Chr.3: 134191862-134191862 on Build GRCh38
rs79798304 Chr.3: 134192176-134192176 on Build GRCh38
rs769297012 Chr.3: 134191802-134191802 on Build GRCh38
rs563051019 Chr.3: 134192120-134192120 on Build GRCh38
rs76803140 Chr.3: 134192154-134192154 on Build GRCh38
rs756934212 Chr.3: 134192027-134192027 on Build GRCh38
rs770656029 Chr.3: 134191913-134191913 on Build GRCh38
rs1039573879 Chr.3: 134192182-134192182 on Build GRCh38
rs1004797021 Chr.3: 134192235-134192235 on Build GRCh38
rs575814639 Chr.3: 134192044-134192044 on Build GRCh38
rs937537260 Chr.3: 134191900-134191900 on Build GRCh38
rs542402568 Chr.3: 134191973-134191973 on Build GRCh38
rs61732795 Chr.3: 134191925-134191925 on Build GRCh38
rs985687139 Chr.3: 134191790-134191790 on Build GRCh38
rs1006744267 Chr.3: 134192069-134192069 on Build GRCh38
rs755514899 Chr.3: 134191976-134191976 on Build GRCh38
rs912662449 Chr.3: 134191795-134191795 on Build GRCh38
rs765713202 Chr.3: 134191865-134191865 on Build GRCh38
rs766785792 Chr.3: 134191971-134191971 on Build GRCh38
rs768783642 Chr.3: 134191931-134191931 on Build GRCh38
rs755710425 Chr.3: 134191888-134191888 on Build GRCh38
rs927586515 Chr.3: 134192209-134192209 on Build GRCh38
rs138457073 Chr.3: 134192123-134192123 on Build GRCh38
rs763428713 Chr.3: 134191967-134191967 on Build GRCh38
rs1049092275 Chr.3: 134192229-134192229 on Build GRCh38
rs777731623 Chr.3: 134192019-134192024 on Build GRCh38
rs995457362 Chr.3: 134191750-134191750 on Build GRCh38
rs748945264 Chr.3: 134191906-134191906 on Build GRCh38
rs879039110 Chr.3: 134192191-134192191 on Build GRCh38
rs537570512 Chr.3: 134191797-134191797 on Build GRCh38
rs577277523 Chr.3: 134191839-134191839 on Build GRCh38
rs573507958 Chr.3: 134191955-134191955 on Build GRCh38
rs368346060 Chr.3: 134191880-134191880 on Build GRCh38
rs966980576 Chr.3: 134192111-134192111 on Build GRCh38
rs770776836 Chr.3: 134191813-134191813 on Build GRCh38
rs776932307 Chr.3: 134191946-134191946 on Build GRCh38
rs989848789 Chr.3: 134192014-134192014 on Build GRCh38
rs922059073 Chr.3: 134192211-134192211 on Build GRCh38
rs568525826 Chr.3: 134191792-134191792 on Build GRCh38
rs957306784 Chr.3: 134192005-134192005 on Build GRCh38
rs915495933 Chr.3: 134191747-134191747 on Build GRCh38
rs753441191 Chr.3: 134192001-134192001 on Build GRCh38
rs775554119 Chr.3: 134191927-134191927 on Build GRCh38
rs7430200 Chr.3: 134192153-134192153 on Build GRCh38
rs759001453 Chr.3: 134191870-134191870 on Build GRCh38
rs886371658 Chr.3: 134191856-134191856 on Build GRCh38
rs543077122 Chr.3: 134192112-134192112 on Build GRCh38
rs945584737 Chr.3: 134191751-134191751 on Build GRCh38
rs886425773 Chr.3: 134192231-134192231 on Build GRCh38
rs772963317 Chr.3: 134191806-134191806 on Build GRCh38
rs375133082 Chr.3: 134191877-134191877 on Build GRCh38
rs528410512 Chr.3: 134192220-134192220 on Build GRCh38
rs55965934 Chr.3: 134191843-134191843 on Build GRCh38
rs1001726536 Chr.3: 134191960-134191960 on Build GRCh38

View Full Product Details