Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 137764485-137765025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOX14
Entrez Gene ID
8403
Gene Name
SRY-box 14
Gene Aliases
SOX28
UniGene
Hs.248184
Gene Chromosome Location
Chr.3: 137764292-137766334 on Build GRCh38
Transcripts
NM_004189.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3123648 c.241C>T p.R81C Chr.3: 137765025-137765025 on Build GRCh38
3123642 c.132C>T p.A44A Chr.3: 137764916-137764916 on Build GRCh38
728209 c.11C>A p.P4H Chr.3: 137764795-137764795 on Build GRCh38
728208 c.53G>T p.W18L Chr.3: 137764837-137764837 on Build GRCh38
5042096 c.197C>T p.A66V Chr.3: 137764981-137764981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005897070 Chr.3: 137764609-137764609 on Build GRCh38
rs143465510 Chr.3: 137764971-137764971 on Build GRCh38
rs996983460 Chr.3: 137764678-137764678 on Build GRCh38
rs549710502 Chr.3: 137764733-137764733 on Build GRCh38
rs188758359 Chr.3: 137764767-137764767 on Build GRCh38
rs868626819 Chr.3: 137764673-137764673 on Build GRCh38
rs375023583 Chr.3: 137764846-137764846 on Build GRCh38
rs929022310 Chr.3: 137764565-137764565 on Build GRCh38
rs767450094 Chr.3: 137764784-137764784 on Build GRCh38
rs200954864 Chr.3: 137764823-137764823 on Build GRCh38
rs573524222 Chr.3: 137764661-137764661 on Build GRCh38
rs891398867 Chr.3: 137764958-137764958 on Build GRCh38
rs141165723 Chr.3: 137764518-137764518 on Build GRCh38
rs903204838 Chr.3: 137764774-137764774 on Build GRCh38
rs779051127 Chr.3: 137764881-137764881 on Build GRCh38
rs555495881 Chr.3: 137764692-137764692 on Build GRCh38
rs1056127758 Chr.3: 137764512-137764512 on Build GRCh38
rs148372198 Chr.3: 137765000-137765000 on Build GRCh38
rs1047403302 Chr.3: 137764989-137764989 on Build GRCh38
rs539847475 Chr.3: 137764599-137764599 on Build GRCh38
rs35434838 Chr.3: 137764828-137764829 on Build GRCh38
rs746167210 Chr.3: 137764766-137764766 on Build GRCh38
rs757336135 Chr.3: 137764855-137764855 on Build GRCh38
rs747585370 Chr.3: 137764769-137764769 on Build GRCh38
rs201290750 Chr.3: 137764772-137764772 on Build GRCh38
rs371601436 Chr.3: 137764776-137764776 on Build GRCh38
rs760715874 Chr.3: 137764817-137764817 on Build GRCh38
rs568330763 Chr.3: 137764529-137764529 on Build GRCh38
rs955647643 Chr.3: 137764701-137764701 on Build GRCh38
rs192855180 Chr.3: 137764851-137764851 on Build GRCh38
rs148822399 Chr.3: 137764950-137764950 on Build GRCh38
rs1033072111 Chr.3: 137764610-137764610 on Build GRCh38
rs929196690 Chr.3: 137764943-137764943 on Build GRCh38
rs553373486 Chr.3: 137764605-137764605 on Build GRCh38
rs1016234334 Chr.3: 137764990-137764990 on Build GRCh38
rs542018425 Chr.3: 137764664-137764664 on Build GRCh38
rs747454336 Chr.3: 137764946-137764946 on Build GRCh38
rs201718 Chr.3: 137764520-137764520 on Build GRCh38
rs1014712604 Chr.3: 137764528-137764528 on Build GRCh38
rs964220446 Chr.3: 137764619-137764619 on Build GRCh38
rs899879431 Chr.3: 137764802-137764802 on Build GRCh38
rs752546426 Chr.3: 137764808-137764808 on Build GRCh38
rs375889281 Chr.3: 137764713-137764713 on Build GRCh38
rs750669644 Chr.3: 137764916-137764916 on Build GRCh38
rs749447558 Chr.3: 137764745-137764745 on Build GRCh38
rs774157859 Chr.3: 137764779-137764779 on Build GRCh38
rs777948477 Chr.3: 137764741-137764741 on Build GRCh38
rs1004449220 Chr.3: 137764978-137764978 on Build GRCh38
rs937181298 Chr.3: 137764762-137764762 on Build GRCh38
rs370962798 Chr.3: 137764587-137764587 on Build GRCh38
rs907520750 Chr.3: 137764720-137764720 on Build GRCh38
rs758724387 Chr.3: 137764931-137764931 on Build GRCh38
rs915752014 Chr.3: 137764781-137764781 on Build GRCh38
rs758195197 Chr.3: 137764749-137764750 on Build GRCh38
rs753135462 Chr.3: 137764775-137764775 on Build GRCh38
rs757558858 Chr.3: 137764760-137764760 on Build GRCh38
rs371764590 Chr.3: 137764738-137764738 on Build GRCh38
rs920313913 Chr.3: 137764751-137764751 on Build GRCh38
rs948638522 Chr.3: 137764528-137764528 on Build GRCh38
rs117725617 Chr.3: 137764934-137764934 on Build GRCh38
rs201994373 Chr.3: 137764763-137764763 on Build GRCh38

View Full Product Details