Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113096082-113096608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRIG2
Entrez Gene ID
9860
Gene Name
leucine rich repeats and immunoglobulin like domains 2
Gene Aliases
LIG-2, LIG2, UFS2
UniGene
Hs.448972
Gene Chromosome Location
Chr.1: 113073170-113132260 on Build GRCh38
Transcripts
NM_001312686.1 NM_014813.2 XM_005271369.2 XM_017002952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3704841 c.1021A>C p.N341H Chr.1: 113096295-113096295 on Build GRCh38
3704842 c.1022A>C p.N341T Chr.1: 113096296-113096296 on Build GRCh38
3862251 c.1084C>T p.Q362* Chr.1: 113096358-113096358 on Build GRCh38
4698756 c.950A>G p.D317G Chr.1: 113096224-113096224 on Build GRCh38
4020759 c.973C>T p.R325C Chr.1: 113096247-113096247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755111297 Chr.1: 113096313-113096313 on Build GRCh38
rs200968672 Chr.1: 113096262-113096262 on Build GRCh38
rs768201490 Chr.1: 113096405-113096409 on Build GRCh38
rs372448823 Chr.1: 113096289-113096289 on Build GRCh38
rs749856547 Chr.1: 113096379-113096397 on Build GRCh38
rs780508652 Chr.1: 113096370-113096370 on Build GRCh38
rs185234760 Chr.1: 113096390-113096390 on Build GRCh38
rs987359663 Chr.1: 113096140-113096140 on Build GRCh38
rs200647112 Chr.1: 113096393-113096393 on Build GRCh38
rs776152723 Chr.1: 113096404-113096405 on Build GRCh38
rs545354116 Chr.1: 113096516-113096516 on Build GRCh38
rs770532042 Chr.1: 113096300-113096300 on Build GRCh38
rs770298704 Chr.1: 113096248-113096248 on Build GRCh38
rs760970098 Chr.1: 113096295-113096295 on Build GRCh38
rs371609848 Chr.1: 113096344-113096344 on Build GRCh38
rs753954840 Chr.1: 113096304-113096304 on Build GRCh38
rs776238599 Chr.1: 113096255-113096255 on Build GRCh38
rs958363353 Chr.1: 113096438-113096438 on Build GRCh38
rs557128850 Chr.1: 113096319-113096319 on Build GRCh38
rs564884945 Chr.1: 113096527-113096527 on Build GRCh38
rs756112567 Chr.1: 113096189-113096189 on Build GRCh38
rs746603348 Chr.1: 113096353-113096353 on Build GRCh38
rs762263655 Chr.1: 113096266-113096266 on Build GRCh38
rs774743583 Chr.1: 113096399-113096400 on Build GRCh38
rs989530517 Chr.1: 113096448-113096448 on Build GRCh38
rs186434990 Chr.1: 113096551-113096551 on Build GRCh38
rs374559020 Chr.1: 113096357-113096357 on Build GRCh38
rs200140695 Chr.1: 113096263-113096263 on Build GRCh38
rs987287531 Chr.1: 113096544-113096544 on Build GRCh38
rs768033222 Chr.1: 113096271-113096271 on Build GRCh38
rs1054944474 Chr.1: 113096178-113096178 on Build GRCh38
rs772823741 Chr.1: 113096231-113096231 on Build GRCh38
rs760323257 Chr.1: 113096405-113096405 on Build GRCh38
rs971612093 Chr.1: 113096558-113096558 on Build GRCh38
rs377413842 Chr.1: 113096366-113096366 on Build GRCh38
rs1027811309 Chr.1: 113096440-113096440 on Build GRCh38
rs542808569 Chr.1: 113096415-113096415 on Build GRCh38
rs771250889 Chr.1: 113096391-113096393 on Build GRCh38
rs749128674 Chr.1: 113096207-113096207 on Build GRCh38
rs201812094 Chr.1: 113096203-113096203 on Build GRCh38
rs376102475 Chr.1: 113096341-113096341 on Build GRCh38
rs771925376 Chr.1: 113096229-113096229 on Build GRCh38
rs762454185 Chr.1: 113096397-113096397 on Build GRCh38
rs931864310 Chr.1: 113096115-113096115 on Build GRCh38
rs190184231 Chr.1: 113096453-113096453 on Build GRCh38
rs182236190 Chr.1: 113096488-113096488 on Build GRCh38
rs1005017452 Chr.1: 113096510-113096510 on Build GRCh38
rs994553188 Chr.1: 113096436-113096436 on Build GRCh38
rs750740698 Chr.1: 113096172-113096172 on Build GRCh38
rs200359864 Chr.1: 113096247-113096247 on Build GRCh38
rs752676342 Chr.1: 113096322-113096322 on Build GRCh38
rs201466448 Chr.1: 113096377-113096377 on Build GRCh38
rs566000019 Chr.1: 113096149-113096149 on Build GRCh38
rs772512173 Chr.1: 113096403-113096403 on Build GRCh38
rs769495792 Chr.1: 113096391-113096391 on Build GRCh38
rs940331224 Chr.1: 113096234-113096234 on Build GRCh38
rs775053825 Chr.1: 113096264-113096264 on Build GRCh38
rs558112559 Chr.1: 113096214-113096214 on Build GRCh38
rs761218940 Chr.1: 113096411-113096412 on Build GRCh38

View Full Product Details