Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149519498-149520001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WWTR1
Entrez Gene ID
25937
Gene Name
WW domain containing transcription regulator 1
Gene Aliases
TAZ
UniGene
Hs.477921
Gene Chromosome Location
Chr.3: 149517235-149724783 on Build GRCh38
Transcripts
NM_001168278.1 NM_001168280.1 NM_015472.4 XM_017006121.1 XM_017006122.1

SNP Detail

SNP ID Chromosome Location
rs1034619515 Chr.3: 149519957-149519957 on Build GRCh38
rs536382360 Chr.3: 149519848-149519848 on Build GRCh38
rs1009857948 Chr.3: 149519875-149519875 on Build GRCh38
rs973799453 Chr.3: 149519568-149519568 on Build GRCh38
rs908366861 Chr.3: 149519558-149519558 on Build GRCh38
rs1045969362 Chr.3: 149519680-149519680 on Build GRCh38
rs768578023 Chr.3: 149519799-149519799 on Build GRCh38
rs934782325 Chr.3: 149519701-149519701 on Build GRCh38
rs949638461 Chr.3: 149519591-149519591 on Build GRCh38
rs949357829 Chr.3: 149519803-149519803 on Build GRCh38
rs925359436 Chr.3: 149519552-149519552 on Build GRCh38
rs143286990 Chr.3: 149519966-149519966 on Build GRCh38
rs1047728258 Chr.3: 149519813-149519813 on Build GRCh38
rs149332903 Chr.3: 149519731-149519731 on Build GRCh38
rs772562035 Chr.3: 149519670-149519670 on Build GRCh38
rs575390325 Chr.3: 149519545-149519545 on Build GRCh38
rs550759744 Chr.3: 149519804-149519804 on Build GRCh38
rs958562806 Chr.3: 149519977-149519977 on Build GRCh38
rs938083732 Chr.3: 149519560-149519560 on Build GRCh38
rs955291771 Chr.3: 149519535-149519535 on Build GRCh38
rs397789319 Chr.3: 149519977-149519978 on Build GRCh38
rs530236785 Chr.3: 149519610-149519610 on Build GRCh38
rs886645221 Chr.3: 149519630-149519630 on Build GRCh38
rs192115805 Chr.3: 149519581-149519581 on Build GRCh38
rs950382888 Chr.3: 149519961-149519961 on Build GRCh38
rs995907340 Chr.3: 149519857-149519857 on Build GRCh38
rs13073865 Chr.3: 149519702-149519702 on Build GRCh38
rs546395179 Chr.3: 149519892-149519892 on Build GRCh38
rs565187430 Chr.3: 149519800-149519800 on Build GRCh38
rs1006810062 Chr.3: 149519645-149519645 on Build GRCh38
rs969051517 Chr.3: 149519891-149519891 on Build GRCh38
rs1051459299 Chr.3: 149519858-149519858 on Build GRCh38
rs937689893 Chr.3: 149519590-149519590 on Build GRCh38
rs1019923130 Chr.3: 149519890-149519890 on Build GRCh38
rs6440619 Chr.3: 149519815-149519815 on Build GRCh38
rs1010150581 Chr.3: 149519698-149519698 on Build GRCh38
rs1055142068 Chr.3: 149519561-149519561 on Build GRCh38
rs551653251 Chr.3: 149519770-149519770 on Build GRCh38
rs540247398 Chr.3: 149519679-149519679 on Build GRCh38
rs780960258 Chr.3: 149519529-149519530 on Build GRCh38
rs532918827 Chr.3: 149519663-149519663 on Build GRCh38
rs776015600 Chr.3: 149519700-149519700 on Build GRCh38
rs973851820 Chr.3: 149519863-149519863 on Build GRCh38
rs1029985032 Chr.3: 149519676-149519676 on Build GRCh38
rs57341972 Chr.3: 149519966-149519967 on Build GRCh38
rs1038291433 Chr.3: 149519827-149519827 on Build GRCh38
rs1000103956 Chr.3: 149519904-149519904 on Build GRCh38
rs187341455 Chr.3: 149519578-149519578 on Build GRCh38
rs990916089 Chr.3: 149519920-149519920 on Build GRCh38
rs1054767305 Chr.3: 149519618-149519618 on Build GRCh38
rs890167964 Chr.3: 149519874-149519874 on Build GRCh38
rs182247039 Chr.3: 149519816-149519816 on Build GRCh38
rs903343480 Chr.3: 149519694-149519694 on Build GRCh38
rs891062686 Chr.3: 149519814-149519814 on Build GRCh38
rs961152736 Chr.3: 149519566-149519566 on Build GRCh38
rs968622883 Chr.3: 149519724-149519724 on Build GRCh38
rs570104522 Chr.3: 149519715-149519715 on Build GRCh38
rs959749882 Chr.3: 149519771-149519771 on Build GRCh38
rs200537343 Chr.3: 149519768-149519768 on Build GRCh38
rs746287822 Chr.3: 149519664-149519664 on Build GRCh38

View Full Product Details