Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150422857-150423371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC22D2
Entrez Gene ID
9819
Gene Name
TSC22 domain family member 2
Gene Aliases
TILZ4a, TILZ4b, TILZ4c
UniGene
Hs.722470
Gene Chromosome Location
Chr.3: 150408268-150460119 on Build GRCh38
Transcripts
NM_001303264.1 NM_014779.3 NR_130136.1 XM_011513337.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1039847 c.1985C>T p.A662V Chr.3: 150423075-150423075 on Build GRCh38
3695865 c.2004A>G p.L668L Chr.3: 150423094-150423094 on Build GRCh38
6810311 c.2025G>A p.W675* Chr.3: 150423115-150423115 on Build GRCh38
7144013 c.2022C>G p.L674L Chr.3: 150423112-150423112 on Build GRCh38
6810312 c.2001G>A p.T667T Chr.3: 150423091-150423091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008144035 Chr.3: 150422915-150422915 on Build GRCh38
rs946636562 Chr.3: 150423237-150423237 on Build GRCh38
rs764138853 Chr.3: 150423169-150423169 on Build GRCh38
rs969209502 Chr.3: 150423021-150423021 on Build GRCh38
rs767398990 Chr.3: 150423129-150423129 on Build GRCh38
rs573796253 Chr.3: 150422895-150422895 on Build GRCh38
rs181961590 Chr.3: 150423130-150423130 on Build GRCh38
rs746649595 Chr.3: 150423044-150423045 on Build GRCh38
rs919230973 Chr.3: 150423083-150423083 on Build GRCh38
rs555759536 Chr.3: 150423037-150423037 on Build GRCh38
rs748008661 Chr.3: 150422977-150422979 on Build GRCh38
rs754370956 Chr.3: 150423047-150423047 on Build GRCh38
rs774317604 Chr.3: 150423123-150423123 on Build GRCh38
rs984795219 Chr.3: 150423059-150423059 on Build GRCh38
rs76533897 Chr.3: 150422949-150422949 on Build GRCh38
rs145029975 Chr.3: 150423073-150423073 on Build GRCh38
rs943502203 Chr.3: 150423241-150423241 on Build GRCh38
rs149343186 Chr.3: 150422960-150422960 on Build GRCh38
rs138874622 Chr.3: 150423102-150423102 on Build GRCh38
rs768218756 Chr.3: 150423159-150423160 on Build GRCh38
rs571428162 Chr.3: 150422891-150422891 on Build GRCh38
rs1049626199 Chr.3: 150422899-150422899 on Build GRCh38
rs767805747 Chr.3: 150423031-150423031 on Build GRCh38
rs966206298 Chr.3: 150422984-150422984 on Build GRCh38
rs1034849456 Chr.3: 150423043-150423043 on Build GRCh38
rs369242785 Chr.3: 150423118-150423118 on Build GRCh38
rs757751765 Chr.3: 150423054-150423054 on Build GRCh38
rs535465256 Chr.3: 150422995-150422995 on Build GRCh38
rs751340388 Chr.3: 150423013-150423013 on Build GRCh38
rs999148732 Chr.3: 150422957-150422957 on Build GRCh38
rs889267494 Chr.3: 150422908-150422908 on Build GRCh38
rs768331193 Chr.3: 150423088-150423088 on Build GRCh38
rs939187778 Chr.3: 150423205-150423205 on Build GRCh38
rs200725903 Chr.3: 150423076-150423076 on Build GRCh38
rs373390033 Chr.3: 150422997-150422997 on Build GRCh38
rs373136693 Chr.3: 150423044-150423044 on Build GRCh38
rs769250272 Chr.3: 150423104-150423104 on Build GRCh38
rs762980512 Chr.3: 150423342-150423342 on Build GRCh38
rs1052298161 Chr.3: 150422913-150422913 on Build GRCh38
rs759354291 Chr.3: 150423127-150423127 on Build GRCh38
rs762996984 Chr.3: 150423109-150423109 on Build GRCh38
rs913809035 Chr.3: 150423234-150423234 on Build GRCh38
rs956386921 Chr.3: 150422961-150422961 on Build GRCh38
rs762939394 Chr.3: 150423008-150423008 on Build GRCh38
rs1010624485 Chr.3: 150422976-150422976 on Build GRCh38
rs888780417 Chr.3: 150423303-150423303 on Build GRCh38
rs1040563761 Chr.3: 150423308-150423308 on Build GRCh38
rs910637478 Chr.3: 150423121-150423121 on Build GRCh38
rs772761749 Chr.3: 150423108-150423108 on Build GRCh38
rs557837365 Chr.3: 150423137-150423137 on Build GRCh38
rs902036998 Chr.3: 150423314-150423314 on Build GRCh38
rs377240420 Chr.3: 150423090-150423090 on Build GRCh38
rs775617637 Chr.3: 150423134-150423134 on Build GRCh38
rs777878148 Chr.3: 150423046-150423046 on Build GRCh38
rs759843002 Chr.3: 150423016-150423016 on Build GRCh38
rs765033401 Chr.3: 150423036-150423036 on Build GRCh38
rs777910982 Chr.3: 150423220-150423220 on Build GRCh38
rs764253270 Chr.3: 150423012-150423012 on Build GRCh38
rs138699932 Chr.3: 150422936-150422936 on Build GRCh38
rs879512107 Chr.3: 150423280-150423280 on Build GRCh38

View Full Product Details