Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160500279-160500821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA4
Entrez Gene ID
3840
Gene Name
karyopherin subunit alpha 4
Gene Aliases
IPOA3, QIP1, SRP3
UniGene
Hs.467866
Gene Chromosome Location
Chr.3: 160494995-160565588 on Build GRCh38
Transcripts
NM_002268.4

SNP Detail

SNP ID Chromosome Location
rs561587097 Chr.3: 160500388-160500388 on Build GRCh38
rs977961067 Chr.3: 160500313-160500326 on Build GRCh38
rs977124821 Chr.3: 160500496-160500496 on Build GRCh38
rs950260590 Chr.3: 160500701-160500701 on Build GRCh38
rs1053205 Chr.3: 160500367-160500367 on Build GRCh38
rs561812249 Chr.3: 160500714-160500714 on Build GRCh38
rs865998455 Chr.3: 160500787-160500787 on Build GRCh38
rs940266972 Chr.3: 160500763-160500763 on Build GRCh38
rs967248618 Chr.3: 160500409-160500409 on Build GRCh38
rs529054373 Chr.3: 160500393-160500393 on Build GRCh38
rs1053269254 Chr.3: 160500720-160500720 on Build GRCh38
rs766016397 Chr.3: 160500621-160500621 on Build GRCh38
rs925602296 Chr.3: 160500540-160500544 on Build GRCh38
rs765252696 Chr.3: 160500778-160500778 on Build GRCh38
rs11919332 Chr.3: 160500697-160500697 on Build GRCh38
rs540506046 Chr.3: 160500327-160500327 on Build GRCh38
rs776193579 Chr.3: 160500715-160500715 on Build GRCh38
rs754110104 Chr.3: 160500393-160500393 on Build GRCh38
rs760345814 Chr.3: 160500581-160500581 on Build GRCh38
rs908804151 Chr.3: 160500722-160500722 on Build GRCh38
rs367554028 Chr.3: 160500480-160500480 on Build GRCh38
rs201233947 Chr.3: 160500365-160500365 on Build GRCh38
rs915623845 Chr.3: 160500642-160500642 on Build GRCh38

View Full Product Details