Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170390272-170390820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIL
Entrez Gene ID
6498
Gene Name
SKI-like proto-oncogene
Gene Aliases
SNO, SnoA, SnoI, SnoN
UniGene
Hs.536655
Gene Chromosome Location
Chr.3: 170357678-170396849 on Build GRCh38
Transcripts
NM_001145097.2 NM_001145098.2 NM_001248008.1 NM_005414.4 XM_005247721.1 XM_006713735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5801320 c.1469_1484del16 p.E491fs*2 Chr.3: 170390262-170390277 on Build GRCh38
4596807 c.1525G>A p.V509I Chr.3: 170390318-170390318 on Build GRCh38
5881028 c.1498G>T p.D500Y Chr.3: 170390291-170390291 on Build GRCh38
6096996 c.1607G>T p.R536I Chr.3: 170390400-170390400 on Build GRCh38
6096997 c.1507A>G p.K503E Chr.3: 170390300-170390300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750512944 Chr.3: 170390398-170390398 on Build GRCh38
rs746651896 Chr.3: 170390548-170390548 on Build GRCh38
rs767517813 Chr.3: 170390419-170390419 on Build GRCh38
rs534892801 Chr.3: 170390450-170390450 on Build GRCh38
rs769059481 Chr.3: 170390497-170390497 on Build GRCh38
rs926246913 Chr.3: 170390714-170390714 on Build GRCh38
rs1033221039 Chr.3: 170390779-170390779 on Build GRCh38
rs568938207 Chr.3: 170390696-170390696 on Build GRCh38
rs745403418 Chr.3: 170390481-170390481 on Build GRCh38
rs753811977 Chr.3: 170390302-170390302 on Build GRCh38
rs777694138 Chr.3: 170390521-170390521 on Build GRCh38
rs368838347 Chr.3: 170390618-170390618 on Build GRCh38
rs868249950 Chr.3: 170390406-170390406 on Build GRCh38
rs754840486 Chr.3: 170390407-170390407 on Build GRCh38
rs770563584 Chr.3: 170390337-170390337 on Build GRCh38
rs61761942 Chr.3: 170390296-170390296 on Build GRCh38
rs760312418 Chr.3: 170390672-170390672 on Build GRCh38
rs1028435199 Chr.3: 170390771-170390771 on Build GRCh38
rs770619300 Chr.3: 170390592-170390592 on Build GRCh38
rs373040113 Chr.3: 170390491-170390491 on Build GRCh38
rs762088937 Chr.3: 170390385-170390385 on Build GRCh38
rs777463243 Chr.3: 170390332-170390332 on Build GRCh38
rs888366465 Chr.3: 170390678-170390678 on Build GRCh38
rs78517885 Chr.3: 170390721-170390721 on Build GRCh38
rs943583074 Chr.3: 170390633-170390633 on Build GRCh38
rs369698762 Chr.3: 170390310-170390310 on Build GRCh38
rs989118312 Chr.3: 170390787-170390787 on Build GRCh38
rs571600999 Chr.3: 170390527-170390527 on Build GRCh38
rs551044831 Chr.3: 170390298-170390298 on Build GRCh38
rs747508959 Chr.3: 170390496-170390496 on Build GRCh38
rs1043261799 Chr.3: 170390404-170390404 on Build GRCh38
rs761963659 Chr.3: 170390507-170390507 on Build GRCh38
rs557114734 Chr.3: 170390611-170390611 on Build GRCh38
rs375044899 Chr.3: 170390338-170390338 on Build GRCh38
rs769607379 Chr.3: 170390366-170390368 on Build GRCh38
rs748937979 Chr.3: 170390461-170390461 on Build GRCh38
rs748991366 Chr.3: 170390334-170390334 on Build GRCh38
rs531274398 Chr.3: 170390329-170390329 on Build GRCh38
rs777035547 Chr.3: 170390505-170390505 on Build GRCh38
rs145818678 Chr.3: 170390447-170390447 on Build GRCh38
rs1852128 Chr.3: 170390517-170390517 on Build GRCh38
rs747975807 Chr.3: 170390328-170390328 on Build GRCh38
rs376769181 Chr.3: 170390526-170390526 on Build GRCh38
rs934900507 Chr.3: 170390716-170390716 on Build GRCh38
rs776090789 Chr.3: 170390612-170390612 on Build GRCh38
rs1037264115 Chr.3: 170390702-170390702 on Build GRCh38
rs989385058 Chr.3: 170390495-170390495 on Build GRCh38
rs774493206 Chr.3: 170390412-170390414 on Build GRCh38
rs772409851 Chr.3: 170390485-170390485 on Build GRCh38
rs937693602 Chr.3: 170390614-170390614 on Build GRCh38
rs1015806905 Chr.3: 170390627-170390627 on Build GRCh38
rs764414749 Chr.3: 170390381-170390381 on Build GRCh38
rs769251727 Chr.3: 170390507-170390507 on Build GRCh38
rs555226834 Chr.3: 170390733-170390733 on Build GRCh38
rs755910931 Chr.3: 170390438-170390438 on Build GRCh38
rs1033273240 Chr.3: 170390487-170390487 on Build GRCh38
rs771701227 Chr.3: 170390386-170390386 on Build GRCh38
rs542664042 Chr.3: 170390525-170390525 on Build GRCh38
rs915592121 Chr.3: 170390695-170390695 on Build GRCh38
rs751492063 Chr.3: 170390322-170390322 on Build GRCh38
rs754964253 Chr.3: 170390326-170390326 on Build GRCh38
rs575185173 Chr.3: 170390376-170390376 on Build GRCh38
rs901045669 Chr.3: 170390707-170390707 on Build GRCh38
rs756986050 Chr.3: 170390462-170390462 on Build GRCh38
rs182102405 Chr.3: 170390778-170390778 on Build GRCh38
rs189808935 Chr.3: 170390543-170390543 on Build GRCh38
rs1026927298 Chr.3: 170390566-170390566 on Build GRCh38
rs1048277089 Chr.3: 170390637-170390637 on Build GRCh38
rs760064099 Chr.3: 170390341-170390341 on Build GRCh38
rs913614614 Chr.3: 170390518-170390518 on Build GRCh38
rs981910150 Chr.3: 170390564-170390564 on Build GRCh38
rs535819524 Chr.3: 170390648-170390648 on Build GRCh38
rs889761366 Chr.3: 170390772-170390772 on Build GRCh38
rs996741608 Chr.3: 170390300-170390300 on Build GRCh38
rs926270447 Chr.3: 170390580-170390580 on Build GRCh38
rs978381000 Chr.3: 170390701-170390701 on Build GRCh38
rs775678905 Chr.3: 170390353-170390353 on Build GRCh38
rs373100959 Chr.3: 170390327-170390327 on Build GRCh38
rs778500915 Chr.3: 170390477-170390477 on Build GRCh38
rs994825239 Chr.3: 170390550-170390550 on Build GRCh38
rs956281662 Chr.3: 170390782-170390782 on Build GRCh38
rs747932092 Chr.3: 170390312-170390326 on Build GRCh38
rs772639158 Chr.3: 170390349-170390349 on Build GRCh38
rs952733890 Chr.3: 170390591-170390591 on Build GRCh38
rs369085416 Chr.3: 170390384-170390384 on Build GRCh38
rs954049759 Chr.3: 170390400-170390400 on Build GRCh38
rs752630359 Chr.3: 170390433-170390433 on Build GRCh38
rs555227200 Chr.3: 170390318-170390318 on Build GRCh38

View Full Product Details