Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183488649-183489152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL6
Entrez Gene ID
89857
Gene Name
kelch like family member 6
Gene Aliases
-
UniGene
Hs.659147
Gene Chromosome Location
Chr.3: 183487531-183555712 on Build GRCh38
Transcripts
NM_130446.2 XM_011513273.2

SNP Detail

SNP ID Chromosome Location
rs1000336731 Chr.3: 183488988-183488988 on Build GRCh38
rs955435655 Chr.3: 183489114-183489114 on Build GRCh38
rs941390785 Chr.3: 183488969-183488969 on Build GRCh38
rs116435332 Chr.3: 183489113-183489113 on Build GRCh38
rs1038477110 Chr.3: 183488825-183488825 on Build GRCh38
rs1038834068 Chr.3: 183488982-183488982 on Build GRCh38
rs985659430 Chr.3: 183488788-183488788 on Build GRCh38
rs918577395 Chr.3: 183488827-183488827 on Build GRCh38
rs763816710 Chr.3: 183488847-183488847 on Build GRCh38
rs60846700 Chr.3: 183489009-183489009 on Build GRCh38
rs1023806637 Chr.3: 183488694-183488694 on Build GRCh38
rs1056496579 Chr.3: 183488877-183488877 on Build GRCh38
rs904058779 Chr.3: 183488961-183488961 on Build GRCh38
rs753474635 Chr.3: 183488873-183488873 on Build GRCh38
rs867612341 Chr.3: 183488816-183488816 on Build GRCh38
rs375638291 Chr.3: 183488893-183488893 on Build GRCh38
rs994362072 Chr.3: 183488708-183488708 on Build GRCh38
rs994300326 Chr.3: 183489004-183489004 on Build GRCh38
rs1017923157 Chr.3: 183489102-183489102 on Build GRCh38
rs139993699 Chr.3: 183488773-183488773 on Build GRCh38
rs563597439 Chr.3: 183489108-183489108 on Build GRCh38
rs559590177 Chr.3: 183488815-183488815 on Build GRCh38
rs922495945 Chr.3: 183488783-183488783 on Build GRCh38
rs896699242 Chr.3: 183488879-183488879 on Build GRCh38
rs1007178433 Chr.3: 183489056-183489056 on Build GRCh38
rs574809522 Chr.3: 183488833-183488833 on Build GRCh38
rs916595742 Chr.3: 183488933-183488933 on Build GRCh38
rs368679003 Chr.3: 183488966-183488966 on Build GRCh38
rs895227419 Chr.3: 183488691-183488691 on Build GRCh38
rs955731394 Chr.3: 183488758-183488758 on Build GRCh38
rs1051304674 Chr.3: 183489040-183489040 on Build GRCh38
rs1012805414 Chr.3: 183488678-183488678 on Build GRCh38
rs532317141 Chr.3: 183489123-183489123 on Build GRCh38
rs3821515 Chr.3: 183488785-183488785 on Build GRCh38
rs1032754225 Chr.3: 183489055-183489055 on Build GRCh38
rs1055039993 Chr.3: 183488932-183488932 on Build GRCh38
rs530898935 Chr.3: 183489046-183489046 on Build GRCh38
rs373913461 Chr.3: 183488880-183488880 on Build GRCh38
rs185367004 Chr.3: 183489090-183489090 on Build GRCh38
rs978566541 Chr.3: 183488711-183488711 on Build GRCh38
rs955804148 Chr.3: 183489106-183489106 on Build GRCh38
rs1020419863 Chr.3: 183489110-183489110 on Build GRCh38
rs541329574 Chr.3: 183488801-183488801 on Build GRCh38
rs1032635120 Chr.3: 183488731-183488731 on Build GRCh38
rs756777361 Chr.3: 183489030-183489030 on Build GRCh38
rs970177019 Chr.3: 183488695-183488695 on Build GRCh38
rs777703461 Chr.3: 183488786-183488786 on Build GRCh38

View Full Product Details