Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186784485-186784994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A2
Entrez Gene ID
1974
Gene Name
eukaryotic translation initiation factor 4A2
Gene Aliases
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
UniGene
Hs.518475
Gene Chromosome Location
Chr.3: 186783572-186789897 on Build GRCh38
Transcripts
NM_001967.3 NR_002587.1

Gene Symbol
SNORD2
Entrez Gene ID
619567
Gene Name
small nucleolar RNA, C/D box 2
Gene Aliases
R39B, SNR39B
UniGene
Gene Chromosome Location
Chr.3: 186784796-186784865 on Build GRCh38
Transcripts
NM_001967.3 NR_002587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4834712 c.160G>C p.E54Q Chr.3: 186784648-186784648 on Build GRCh38
4452690 c.205A>T p.K69* Chr.3: 186784693-186784693 on Build GRCh38
6255481 c.217G>T p.V73L Chr.3: 186784970-186784970 on Build GRCh38
317277 c.178C>T p.Q60* Chr.3: 186784666-186784666 on Build GRCh38
6929959 c.100A>G p.N34D Chr.3: 186784588-186784588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201474676 Chr.3: 186784621-186784621 on Build GRCh38
rs750580382 Chr.3: 186784814-186784814 on Build GRCh38
rs375543403 Chr.3: 186784897-186784897 on Build GRCh38
rs764969586 Chr.3: 186784773-186784773 on Build GRCh38
rs755185062 Chr.3: 186784677-186784679 on Build GRCh38
rs776167777 Chr.3: 186784661-186784661 on Build GRCh38
rs747723757 Chr.3: 186784635-186784635 on Build GRCh38
rs763616993 Chr.3: 186784641-186784641 on Build GRCh38
rs775303809 Chr.3: 186784863-186784866 on Build GRCh38
rs762844796 Chr.3: 186784777-186784777 on Build GRCh38
rs777728886 Chr.3: 186784745-186784746 on Build GRCh38
rs781428901 Chr.3: 186784702-186784703 on Build GRCh38
rs577099369 Chr.3: 186784676-186784676 on Build GRCh38
rs367643607 Chr.3: 186784731-186784731 on Build GRCh38
rs753913348 Chr.3: 186784795-186784795 on Build GRCh38
rs755888390 Chr.3: 186784712-186784713 on Build GRCh38
rs771307950 Chr.3: 186784596-186784596 on Build GRCh38
rs766158427 Chr.3: 186784783-186784783 on Build GRCh38
rs771820404 Chr.3: 186784846-186784847 on Build GRCh38
rs768097577 Chr.3: 186784757-186784757 on Build GRCh38
rs751445613 Chr.3: 186784717-186784717 on Build GRCh38
rs953593626 Chr.3: 186784597-186784597 on Build GRCh38
rs201418586 Chr.3: 186784735-186784735 on Build GRCh38
rs747920551 Chr.3: 186784708-186784709 on Build GRCh38
rs547575338 Chr.3: 186784852-186784852 on Build GRCh38
rs192246586 Chr.3: 186784904-186784904 on Build GRCh38
rs1009087477 Chr.3: 186784853-186784853 on Build GRCh38
rs916056628 Chr.3: 186784771-186784771 on Build GRCh38
rs566093664 Chr.3: 186784557-186784557 on Build GRCh38
rs17851348 Chr.3: 186784568-186784568 on Build GRCh38
rs377425549 Chr.3: 186784813-186784813 on Build GRCh38
rs775869166 Chr.3: 186784741-186784741 on Build GRCh38
rs552816121 Chr.3: 186784740-186784740 on Build GRCh38
rs762227302 Chr.3: 186784949-186784951 on Build GRCh38
rs773029428 Chr.3: 186784845-186784845 on Build GRCh38
rs377269772 Chr.3: 186784926-186784926 on Build GRCh38
rs775441076 Chr.3: 186784952-186784952 on Build GRCh38
rs368094046 Chr.3: 186784541-186784541 on Build GRCh38
rs898121540 Chr.3: 186784900-186784900 on Build GRCh38
rs565320884 Chr.3: 186784918-186784918 on Build GRCh38
rs781765741 Chr.3: 186784905-186784905 on Build GRCh38
rs763238505 Chr.3: 186784518-186784518 on Build GRCh38
rs771792470 Chr.3: 186784929-186784929 on Build GRCh38
rs770682987 Chr.3: 186784860-186784860 on Build GRCh38
rs751692924 Chr.3: 186784560-186784561 on Build GRCh38
rs148354660 Chr.3: 186784629-186784629 on Build GRCh38
rs774164716 Chr.3: 186784863-186784863 on Build GRCh38
rs369392748 Chr.3: 186784801-186784801 on Build GRCh38
rs780906671 Chr.3: 186784832-186784832 on Build GRCh38
rs532670729 Chr.3: 186784512-186784512 on Build GRCh38
rs751936510 Chr.3: 186784903-186784903 on Build GRCh38
rs749561100 Chr.3: 186784729-186784729 on Build GRCh38
rs759495727 Chr.3: 186784791-186784791 on Build GRCh38
rs750690230 Chr.3: 186784882-186784882 on Build GRCh38
rs753475251 Chr.3: 186784539-186784539 on Build GRCh38
rs547805544 Chr.3: 186784525-186784525 on Build GRCh38
rs36060455 Chr.3: 186784894-186784895 on Build GRCh38
rs750238082 Chr.3: 186784681-186784681 on Build GRCh38
rs748935479 Chr.3: 186784915-186784915 on Build GRCh38
rs757440601 Chr.3: 186784730-186784730 on Build GRCh38
rs747506625 Chr.3: 186784826-186784826 on Build GRCh38
rs541382107 Chr.3: 186784797-186784797 on Build GRCh38
rs893849663 Chr.3: 186784817-186784817 on Build GRCh38
rs781469768 Chr.3: 186784514-186784514 on Build GRCh38
rs200752693 Chr.3: 186784651-186784651 on Build GRCh38
rs916465363 Chr.3: 186784725-186784725 on Build GRCh38
rs377637826 Chr.3: 186784763-186784763 on Build GRCh38
rs952404904 Chr.3: 186784889-186784889 on Build GRCh38
rs199618892 Chr.3: 186784620-186784620 on Build GRCh38
rs765521699 Chr.3: 186784952-186784952 on Build GRCh38
rs747497696 Chr.3: 186784749-186784749 on Build GRCh38
rs3816112 Chr.3: 186784876-186784876 on Build GRCh38
rs758468090 Chr.3: 186784686-186784686 on Build GRCh38
rs375951775 Chr.3: 186784590-186784590 on Build GRCh38
rs752121419 Chr.3: 186784534-186784534 on Build GRCh38
rs906586830 Chr.3: 186784516-186784516 on Build GRCh38
rs972376732 Chr.3: 186784630-186784630 on Build GRCh38
rs926024001 Chr.3: 186784754-186784755 on Build GRCh38
rs367742026 Chr.3: 186784711-186784711 on Build GRCh38
rs767433110 Chr.3: 186784871-186784871 on Build GRCh38
rs780441673 Chr.3: 186784825-186784825 on Build GRCh38
rs759987412 Chr.3: 186784526-186784526 on Build GRCh38
rs758434613 Chr.3: 186784543-186784545 on Build GRCh38
rs570125990 Chr.3: 186784608-186784608 on Build GRCh38
rs762317015 Chr.3: 186784509-186784510 on Build GRCh38
rs867137018 Chr.3: 186784628-186784628 on Build GRCh38
rs765159897 Chr.3: 186784678-186784678 on Build GRCh38
rs773986749 Chr.3: 186784812-186784814 on Build GRCh38
rs558469535 Chr.3: 186784638-186784638 on Build GRCh38
rs77070252 Chr.3: 186784738-186784738 on Build GRCh38
rs745439958 Chr.3: 186784838-186784840 on Build GRCh38
rs749190693 Chr.3: 186784763-186784764 on Build GRCh38
rs556676353 Chr.3: 186784815-186784815 on Build GRCh38
rs770418129 Chr.3: 186784794-186784795 on Build GRCh38
rs761531889 Chr.3: 186784668-186784668 on Build GRCh38
rs769198532 Chr.3: 186784830-186784830 on Build GRCh38
rs1011303416 Chr.3: 186784839-186784839 on Build GRCh38
rs187041507 Chr.3: 186784569-186784569 on Build GRCh38
rs765725786 Chr.3: 186784514-186784517 on Build GRCh38
rs199724027 Chr.3: 186784954-186784954 on Build GRCh38
rs75553325 Chr.3: 186784908-186784908 on Build GRCh38
rs756180145 Chr.3: 186784718-186784718 on Build GRCh38
rs781399553 Chr.3: 186784561-186784561 on Build GRCh38
rs767063379 Chr.3: 186784529-186784529 on Build GRCh38
rs765073688 Chr.3: 186784546-186784546 on Build GRCh38
rs766380521 Chr.3: 186784546-186784547 on Build GRCh38
rs369787126 Chr.3: 186784726-186784726 on Build GRCh38
rs747674776 Chr.3: 186784840-186784840 on Build GRCh38
rs761621888 Chr.3: 186784765-186784765 on Build GRCh38
rs758375877 Chr.3: 186784560-186784560 on Build GRCh38
rs751476534 Chr.3: 186784785-186784785 on Build GRCh38
rs11538618 Chr.3: 186784631-186784631 on Build GRCh38
rs765264400 Chr.3: 186784880-186784880 on Build GRCh38
rs866637500 Chr.3: 186784685-186784685 on Build GRCh38
rs757147052 Chr.3: 186784543-186784543 on Build GRCh38
rs750924119 Chr.3: 186784518-186784519 on Build GRCh38
rs764120426 Chr.3: 186784792-186784792 on Build GRCh38
rs371691662 Chr.3: 186784566-186784566 on Build GRCh38
rs993912320 Chr.3: 186784873-186784873 on Build GRCh38
rs772941758 Chr.3: 186784776-186784776 on Build GRCh38
rs763643002 Chr.3: 186784537-186784537 on Build GRCh38
rs1052512682 Chr.3: 186784807-186784807 on Build GRCh38
rs141851340 Chr.3: 186784920-186784920 on Build GRCh38
rs1050726749 Chr.3: 186784850-186784850 on Build GRCh38
rs777400639 Chr.3: 186784910-186784910 on Build GRCh38
rs758612890 Chr.3: 186784886-186784886 on Build GRCh38
rs538734682 Chr.3: 186784877-186784877 on Build GRCh38

View Full Product Details