Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 189171832-189172323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPRG1
Entrez Gene ID
285386
Gene Name
tumor protein p63 regulated 1
Gene Aliases
FAM79B
UniGene
Hs.729544
Gene Chromosome Location
Chr.3: 189000774-189325304 on Build GRCh38
Transcripts
NM_198485.3 XM_011512732.2 XM_017006257.1 XM_017006258.1 XM_017006259.1 XM_017006260.1 XM_017006261.1

SNP Detail

SNP ID Chromosome Location
rs920421594 Chr.3: 189172213-189172213 on Build GRCh38
rs962860948 Chr.3: 189171934-189171934 on Build GRCh38
rs992143045 Chr.3: 189172004-189172004 on Build GRCh38
rs974185811 Chr.3: 189172201-189172201 on Build GRCh38
rs886091343 Chr.3: 189172116-189172116 on Build GRCh38
rs941411117 Chr.3: 189172197-189172198 on Build GRCh38
rs187238155 Chr.3: 189172022-189172022 on Build GRCh38
rs980908894 Chr.3: 189172101-189172101 on Build GRCh38
rs772052165 Chr.3: 189172103-189172103 on Build GRCh38
rs1047412693 Chr.3: 189172107-189172107 on Build GRCh38
rs981303504 Chr.3: 189171982-189171982 on Build GRCh38
rs890378756 Chr.3: 189171950-189171950 on Build GRCh38
rs560674409 Chr.3: 189171887-189171887 on Build GRCh38
rs1004500186 Chr.3: 189172129-189172129 on Build GRCh38
rs1031104610 Chr.3: 189172017-189172017 on Build GRCh38
rs367935204 Chr.3: 189172272-189172272 on Build GRCh38
rs908560672 Chr.3: 189172160-189172160 on Build GRCh38
rs960798549 Chr.3: 189172233-189172233 on Build GRCh38
rs535491471 Chr.3: 189171907-189171907 on Build GRCh38
rs780267742 Chr.3: 189171870-189171870 on Build GRCh38
rs529497789 Chr.3: 189171896-189171896 on Build GRCh38
rs76211722 Chr.3: 189171925-189171925 on Build GRCh38
rs118092872 Chr.3: 189172210-189172210 on Build GRCh38
rs189788627 Chr.3: 189172177-189172177 on Build GRCh38
rs1033729564 Chr.3: 189171987-189171987 on Build GRCh38
rs1030341571 Chr.3: 189171911-189171911 on Build GRCh38
rs140125711 Chr.3: 189172140-189172140 on Build GRCh38
rs1034191502 Chr.3: 189172218-189172218 on Build GRCh38
rs151140058 Chr.3: 189172058-189172058 on Build GRCh38
rs370620961 Chr.3: 189171965-189171965 on Build GRCh38
rs887332741 Chr.3: 189171920-189171920 on Build GRCh38
rs943179897 Chr.3: 189172068-189172068 on Build GRCh38
rs1017141543 Chr.3: 189171930-189171930 on Build GRCh38
rs982779599 Chr.3: 189172158-189172158 on Build GRCh38
rs1037519042 Chr.3: 189171866-189171866 on Build GRCh38
rs9847957 Chr.3: 189172047-189172047 on Build GRCh38
rs34021464 Chr.3: 189171879-189171880 on Build GRCh38
rs1038627787 Chr.3: 189172130-189172130 on Build GRCh38
rs540873815 Chr.3: 189171880-189171880 on Build GRCh38
rs966628176 Chr.3: 189172041-189172041 on Build GRCh38
rs1039205866 Chr.3: 189172084-189172084 on Build GRCh38
rs941443965 Chr.3: 189171863-189171863 on Build GRCh38
rs933001139 Chr.3: 189172104-189172104 on Build GRCh38
rs1050401784 Chr.3: 189171938-189171938 on Build GRCh38
rs751379581 Chr.3: 189171991-189171991 on Build GRCh38

View Full Product Details