Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2659733-2660244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM193A
Entrez Gene ID
8603
Gene Name
family with sequence similarity 193 member A
Gene Aliases
C4orf8, RES4-22
UniGene
Hs.652364
Gene Chromosome Location
Chr.4: 2596288-2732575 on Build GRCh38
Transcripts
NM_001256666.1 NM_001256667.1 NM_001256668.1 NM_003704.3 NR_046335.1 NR_046336.1 XM_006713930.2 XM_006713932.2 XM_011513590.1 XM_011513593.2 XM_017008788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6535260 c.727C>G p.Q243E Chr.4: 2659909-2659909 on Build GRCh38
7086249 c.762G>A p.R254R Chr.4: 2659944-2659944 on Build GRCh38
5927327 c.689C>T p.P230L Chr.4: 2659871-2659871 on Build GRCh38
74592 c.775G>A p.V259M Chr.4: 2659957-2659957 on Build GRCh38
5927326 c.689C>T p.P230L Chr.4: 2659871-2659871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141633773 Chr.4: 2659950-2659950 on Build GRCh38
rs939575507 Chr.4: 2660168-2660168 on Build GRCh38
rs781477880 Chr.4: 2659778-2659778 on Build GRCh38
rs372722690 Chr.4: 2659913-2659913 on Build GRCh38
rs201125809 Chr.4: 2659787-2659787 on Build GRCh38
rs748099791 Chr.4: 2659865-2659866 on Build GRCh38
rs779008269 Chr.4: 2659806-2659806 on Build GRCh38
rs868133441 Chr.4: 2660044-2660044 on Build GRCh38
rs763126217 Chr.4: 2660083-2660083 on Build GRCh38
rs76312567 Chr.4: 2660212-2660212 on Build GRCh38
rs777755872 Chr.4: 2660105-2660105 on Build GRCh38
rs769059938 Chr.4: 2660060-2660060 on Build GRCh38
rs956474381 Chr.4: 2659826-2659826 on Build GRCh38
rs550227977 Chr.4: 2660042-2660042 on Build GRCh38
rs867864099 Chr.4: 2659962-2659962 on Build GRCh38
rs756373943 Chr.4: 2659773-2659773 on Build GRCh38
rs747969103 Chr.4: 2660009-2660009 on Build GRCh38
rs764951211 Chr.4: 2659869-2659869 on Build GRCh38
rs1032226437 Chr.4: 2660065-2660065 on Build GRCh38
rs752638374 Chr.4: 2660100-2660100 on Build GRCh38
rs983953012 Chr.4: 2660203-2660203 on Build GRCh38
rs980097819 Chr.4: 2660136-2660136 on Build GRCh38
rs561355288 Chr.4: 2659873-2659873 on Build GRCh38
rs773879823 Chr.4: 2659850-2659850 on Build GRCh38
rs757347828 Chr.4: 2659795-2659795 on Build GRCh38
rs765538883 Chr.4: 2660080-2660080 on Build GRCh38
rs769125391 Chr.4: 2659866-2659866 on Build GRCh38
rs762197551 Chr.4: 2660073-2660073 on Build GRCh38
rs140213757 Chr.4: 2659973-2659973 on Build GRCh38
rs754056839 Chr.4: 2660062-2660062 on Build GRCh38
rs928165230 Chr.4: 2659853-2659853 on Build GRCh38
rs753162223 Chr.4: 2659766-2659766 on Build GRCh38
rs942361586 Chr.4: 2660000-2660000 on Build GRCh38
rs200559674 Chr.4: 2659870-2659870 on Build GRCh38
rs202003033 Chr.4: 2659909-2659909 on Build GRCh38
rs765368271 Chr.4: 2660096-2660096 on Build GRCh38
rs61731404 Chr.4: 2659974-2659974 on Build GRCh38
rs761131118 Chr.4: 2659966-2659966 on Build GRCh38
rs144441001 Chr.4: 2659822-2659822 on Build GRCh38
rs928251550 Chr.4: 2660150-2660150 on Build GRCh38
rs1040725240 Chr.4: 2660022-2660022 on Build GRCh38
rs970333927 Chr.4: 2660132-2660132 on Build GRCh38
rs758448018 Chr.4: 2660178-2660178 on Build GRCh38
rs781184398 Chr.4: 2660186-2660186 on Build GRCh38
rs770304794 Chr.4: 2659953-2659953 on Build GRCh38
rs1025675951 Chr.4: 2659982-2659982 on Build GRCh38
rs1035719099 Chr.4: 2660139-2660139 on Build GRCh38
rs764661497 Chr.4: 2659967-2659967 on Build GRCh38
rs980150320 Chr.4: 2659917-2659917 on Build GRCh38
rs758528382 Chr.4: 2659981-2659981 on Build GRCh38
rs368956480 Chr.4: 2659911-2659911 on Build GRCh38
rs371056145 Chr.4: 2660011-2660011 on Build GRCh38
rs772058091 Chr.4: 2659815-2659815 on Build GRCh38
rs533013880 Chr.4: 2660003-2660003 on Build GRCh38
rs757545418 Chr.4: 2659889-2659889 on Build GRCh38
rs755992082 Chr.4: 2660102-2660102 on Build GRCh38
rs1045357116 Chr.4: 2659827-2659827 on Build GRCh38
rs747952548 Chr.4: 2659931-2659931 on Build GRCh38
rs1031861327 Chr.4: 2659774-2659774 on Build GRCh38
rs754938002 Chr.4: 2659999-2659999 on Build GRCh38
rs751648630 Chr.4: 2660089-2660089 on Build GRCh38
rs201879000 Chr.4: 2660084-2660084 on Build GRCh38
rs764312520 Chr.4: 2659883-2659883 on Build GRCh38
rs780048454 Chr.4: 2659988-2659988 on Build GRCh38
rs541390001 Chr.4: 2659872-2659872 on Build GRCh38
rs369254012 Chr.4: 2660095-2660095 on Build GRCh38
rs768297494 Chr.4: 2659851-2659851 on Build GRCh38
rs751626107 Chr.4: 2659997-2659997 on Build GRCh38
rs370202929 Chr.4: 2660142-2660142 on Build GRCh38
rs761605899 Chr.4: 2659867-2659867 on Build GRCh38
rs750539957 Chr.4: 2660141-2660141 on Build GRCh38
rs566085187 Chr.4: 2660116-2660116 on Build GRCh38
rs575751511 Chr.4: 2659821-2659821 on Build GRCh38
rs931151172 Chr.4: 2660187-2660187 on Build GRCh38
rs765452273 Chr.4: 2659976-2659976 on Build GRCh38
rs546877490 Chr.4: 2660088-2660088 on Build GRCh38
rs776083440 Chr.4: 2659956-2659956 on Build GRCh38
rs61731403 Chr.4: 2659980-2659980 on Build GRCh38
rs372399953 Chr.4: 2659757-2659757 on Build GRCh38
rs199796324 Chr.4: 2660070-2660070 on Build GRCh38
rs759078074 Chr.4: 2660053-2660053 on Build GRCh38
rs773803357 Chr.4: 2660050-2660050 on Build GRCh38
rs938820326 Chr.4: 2659932-2659932 on Build GRCh38
rs147503078 Chr.4: 2659949-2659949 on Build GRCh38
rs776368968 Chr.4: 2659860-2659860 on Build GRCh38
rs745946997 Chr.4: 2659812-2659812 on Build GRCh38
rs928706053 Chr.4: 2659924-2659924 on Build GRCh38
rs769627283 Chr.4: 2659939-2659939 on Build GRCh38
rs754203067 Chr.4: 2659885-2659885 on Build GRCh38
rs750517238 Chr.4: 2659899-2659899 on Build GRCh38
rs768133102 Chr.4: 2659764-2659764 on Build GRCh38
rs758408323 Chr.4: 2659902-2659902 on Build GRCh38
rs748896983 Chr.4: 2660043-2660043 on Build GRCh38
rs375688554 Chr.4: 2659891-2659891 on Build GRCh38

View Full Product Details