Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119514752-119515247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD3B1
Entrez Gene ID
3283
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
Gene Aliases
3BETAHSD, HSD3B, HSDB3, HSDB3A, I, SDR11E1
UniGene
Hs.364941
Gene Chromosome Location
Chr.1: 119507112-119515060 on Build GRCh38
Transcripts
NM_000862.2 XM_011541314.2 XM_017001119.1

SNP Detail

SNP ID Chromosome Location
rs963616014 Chr.1: 119514809-119514809 on Build GRCh38
rs186582654 Chr.1: 119514938-119514938 on Build GRCh38
rs746605009 Chr.1: 119514817-119514817 on Build GRCh38
rs587616510 Chr.1: 119515033-119515033 on Build GRCh38
rs1001647666 Chr.1: 119514810-119514810 on Build GRCh38
rs138915177 Chr.1: 119514983-119514983 on Build GRCh38
rs759193217 Chr.1: 119514982-119514982 on Build GRCh38
rs1037989007 Chr.1: 119515109-119515109 on Build GRCh38
rs1001659066 Chr.1: 119515155-119515155 on Build GRCh38
rs184188850 Chr.1: 119514867-119514867 on Build GRCh38
rs587716083 Chr.1: 119514897-119514897 on Build GRCh38
rs978058473 Chr.1: 119514937-119514937 on Build GRCh38
rs766899806 Chr.1: 119514805-119514805 on Build GRCh38
rs770429043 Chr.1: 119515019-119515019 on Build GRCh38
rs1013319634 Chr.1: 119515180-119515180 on Build GRCh38
rs924244848 Chr.1: 119515174-119515174 on Build GRCh38
rs879430202 Chr.1: 119515212-119515222 on Build GRCh38
rs991805946 Chr.1: 119514861-119514861 on Build GRCh38
rs1005162565 Chr.1: 119514793-119514793 on Build GRCh38
rs1034458413 Chr.1: 119515156-119515156 on Build GRCh38
rs886465372 Chr.1: 119515072-119515072 on Build GRCh38
rs972167753 Chr.1: 119515048-119515048 on Build GRCh38
rs760107898 Chr.1: 119515196-119515196 on Build GRCh38
rs375382280 Chr.1: 119514919-119514919 on Build GRCh38
rs868202752 Chr.1: 119514832-119514832 on Build GRCh38
rs986395303 Chr.1: 119515152-119515152 on Build GRCh38
rs940682470 Chr.1: 119515085-119515085 on Build GRCh38
rs1025129406 Chr.1: 119515195-119515195 on Build GRCh38
rs986065504 Chr.1: 119514800-119514800 on Build GRCh38
rs1056880421 Chr.1: 119515201-119515201 on Build GRCh38
rs781484092 Chr.1: 119514935-119514935 on Build GRCh38
rs922028762 Chr.1: 119515005-119515005 on Build GRCh38
rs745518879 Chr.1: 119515166-119515166 on Build GRCh38
rs1016234714 Chr.1: 119515000-119515000 on Build GRCh38
rs936524809 Chr.1: 119514951-119514951 on Build GRCh38
rs954552847 Chr.1: 119515140-119515140 on Build GRCh38
rs587747736 Chr.1: 119515015-119515015 on Build GRCh38
rs910490951 Chr.1: 119514807-119514807 on Build GRCh38
rs894913971 Chr.1: 119515157-119515157 on Build GRCh38
rs587653960 Chr.1: 119514952-119514952 on Build GRCh38
rs959075024 Chr.1: 119514845-119514845 on Build GRCh38
rs978154145 Chr.1: 119515207-119515207 on Build GRCh38
rs35645491 Chr.1: 119515222-119515222 on Build GRCh38
rs917563497 Chr.1: 119514909-119514909 on Build GRCh38
rs1051836302 Chr.1: 119515068-119515068 on Build GRCh38
rs115746802 Chr.1: 119514917-119514917 on Build GRCh38
rs746453751 Chr.1: 119515004-119515004 on Build GRCh38
rs780746704 Chr.1: 119515119-119515119 on Build GRCh38

View Full Product Details