Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8215990-8216483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC1
Entrez Gene ID
54436
Gene Name
SH3 domain and tetratricopeptide repeats 1
Gene Aliases
-
UniGene
Hs.479116
Gene Chromosome Location
Chr.4: 8182072-8241103 on Build GRCh38
Transcripts
NM_001318480.1 NM_018986.4 NR_134639.1 XM_006713889.2 XM_011513488.1 XM_011513489.2 XM_017008300.1 XM_017008301.1 XM_017008302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6189031 c.547G>A p.E183K Chr.4: 8216176-8216176 on Build GRCh38
6777386 c.517G>A p.D173N Chr.4: 8216146-8216146 on Build GRCh38
3606479 c.552G>A p.E184E Chr.4: 8216181-8216181 on Build GRCh38
1058101 c.482-1G>T p.? Chr.4: 8216110-8216110 on Build GRCh38
3606480 c.603C>T p.T201T Chr.4: 8216232-8216232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746440266 Chr.4: 8216138-8216138 on Build GRCh38
rs761748639 Chr.4: 8216205-8216205 on Build GRCh38
rs1021261025 Chr.4: 8216044-8216044 on Build GRCh38
rs758408435 Chr.4: 8216128-8216128 on Build GRCh38
rs1001418361 Chr.4: 8216434-8216434 on Build GRCh38
rs894371404 Chr.4: 8216033-8216033 on Build GRCh38
rs749792356 Chr.4: 8216226-8216226 on Build GRCh38
rs758524420 Chr.4: 8216211-8216211 on Build GRCh38
rs915706682 Chr.4: 8216387-8216387 on Build GRCh38
rs907008199 Chr.4: 8216017-8216017 on Build GRCh38
rs374629121 Chr.4: 8216230-8216230 on Build GRCh38
rs566552104 Chr.4: 8216349-8216349 on Build GRCh38
rs371436398 Chr.4: 8216216-8216216 on Build GRCh38
rs974313847 Chr.4: 8216317-8216317 on Build GRCh38
rs539172619 Chr.4: 8216309-8216309 on Build GRCh38
rs753621587 Chr.4: 8216192-8216192 on Build GRCh38
rs998795385 Chr.4: 8216417-8216417 on Build GRCh38
rs1047104128 Chr.4: 8216189-8216189 on Build GRCh38
rs879920274 Chr.4: 8216249-8216249 on Build GRCh38
rs771157387 Chr.4: 8216063-8216063 on Build GRCh38
rs550492779 Chr.4: 8216117-8216117 on Build GRCh38
rs756567481 Chr.4: 8216283-8216283 on Build GRCh38
rs547986531 Chr.4: 8216333-8216333 on Build GRCh38
rs768672356 Chr.4: 8216094-8216094 on Build GRCh38
rs74935855 Chr.4: 8216067-8216067 on Build GRCh38
rs534051835 Chr.4: 8216369-8216369 on Build GRCh38
rs750110207 Chr.4: 8216232-8216232 on Build GRCh38
rs951617085 Chr.4: 8216325-8216325 on Build GRCh38
rs371754143 Chr.4: 8216300-8216300 on Build GRCh38
rs771374243 Chr.4: 8216135-8216135 on Build GRCh38
rs928666323 Chr.4: 8216338-8216338 on Build GRCh38
rs761107990 Chr.4: 8216061-8216061 on Build GRCh38
rs367921811 Chr.4: 8216297-8216297 on Build GRCh38
rs760094550 Chr.4: 8216070-8216070 on Build GRCh38
rs1011934417 Chr.4: 8216035-8216035 on Build GRCh38
rs907103120 Chr.4: 8216429-8216429 on Build GRCh38
rs1006939575 Chr.4: 8216047-8216047 on Build GRCh38
rs555609236 Chr.4: 8216282-8216282 on Build GRCh38
rs141095376 Chr.4: 8216218-8216218 on Build GRCh38
rs757303151 Chr.4: 8216269-8216269 on Build GRCh38
rs577348513 Chr.4: 8216085-8216085 on Build GRCh38
rs752594588 Chr.4: 8216208-8216208 on Build GRCh38
rs1032910404 Chr.4: 8216093-8216093 on Build GRCh38
rs555388514 Chr.4: 8216413-8216413 on Build GRCh38
rs998513818 Chr.4: 8216277-8216277 on Build GRCh38
rs147709035 Chr.4: 8216294-8216294 on Build GRCh38
rs751006775 Chr.4: 8216336-8216336 on Build GRCh38
rs145544836 Chr.4: 8216238-8216238 on Build GRCh38
rs376429563 Chr.4: 8216308-8216308 on Build GRCh38
rs761410146 Chr.4: 8216144-8216144 on Build GRCh38
rs761615889 Chr.4: 8216083-8216083 on Build GRCh38
rs140242226 Chr.4: 8216217-8216217 on Build GRCh38
rs770410200 Chr.4: 8216141-8216141 on Build GRCh38
rs925289600 Chr.4: 8216350-8216350 on Build GRCh38
rs764965167 Chr.4: 8216247-8216247 on Build GRCh38
rs377112693 Chr.4: 8216075-8216075 on Build GRCh38
rs183071768 Chr.4: 8216264-8216264 on Build GRCh38
rs555710448 Chr.4: 8216092-8216097 on Build GRCh38
rs772967425 Chr.4: 8216151-8216151 on Build GRCh38
rs751575157 Chr.4: 8216265-8216265 on Build GRCh38
rs543868013 Chr.4: 8216037-8216037 on Build GRCh38
rs770215839 Chr.4: 8216219-8216219 on Build GRCh38
rs974621368 Chr.4: 8216185-8216185 on Build GRCh38
rs751066905 Chr.4: 8216303-8216303 on Build GRCh38
rs756648291 Chr.4: 8216460-8216460 on Build GRCh38
rs766067697 Chr.4: 8216087-8216087 on Build GRCh38
rs971023223 Chr.4: 8216288-8216288 on Build GRCh38
rs377378834 Chr.4: 8216031-8216031 on Build GRCh38
rs2291070 Chr.4: 8216357-8216357 on Build GRCh38
rs1024358905 Chr.4: 8216124-8216124 on Build GRCh38
rs551548207 Chr.4: 8216335-8216335 on Build GRCh38
rs375047951 Chr.4: 8216131-8216131 on Build GRCh38
rs1051415661 Chr.4: 8216431-8216431 on Build GRCh38
rs1056298729 Chr.4: 8216362-8216362 on Build GRCh38
rs368466334 Chr.4: 8216153-8216153 on Build GRCh38
rs1712290 Chr.4: 8216102-8216102 on Build GRCh38
rs550297805 Chr.4: 8216190-8216190 on Build GRCh38
rs754713960 Chr.4: 8216204-8216204 on Build GRCh38
rs769031423 Chr.4: 8216298-8216298 on Build GRCh38
rs929982389 Chr.4: 8216170-8216170 on Build GRCh38
rs1034632801 Chr.4: 8216032-8216032 on Build GRCh38
rs1039704477 Chr.4: 8216339-8216339 on Build GRCh38
rs766341280 Chr.4: 8216252-8216254 on Build GRCh38
rs760433690 Chr.4: 8216235-8216235 on Build GRCh38
rs187059321 Chr.4: 8216321-8216321 on Build GRCh38
rs891622793 Chr.4: 8216393-8216393 on Build GRCh38
rs781568992 Chr.4: 8216274-8216274 on Build GRCh38
rs766895282 Chr.4: 8216149-8216149 on Build GRCh38
rs776082223 Chr.4: 8216142-8216142 on Build GRCh38
rs962830497 Chr.4: 8216178-8216178 on Build GRCh38
rs748244165 Chr.4: 8216112-8216112 on Build GRCh38
rs144802316 Chr.4: 8216236-8216236 on Build GRCh38
rs779026576 Chr.4: 8216104-8216104 on Build GRCh38
rs1042928090 Chr.4: 8216396-8216396 on Build GRCh38
rs752676640 Chr.4: 8216252-8216252 on Build GRCh38
rs773994129 Chr.4: 8216082-8216082 on Build GRCh38
rs776825018 Chr.4: 8216136-8216136 on Build GRCh38
rs762765710 Chr.4: 8216258-8216258 on Build GRCh38
rs780736932 Chr.4: 8216222-8216222 on Build GRCh38
rs890139218 Chr.4: 8216449-8216449 on Build GRCh38
rs947215786 Chr.4: 8216394-8216394 on Build GRCh38
rs189047872 Chr.4: 8216458-8216458 on Build GRCh38

View Full Product Details