Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8218193-8218696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC1
Entrez Gene ID
54436
Gene Name
SH3 domain and tetratricopeptide repeats 1
Gene Aliases
-
UniGene
Hs.479116
Gene Chromosome Location
Chr.4: 8182072-8241103 on Build GRCh38
Transcripts
NM_001318480.1 NM_018986.4 NR_134639.1 XM_006713889.2 XM_006713891.3 XM_011513488.1 XM_011513489.2 XM_017008300.1 XM_017008301.1 XM_017008302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3768019 c.901G>A p.G301S Chr.4: 8218332-8218332 on Build GRCh38
4511362 c.865C>T p.P289S Chr.4: 8218296-8218296 on Build GRCh38
5866838 c.856C>T p.P286S Chr.4: 8218287-8218287 on Build GRCh38
4399335 c.916G>A p.A306T Chr.4: 8218347-8218347 on Build GRCh38
6777390 c.854T>C p.I285T Chr.4: 8218285-8218285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528707821 Chr.4: 8218588-8218588 on Build GRCh38
rs767411858 Chr.4: 8218376-8218376 on Build GRCh38
rs767193388 Chr.4: 8218314-8218314 on Build GRCh38
rs749314238 Chr.4: 8218659-8218659 on Build GRCh38
rs1053417497 Chr.4: 8218449-8218449 on Build GRCh38
rs144885198 Chr.4: 8218301-8218301 on Build GRCh38
rs917978138 Chr.4: 8218625-8218625 on Build GRCh38
rs756001440 Chr.4: 8218329-8218329 on Build GRCh38
rs777818853 Chr.4: 8218422-8218422 on Build GRCh38
rs949253462 Chr.4: 8218360-8218360 on Build GRCh38
rs779371781 Chr.4: 8218348-8218351 on Build GRCh38
rs756933443 Chr.4: 8218248-8218248 on Build GRCh38
rs777207892 Chr.4: 8218352-8218352 on Build GRCh38
rs1281138 Chr.4: 8218302-8218302 on Build GRCh38
rs773821923 Chr.4: 8218293-8218293 on Build GRCh38
rs774071418 Chr.4: 8218511-8218511 on Build GRCh38
rs560359933 Chr.4: 8218498-8218498 on Build GRCh38
rs949537448 Chr.4: 8218626-8218626 on Build GRCh38
rs768019764 Chr.4: 8218658-8218658 on Build GRCh38
rs747613749 Chr.4: 8218346-8218346 on Build GRCh38
rs751409634 Chr.4: 8218246-8218246 on Build GRCh38
rs758157912 Chr.4: 8218245-8218245 on Build GRCh38
rs892108607 Chr.4: 8218660-8218660 on Build GRCh38
rs190297953 Chr.4: 8218491-8218491 on Build GRCh38
rs1281139 Chr.4: 8218642-8218642 on Build GRCh38
rs201346781 Chr.4: 8218262-8218262 on Build GRCh38
rs149301498 Chr.4: 8218594-8218594 on Build GRCh38
rs1000820259 Chr.4: 8218448-8218448 on Build GRCh38
rs998075621 Chr.4: 8218506-8218506 on Build GRCh38
rs376095673 Chr.4: 8218332-8218332 on Build GRCh38
rs373637959 Chr.4: 8218334-8218334 on Build GRCh38
rs932005384 Chr.4: 8218518-8218518 on Build GRCh38
rs750412446 Chr.4: 8218320-8218320 on Build GRCh38
rs773447586 Chr.4: 8218344-8218344 on Build GRCh38
rs780081627 Chr.4: 8218531-8218531 on Build GRCh38
rs182812422 Chr.4: 8218510-8218510 on Build GRCh38
rs771575106 Chr.4: 8218347-8218347 on Build GRCh38
rs902365709 Chr.4: 8218598-8218598 on Build GRCh38
rs766845129 Chr.4: 8218252-8218252 on Build GRCh38
rs750171531 Chr.4: 8218379-8218379 on Build GRCh38
rs139298992 Chr.4: 8218339-8218339 on Build GRCh38
rs775001182 Chr.4: 8218560-8218560 on Build GRCh38
rs892139911 Chr.4: 8218455-8218455 on Build GRCh38
rs372034893 Chr.4: 8218397-8218397 on Build GRCh38
rs774490040 Chr.4: 8218276-8218276 on Build GRCh38
rs377622777 Chr.4: 8218384-8218384 on Build GRCh38
rs768879499 Chr.4: 8218269-8218269 on Build GRCh38
rs151285463 Chr.4: 8218304-8218304 on Build GRCh38
rs570144613 Chr.4: 8218593-8218593 on Build GRCh38
rs1053722691 Chr.4: 8218653-8218653 on Build GRCh38
rs199605281 Chr.4: 8218340-8218340 on Build GRCh38
rs748578841 Chr.4: 8218281-8218281 on Build GRCh38
rs750856944 Chr.4: 8218225-8218225 on Build GRCh38
rs1036406075 Chr.4: 8218326-8218326 on Build GRCh38
rs534905551 Chr.4: 8218607-8218607 on Build GRCh38
rs761057545 Chr.4: 8218297-8218297 on Build GRCh38
rs200478204 Chr.4: 8218364-8218364 on Build GRCh38
rs113142756 Chr.4: 8218464-8218464 on Build GRCh38
rs764035366 Chr.4: 8218242-8218242 on Build GRCh38
rs551774383 Chr.4: 8218532-8218532 on Build GRCh38
rs376330323 Chr.4: 8218317-8218317 on Build GRCh38
rs185483665 Chr.4: 8218263-8218263 on Build GRCh38
rs1045294485 Chr.4: 8218630-8218630 on Build GRCh38
rs35103792 Chr.4: 8218632-8218633 on Build GRCh38
rs1009298287 Chr.4: 8218460-8218460 on Build GRCh38
rs889611263 Chr.4: 8218261-8218261 on Build GRCh38
rs745521828 Chr.4: 8218529-8218529 on Build GRCh38
rs905108896 Chr.4: 8218385-8218385 on Build GRCh38
rs1015520373 Chr.4: 8218458-8218458 on Build GRCh38
rs368180593 Chr.4: 8218300-8218300 on Build GRCh38
rs568724481 Chr.4: 8218597-8218597 on Build GRCh38
rs574103104 Chr.4: 8218381-8218381 on Build GRCh38
rs965186701 Chr.4: 8218474-8218474 on Build GRCh38
rs926487175 Chr.4: 8218631-8218631 on Build GRCh38
rs752441931 Chr.4: 8218241-8218241 on Build GRCh38
rs371090550 Chr.4: 8218331-8218331 on Build GRCh38
rs201214130 Chr.4: 8218240-8218240 on Build GRCh38
rs772530771 Chr.4: 8218343-8218343 on Build GRCh38
rs755194202 Chr.4: 8218264-8218264 on Build GRCh38
rs1298145 Chr.4: 8218673-8218673 on Build GRCh38
rs759109171 Chr.4: 8218226-8218226 on Build GRCh38
rs772277318 Chr.4: 8218285-8218285 on Build GRCh38
rs541835923 Chr.4: 8218463-8218463 on Build GRCh38
rs1011937090 Chr.4: 8218635-8218635 on Build GRCh38

View Full Product Details