Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48067590-48068073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXK
Entrez Gene ID
7294
Gene Name
TXK tyrosine kinase
Gene Aliases
BTKL, PSCTK5, PTK4, RLK, TKL
UniGene
Hs.479669
Gene Chromosome Location
Chr.4: 48066393-48135322 on Build GRCh38
Transcripts
NM_003328.2 XM_011513747.2 XM_011513748.2 XM_017008580.1 XM_017008581.1 XM_017008582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5034188 c.1516-10C>T p.? Chr.4: 48067715-48067715 on Build GRCh38
6476607 c.1578C>G p.T526T Chr.4: 48067643-48067643 on Build GRCh38
7238887 c.1542C>G p.A514A Chr.4: 48067679-48067679 on Build GRCh38
7238949 c.1516-3C>T p.? Chr.4: 48067708-48067708 on Build GRCh38
6814462 c.1528C>T p.R510C Chr.4: 48067693-48067693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763222681 Chr.4: 48067634-48067634 on Build GRCh38
rs766180183 Chr.4: 48067644-48067644 on Build GRCh38
rs759605449 Chr.4: 48067733-48067733 on Build GRCh38
rs770134818 Chr.4: 48067705-48067705 on Build GRCh38
rs768722944 Chr.4: 48067709-48067709 on Build GRCh38
rs752472445 Chr.4: 48067750-48067750 on Build GRCh38
rs774658130 Chr.4: 48067710-48067710 on Build GRCh38
rs764423181 Chr.4: 48067687-48067687 on Build GRCh38
rs1043910898 Chr.4: 48067954-48067954 on Build GRCh38
rs891219456 Chr.4: 48067977-48067977 on Build GRCh38
rs565843003 Chr.4: 48067720-48067720 on Build GRCh38
rs574648060 Chr.4: 48068010-48068010 on Build GRCh38
rs912947650 Chr.4: 48067889-48067889 on Build GRCh38
rs1006653918 Chr.4: 48067997-48067997 on Build GRCh38
rs925464710 Chr.4: 48067981-48067981 on Build GRCh38
rs752521636 Chr.4: 48067679-48067679 on Build GRCh38
rs967956192 Chr.4: 48067649-48067649 on Build GRCh38
rs2230594 Chr.4: 48067679-48067679 on Build GRCh38
rs1029695000 Chr.4: 48067971-48067971 on Build GRCh38
rs758679691 Chr.4: 48067811-48067811 on Build GRCh38
rs980068044 Chr.4: 48067826-48067826 on Build GRCh38
rs746410519 Chr.4: 48067693-48067693 on Build GRCh38
rs770493653 Chr.4: 48067680-48067680 on Build GRCh38
rs906093625 Chr.4: 48067844-48067844 on Build GRCh38
rs777485377 Chr.4: 48067671-48067671 on Build GRCh38
rs539408241 Chr.4: 48067775-48067775 on Build GRCh38
rs762022341 Chr.4: 48067678-48067678 on Build GRCh38
rs376401047 Chr.4: 48067737-48067737 on Build GRCh38
rs1018076336 Chr.4: 48067999-48067999 on Build GRCh38
rs967882898 Chr.4: 48068006-48068006 on Build GRCh38
rs1007845789 Chr.4: 48067631-48067631 on Build GRCh38
rs941635547 Chr.4: 48067866-48067866 on Build GRCh38
rs991919362 Chr.4: 48067732-48067732 on Build GRCh38
rs777214729 Chr.4: 48067635-48067635 on Build GRCh38
rs369754627 Chr.4: 48067883-48067883 on Build GRCh38
rs778918085 Chr.4: 48067666-48067666 on Build GRCh38
rs777523956 Chr.4: 48068017-48068017 on Build GRCh38
rs202057212 Chr.4: 48067668-48067668 on Build GRCh38
rs373037539 Chr.4: 48068018-48068018 on Build GRCh38
rs1016082953 Chr.4: 48067792-48067792 on Build GRCh38
rs917704489 Chr.4: 48067767-48067767 on Build GRCh38
rs9996535 Chr.4: 48067708-48067708 on Build GRCh38
rs375788286 Chr.4: 48067692-48067692 on Build GRCh38
rs549984322 Chr.4: 48067659-48067659 on Build GRCh38
rs879183436 Chr.4: 48067820-48067820 on Build GRCh38
rs142307382 Chr.4: 48067630-48067630 on Build GRCh38
rs766757985 Chr.4: 48067870-48067870 on Build GRCh38
rs1018107506 Chr.4: 48067638-48067638 on Build GRCh38
rs969933464 Chr.4: 48067808-48067808 on Build GRCh38
rs369204768 Chr.4: 48067749-48067749 on Build GRCh38
rs553089920 Chr.4: 48067890-48067890 on Build GRCh38
rs116501833 Chr.4: 48067810-48067810 on Build GRCh38
rs867400858 Chr.4: 48067650-48067650 on Build GRCh38
rs755118984 Chr.4: 48067888-48067888 on Build GRCh38
rs761986798 Chr.4: 48067636-48067636 on Build GRCh38
rs375701019 Chr.4: 48067871-48067871 on Build GRCh38
rs770189847 Chr.4: 48067621-48067621 on Build GRCh38
rs200170089 Chr.4: 48067654-48067654 on Build GRCh38
rs775778228 Chr.4: 48067707-48067707 on Build GRCh38
rs767677274 Chr.4: 48067721-48067721 on Build GRCh38

View Full Product Details