Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52866326-52866854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASL11B
Entrez Gene ID
65997
Gene Name
RAS like family 11 member B
Gene Aliases
-
UniGene
Hs.8035
Gene Chromosome Location
Chr.4: 52862328-52866835 on Build GRCh38
Transcripts
NM_023940.2

SNP Detail

SNP ID Chromosome Location
rs1004718473 Chr.4: 52866413-52866413 on Build GRCh38
rs112773145 Chr.4: 52866451-52866451 on Build GRCh38
rs368342070 Chr.4: 52866563-52866564 on Build GRCh38
rs543224075 Chr.4: 52866542-52866542 on Build GRCh38
rs1017667384 Chr.4: 52866708-52866708 on Build GRCh38
rs1004685817 Chr.4: 52866601-52866601 on Build GRCh38
rs564986129 Chr.4: 52866666-52866666 on Build GRCh38
rs966869407 Chr.4: 52866488-52866488 on Build GRCh38
rs768998672 Chr.4: 52866591-52866591 on Build GRCh38
rs955941802 Chr.4: 52866751-52866751 on Build GRCh38
rs887518300 Chr.4: 52866676-52866676 on Build GRCh38
rs761244205 Chr.4: 52866749-52866749 on Build GRCh38
rs966351403 Chr.4: 52866422-52866422 on Build GRCh38
rs565269636 Chr.4: 52866599-52866599 on Build GRCh38
rs918053864 Chr.4: 52866466-52866466 on Build GRCh38
rs928614420 Chr.4: 52866778-52866778 on Build GRCh38
rs545214452 Chr.4: 52866595-52866595 on Build GRCh38
rs975055707 Chr.4: 52866803-52866803 on Build GRCh38
rs775761075 Chr.4: 52866528-52866528 on Build GRCh38
rs886281932 Chr.4: 52866381-52866381 on Build GRCh38
rs965767546 Chr.4: 52866713-52866713 on Build GRCh38
rs921656909 Chr.4: 52866563-52866563 on Build GRCh38
rs997604960 Chr.4: 52866747-52866747 on Build GRCh38
rs557611040 Chr.4: 52866398-52866398 on Build GRCh38
rs550719868 Chr.4: 52866639-52866639 on Build GRCh38
rs978313437 Chr.4: 52866501-52866501 on Build GRCh38
rs149879605 Chr.4: 52866814-52866814 on Build GRCh38
rs576305483 Chr.4: 52866491-52866491 on Build GRCh38
rs750059327 Chr.4: 52866782-52866782 on Build GRCh38
rs1007347808 Chr.4: 52866683-52866683 on Build GRCh38
rs900236878 Chr.4: 52866352-52866352 on Build GRCh38
rs531011338 Chr.4: 52866600-52866600 on Build GRCh38
rs1037528020 Chr.4: 52866607-52866607 on Build GRCh38
rs8287 Chr.4: 52866419-52866419 on Build GRCh38
rs138991809 Chr.4: 52866527-52866527 on Build GRCh38
rs869111122 Chr.4: 52866706-52866706 on Build GRCh38
rs1007600094 Chr.4: 52866368-52866368 on Build GRCh38
rs546841307 Chr.4: 52866758-52866758 on Build GRCh38
rs970545286 Chr.4: 52866770-52866770 on Build GRCh38
rs931574964 Chr.4: 52866822-52866822 on Build GRCh38
rs567615981 Chr.4: 52866644-52866644 on Build GRCh38
rs534413026 Chr.4: 52866358-52866358 on Build GRCh38
rs992291770 Chr.4: 52866457-52866457 on Build GRCh38
rs925536806 Chr.4: 52866504-52866504 on Build GRCh38
rs559598653 Chr.4: 52866556-52866556 on Build GRCh38
rs879134545 Chr.4: 52866585-52866585 on Build GRCh38
rs574008095 Chr.4: 52866521-52866521 on Build GRCh38
rs1037317618 Chr.4: 52866620-52866620 on Build GRCh38
rs761226315 Chr.4: 52866547-52866547 on Build GRCh38
rs540967230 Chr.4: 52866821-52866821 on Build GRCh38
rs1043975 Chr.4: 52866444-52866444 on Build GRCh38
rs955827859 Chr.4: 52866502-52866502 on Build GRCh38
rs9646 Chr.4: 52866776-52866776 on Build GRCh38
rs5858187 Chr.4: 52866705-52866705 on Build GRCh38
rs528249185 Chr.4: 52866572-52866572 on Build GRCh38
rs989947688 Chr.4: 52866757-52866757 on Build GRCh38
rs925498563 Chr.4: 52866772-52866772 on Build GRCh38
rs961465855 Chr.4: 52866732-52866732 on Build GRCh38
rs1044784465 Chr.4: 52866574-52866574 on Build GRCh38
rs974469475 Chr.4: 52866441-52866441 on Build GRCh38

View Full Product Details