Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 95164190-95164698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UNC5C
Entrez Gene ID
8633
Gene Name
unc-5 netrin receptor C
Gene Aliases
UNC5H3
UniGene
Hs.388565
Gene Chromosome Location
Chr.4: 95162504-95549210 on Build GRCh38
Transcripts
NM_003728.3 XM_005263321.3 XM_017008789.1 XM_017008790.1

SNP Detail

SNP ID Chromosome Location
rs1031905030 Chr.4: 95164644-95164644 on Build GRCh38
rs892219259 Chr.4: 95164515-95164515 on Build GRCh38
rs942172196 Chr.4: 95164284-95164284 on Build GRCh38
rs1057445796 Chr.4: 95164403-95164403 on Build GRCh38
rs1021235278 Chr.4: 95164375-95164375 on Build GRCh38
rs1030424788 Chr.4: 95164571-95164572 on Build GRCh38
rs189942043 Chr.4: 95164641-95164641 on Build GRCh38
rs9985855 Chr.4: 95164619-95164619 on Build GRCh38
rs944859900 Chr.4: 95164448-95164448 on Build GRCh38
rs560720810 Chr.4: 95164389-95164389 on Build GRCh38
rs989800058 Chr.4: 95164363-95164363 on Build GRCh38
rs972227537 Chr.4: 95164666-95164666 on Build GRCh38
rs919347801 Chr.4: 95164306-95164306 on Build GRCh38
rs907105156 Chr.4: 95164637-95164637 on Build GRCh38
rs1049104176 Chr.4: 95164523-95164523 on Build GRCh38
rs986371310 Chr.4: 95164232-95164232 on Build GRCh38
rs976650218 Chr.4: 95164455-95164455 on Build GRCh38
rs115955515 Chr.4: 95164391-95164391 on Build GRCh38
rs116545334 Chr.4: 95164215-95164215 on Build GRCh38
rs1003041844 Chr.4: 95164282-95164282 on Build GRCh38
rs1044967815 Chr.4: 95164469-95164469 on Build GRCh38
rs553735704 Chr.4: 95164481-95164481 on Build GRCh38
rs986322102 Chr.4: 95164404-95164404 on Build GRCh38
rs909399851 Chr.4: 95164262-95164263 on Build GRCh38
rs903738697 Chr.4: 95164477-95164477 on Build GRCh38
rs756927858 Chr.4: 95164554-95164554 on Build GRCh38
rs555243658 Chr.4: 95164546-95164546 on Build GRCh38
rs891573270 Chr.4: 95164650-95164650 on Build GRCh38
rs906994262 Chr.4: 95164276-95164276 on Build GRCh38
rs1044300137 Chr.4: 95164624-95164624 on Build GRCh38
rs910714119 Chr.4: 95164438-95164438 on Build GRCh38
rs28491664 Chr.4: 95164497-95164497 on Build GRCh38
rs972176650 Chr.4: 95164292-95164292 on Build GRCh38
rs1031452285 Chr.4: 95164345-95164345 on Build GRCh38
rs953777195 Chr.4: 95164225-95164225 on Build GRCh38
rs1022822802 Chr.4: 95164237-95164237 on Build GRCh38
rs955982278 Chr.4: 95164355-95164355 on Build GRCh38

View Full Product Details