Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99061070-99061580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METAP1
Entrez Gene ID
23173
Gene Name
methionyl aminopeptidase 1
Gene Aliases
MAP1A, MetAP1A
UniGene
Hs.480364
Gene Chromosome Location
Chr.4: 98995637-99062809 on Build GRCh38
Transcripts
NM_015143.2 XM_011531777.2 XM_011531778.1 XM_011531779.1 XM_017007914.1 XM_017007915.1 XM_017007916.1 XM_017007917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6711554 c.1135C>T p.R379W Chr.4: 99061291-99061291 on Build GRCh38
5401870 c.1016C>T p.T339I Chr.4: 99061172-99061172 on Build GRCh38
1193166 c.1036G>T p.A346S Chr.4: 99061192-99061192 on Build GRCh38
6883723 c.1039G>A p.V347M Chr.4: 99061195-99061195 on Build GRCh38
4701790 c.1137G>A p.R379R Chr.4: 99061293-99061293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145448780 Chr.4: 99061258-99061259 on Build GRCh38
rs149360928 Chr.4: 99061360-99061360 on Build GRCh38
rs979754764 Chr.4: 99061211-99061211 on Build GRCh38
rs748247308 Chr.4: 99061261-99061261 on Build GRCh38
rs958919131 Chr.4: 99061538-99061538 on Build GRCh38
rs1037265839 Chr.4: 99061239-99061239 on Build GRCh38
rs992297178 Chr.4: 99061462-99061462 on Build GRCh38
rs142456262 Chr.4: 99061206-99061206 on Build GRCh38
rs969802376 Chr.4: 99061469-99061469 on Build GRCh38
rs776450607 Chr.4: 99061282-99061282 on Build GRCh38
rs539287912 Chr.4: 99061528-99061528 on Build GRCh38
rs998073773 Chr.4: 99061291-99061291 on Build GRCh38
rs1023982692 Chr.4: 99061466-99061466 on Build GRCh38
rs750830276 Chr.4: 99061207-99061207 on Build GRCh38
rs917004561 Chr.4: 99061200-99061200 on Build GRCh38
rs560095874 Chr.4: 99061547-99061547 on Build GRCh38
rs531050033 Chr.4: 99061484-99061505 on Build GRCh38
rs897027898 Chr.4: 99061247-99061247 on Build GRCh38
rs79876289 Chr.4: 99061290-99061290 on Build GRCh38
rs754289702 Chr.4: 99061333-99061333 on Build GRCh38
rs75648569 Chr.4: 99061260-99061260 on Build GRCh38
rs780343100 Chr.4: 99061246-99061246 on Build GRCh38
rs534093387 Chr.4: 99061273-99061273 on Build GRCh38
rs868450356 Chr.4: 99061231-99061231 on Build GRCh38
rs76763924 Chr.4: 99061262-99061262 on Build GRCh38
rs771976847 Chr.4: 99061269-99061269 on Build GRCh38
rs941318821 Chr.4: 99061220-99061220 on Build GRCh38
rs1031673824 Chr.4: 99061362-99061362 on Build GRCh38
rs755294489 Chr.4: 99061259-99061259 on Build GRCh38
rs748445323 Chr.4: 99061353-99061353 on Build GRCh38
rs749653198 Chr.4: 99061253-99061253 on Build GRCh38
rs941274329 Chr.4: 99061544-99061544 on Build GRCh38
rs768989070 Chr.4: 99061108-99061108 on Build GRCh38
rs775066290 Chr.4: 99061292-99061292 on Build GRCh38
rs772065310 Chr.4: 99061150-99061150 on Build GRCh38
rs568241453 Chr.4: 99061106-99061106 on Build GRCh38
rs1022319708 Chr.4: 99061336-99061336 on Build GRCh38
rs867351518 Chr.4: 99061548-99061548 on Build GRCh38
rs750841176 Chr.4: 99061114-99061114 on Build GRCh38
rs535426460 Chr.4: 99061178-99061178 on Build GRCh38
rs577099767 Chr.4: 99061303-99061303 on Build GRCh38
rs371960839 Chr.4: 99061156-99061156 on Build GRCh38
rs762530332 Chr.4: 99061297-99061297 on Build GRCh38
rs766882314 Chr.4: 99061324-99061324 on Build GRCh38
rs746975345 Chr.4: 99061274-99061274 on Build GRCh38
rs753652866 Chr.4: 99061173-99061173 on Build GRCh38
rs972639632 Chr.4: 99061550-99061550 on Build GRCh38
rs946112618 Chr.4: 99061143-99061143 on Build GRCh38
rs752236905 Chr.4: 99061197-99061197 on Build GRCh38
rs759286171 Chr.4: 99061288-99061288 on Build GRCh38
rs1017090227 Chr.4: 99061447-99061447 on Build GRCh38
rs139041697 Chr.4: 99061311-99061311 on Build GRCh38
rs779277758 Chr.4: 99061348-99061348 on Build GRCh38
rs756631478 Chr.4: 99061321-99061321 on Build GRCh38
rs766153447 Chr.4: 99061170-99061170 on Build GRCh38
rs200609570 Chr.4: 99061213-99061213 on Build GRCh38
rs181684369 Chr.4: 99061426-99061426 on Build GRCh38
rs918509732 Chr.4: 99061552-99061552 on Build GRCh38
rs1057125703 Chr.4: 99061103-99061103 on Build GRCh38
rs755333115 Chr.4: 99061342-99061342 on Build GRCh38
rs202113897 Chr.4: 99061276-99061276 on Build GRCh38
rs909585475 Chr.4: 99061515-99061515 on Build GRCh38
rs980160867 Chr.4: 99061479-99061479 on Build GRCh38
rs368960333 Chr.4: 99061142-99061142 on Build GRCh38
rs200940950 Chr.4: 99061155-99061155 on Build GRCh38
rs541421673 Chr.4: 99061432-99061432 on Build GRCh38
rs375237670 Chr.4: 99061194-99061194 on Build GRCh38
rs774626346 Chr.4: 99061136-99061136 on Build GRCh38

View Full Product Details