Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109869561-109870058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRIT3
Entrez Gene ID
345193
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Gene Aliases
CSNB1F, FIGLER4
UniGene
Hs.308127
Gene Chromosome Location
Chr.4: 109848182-109872315 on Build GRCh38
Transcripts
NM_198506.4 XM_017008167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725655 c.1084G>T p.G362* Chr.4: 109869833-109869833 on Build GRCh38
1213709 c.832G>A p.A278T Chr.4: 109869716-109869716 on Build GRCh38
4328746 c.615C>G p.S205S Chr.4: 109869913-109869913 on Build GRCh38
5060057 c.992C>G p.S331C Chr.4: 109869876-109869876 on Build GRCh38
4584727 c.1041C>T p.G347G Chr.4: 109869790-109869790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778669769 Chr.4: 109869745-109869745 on Build GRCh38
rs397509379 Chr.4: 109869900-109869900 on Build GRCh38
rs141753583 Chr.4: 109869715-109869715 on Build GRCh38
rs781074134 Chr.4: 109869965-109869965 on Build GRCh38
rs752069625 Chr.4: 109869730-109869730 on Build GRCh38
rs778238467 Chr.4: 109869858-109869858 on Build GRCh38
rs1043790079 Chr.4: 109869903-109869903 on Build GRCh38
rs545573146 Chr.4: 109869697-109869697 on Build GRCh38
rs763421788 Chr.4: 109869731-109869731 on Build GRCh38
rs139536875 Chr.4: 109869632-109869632 on Build GRCh38
rs199943498 Chr.4: 109869778-109869778 on Build GRCh38
rs201205234 Chr.4: 109869616-109869616 on Build GRCh38
rs145673663 Chr.4: 109869817-109869817 on Build GRCh38
rs762022717 Chr.4: 109869790-109869790 on Build GRCh38
rs765373112 Chr.4: 109869603-109869603 on Build GRCh38
rs766767705 Chr.4: 109869797-109869797 on Build GRCh38
rs984033485 Chr.4: 109869838-109869838 on Build GRCh38
rs757902583 Chr.4: 109869925-109869925 on Build GRCh38
rs769927181 Chr.4: 109869886-109869886 on Build GRCh38
rs938008451 Chr.4: 109869928-109869928 on Build GRCh38
rs755626069 Chr.4: 109869971-109869971 on Build GRCh38
rs775531246 Chr.4: 109869685-109869685 on Build GRCh38
rs138218384 Chr.4: 109870022-109870022 on Build GRCh38
rs747431079 Chr.4: 109869615-109869615 on Build GRCh38
rs749381848 Chr.4: 109869985-109869985 on Build GRCh38
rs764205 Chr.4: 109869755-109869755 on Build GRCh38
rs150560307 Chr.4: 109869855-109869855 on Build GRCh38
rs1014032014 Chr.4: 109869722-109869722 on Build GRCh38
rs780648233 Chr.4: 109869611-109869611 on Build GRCh38
rs754839429 Chr.4: 109869961-109869961 on Build GRCh38
rs748116837 Chr.4: 109869966-109869966 on Build GRCh38
rs759885114 Chr.4: 109869811-109869811 on Build GRCh38
rs779839287 Chr.4: 109869867-109869867 on Build GRCh38
rs1011024544 Chr.4: 109869651-109869651 on Build GRCh38
rs781307155 Chr.4: 109869879-109869879 on Build GRCh38
rs753175023 Chr.4: 109869840-109869840 on Build GRCh38
rs777203063 Chr.4: 109869624-109869624 on Build GRCh38
rs184650144 Chr.4: 109869931-109869931 on Build GRCh38
rs542235762 Chr.4: 109869670-109869670 on Build GRCh38
rs895732426 Chr.4: 109869943-109869943 on Build GRCh38
rs775765889 Chr.4: 109870001-109870001 on Build GRCh38
rs750103732 Chr.4: 109869861-109869861 on Build GRCh38
rs746795751 Chr.4: 109869767-109869767 on Build GRCh38
rs878896031 Chr.4: 109869635-109869635 on Build GRCh38
rs538065711 Chr.4: 109869836-109869836 on Build GRCh38
rs768539302 Chr.4: 109869874-109869874 on Build GRCh38
rs373408851 Chr.4: 109869716-109869716 on Build GRCh38
rs758666361 Chr.4: 109869609-109869609 on Build GRCh38
rs199662798 Chr.4: 109869795-109869795 on Build GRCh38
rs774590264 Chr.4: 109869988-109869988 on Build GRCh38
rs996641458 Chr.4: 109869649-109869649 on Build GRCh38
rs373003106 Chr.4: 109869871-109869871 on Build GRCh38
rs140494004 Chr.4: 109869908-109869908 on Build GRCh38
rs112980116 Chr.4: 109869660-109869660 on Build GRCh38
rs770968977 Chr.4: 109869987-109869987 on Build GRCh38
rs777757196 Chr.4: 109869981-109869981 on Build GRCh38
rs761969518 Chr.4: 109869705-109869705 on Build GRCh38
rs761097698 Chr.4: 109870017-109870017 on Build GRCh38
rs777279684 Chr.4: 109869699-109869699 on Build GRCh38
rs768772325 Chr.4: 109869777-109869777 on Build GRCh38
rs751215450 Chr.4: 109869936-109869936 on Build GRCh38
rs1052627539 Chr.4: 109869622-109869622 on Build GRCh38
rs368638041 Chr.4: 109869904-109869904 on Build GRCh38
rs548812499 Chr.4: 109869926-109869926 on Build GRCh38
rs545747114 Chr.4: 109869741-109869741 on Build GRCh38
rs181208057 Chr.4: 109869862-109869862 on Build GRCh38
rs1052597074 Chr.4: 109869809-109869809 on Build GRCh38
rs766599778 Chr.4: 109869725-109869725 on Build GRCh38
rs145061437 Chr.4: 109869668-109869668 on Build GRCh38
rs773503246 Chr.4: 109869794-109869794 on Build GRCh38
rs532945434 Chr.4: 109869889-109869889 on Build GRCh38
rs140239777 Chr.4: 109869760-109869760 on Build GRCh38
rs777163396 Chr.4: 109870023-109870023 on Build GRCh38
rs794727660 Chr.4: 109869936-109869938 on Build GRCh38
rs760642322 Chr.4: 109869687-109869687 on Build GRCh38
rs371623354 Chr.4: 109869901-109869901 on Build GRCh38
rs765187464 Chr.4: 109869791-109869791 on Build GRCh38
rs151283521 Chr.4: 109869906-109869906 on Build GRCh38
rs201714421 Chr.4: 109870035-109870035 on Build GRCh38
rs377073593 Chr.4: 109869703-109869703 on Build GRCh38
rs772441857 Chr.4: 109869994-109869994 on Build GRCh38
rs149197944 Chr.4: 109869865-109869865 on Build GRCh38
rs765543287 Chr.4: 109869710-109869710 on Build GRCh38
rs77966848 Chr.4: 109869975-109869975 on Build GRCh38
rs376610215 Chr.4: 109869732-109869732 on Build GRCh38
rs959295666 Chr.4: 109870007-109870007 on Build GRCh38
rs991241747 Chr.4: 109870019-109870019 on Build GRCh38
rs376430373 Chr.4: 109869884-109869884 on Build GRCh38
rs747984845 Chr.4: 109869880-109869880 on Build GRCh38
rs892540724 Chr.4: 109869637-109869637 on Build GRCh38
rs979342750 Chr.4: 109869751-109869751 on Build GRCh38
rs751991187 Chr.4: 109869806-109869806 on Build GRCh38
rs369521916 Chr.4: 109869764-109869764 on Build GRCh38
rs750856339 Chr.4: 109869605-109869605 on Build GRCh38
rs774537043 Chr.4: 109869895-109869895 on Build GRCh38
rs761341365 Chr.4: 109869893-109869893 on Build GRCh38
rs369314060 Chr.4: 109869654-109869654 on Build GRCh38
rs762400169 Chr.4: 109870030-109870030 on Build GRCh38
rs748949021 Chr.4: 109869625-109869625 on Build GRCh38
rs377733787 Chr.4: 109869892-109869892 on Build GRCh38

View Full Product Details